Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62373673-62374133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB3
Entrez Gene ID
146779
Gene Name
EF-hand calcium binding domain 3
Gene Aliases
-
UniGene
Hs.152670
Gene Chromosome Location
Chr.17: 62370218-62416479 on Build GRCh38
Transcripts
NM_001144933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6523779 c.88C>A p.P30T Chr.17: 62373867-62373867 on Build GRCh38
5686010 c.88+3A>T p.? Chr.17: 62373870-62373870 on Build GRCh38
3820150 c.75G>T p.L25L Chr.17: 62373854-62373854 on Build GRCh38
1385054 c.59C>A p.S20* Chr.17: 62373838-62373838 on Build GRCh38
3820151 c.78G>A p.K26K Chr.17: 62373857-62373857 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6504103 Chr.17: 62373824-62373824 on Build GRCh38
rs184235465 Chr.17: 62373783-62373783 on Build GRCh38
rs541890707 Chr.17: 62373976-62373976 on Build GRCh38
rs532864070 Chr.17: 62373817-62373817 on Build GRCh38
rs1010196846 Chr.17: 62374024-62374024 on Build GRCh38
rs929178708 Chr.17: 62373892-62373892 on Build GRCh38
rs1005999595 Chr.17: 62374043-62374043 on Build GRCh38
rs956277270 Chr.17: 62373737-62373737 on Build GRCh38
rs879453481 Chr.17: 62374000-62374000 on Build GRCh38
rs980210588 Chr.17: 62373853-62373853 on Build GRCh38
rs1057091433 Chr.17: 62373913-62373913 on Build GRCh38
rs909015594 Chr.17: 62373941-62373941 on Build GRCh38
rs981233535 Chr.17: 62373739-62373739 on Build GRCh38
rs919293253 Chr.17: 62373885-62373885 on Build GRCh38
rs562922307 Chr.17: 62373867-62373867 on Build GRCh38
rs578125210 Chr.17: 62373844-62373844 on Build GRCh38
rs1031040755 Chr.17: 62373730-62373730 on Build GRCh38
rs767078931 Chr.17: 62373834-62373834 on Build GRCh38
rs762432427 Chr.17: 62373768-62373768 on Build GRCh38
rs1052764181 Chr.17: 62373987-62373987 on Build GRCh38
rs752103555 Chr.17: 62373799-62373799 on Build GRCh38
rs896398935 Chr.17: 62373930-62373930 on Build GRCh38
rs988988178 Chr.17: 62373791-62373791 on Build GRCh38
rs891477635 Chr.17: 62374004-62374004 on Build GRCh38
rs747998461 Chr.17: 62373957-62373957 on Build GRCh38
rs1048974951 Chr.17: 62373901-62373901 on Build GRCh38
rs780811315 Chr.17: 62373944-62373944 on Build GRCh38
rs188007766 Chr.17: 62374069-62374069 on Build GRCh38
rs1017807993 Chr.17: 62374045-62374045 on Build GRCh38
rs756603881 Chr.17: 62373886-62373886 on Build GRCh38
rs900247796 Chr.17: 62374068-62374068 on Build GRCh38
rs992115172 Chr.17: 62373838-62373838 on Build GRCh38
rs997307242 Chr.17: 62373979-62373979 on Build GRCh38
rs1039308673 Chr.17: 62373969-62373969 on Build GRCh38
rs754774456 Chr.17: 62373721-62373721 on Build GRCh38
rs964763332 Chr.17: 62373715-62373715 on Build GRCh38
rs773752280 Chr.17: 62373819-62373819 on Build GRCh38

View Full Product Details