Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62377825-62378336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB3
Entrez Gene ID
146779
Gene Name
EF-hand calcium binding domain 3
Gene Aliases
-
UniGene
Hs.152670
Gene Chromosome Location
Chr.17: 62370218-62416479 on Build GRCh38
Transcripts
NM_001144933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530358 c.95G>A p.G32E Chr.17: 62377974-62377974 on Build GRCh38
5530359 c.130G>A p.D44N Chr.17: 62378009-62378009 on Build GRCh38
1385055 c.89C>T p.P30L Chr.17: 62377968-62377968 on Build GRCh38
6320596 c.89-9G>T p.? Chr.17: 62377959-62377959 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557993299 Chr.17: 62378108-62378108 on Build GRCh38
rs1012826682 Chr.17: 62378059-62378059 on Build GRCh38
rs1044642292 Chr.17: 62378193-62378193 on Build GRCh38
rs764675263 Chr.17: 62378001-62378001 on Build GRCh38
rs894010162 Chr.17: 62378273-62378274 on Build GRCh38
rs55937716 Chr.17: 62378145-62378145 on Build GRCh38
rs995899716 Chr.17: 62377987-62377987 on Build GRCh38
rs1002931730 Chr.17: 62377887-62377887 on Build GRCh38
rs754985267 Chr.17: 62377905-62377905 on Build GRCh38
rs894006446 Chr.17: 62378056-62378056 on Build GRCh38
rs897261531 Chr.17: 62377917-62377917 on Build GRCh38
rs1032991308 Chr.17: 62377961-62377961 on Build GRCh38
rs1037229334 Chr.17: 62378248-62378248 on Build GRCh38
rs943743579 Chr.17: 62378185-62378185 on Build GRCh38
rs1023723242 Chr.17: 62378080-62378080 on Build GRCh38
rs572240307 Chr.17: 62378213-62378213 on Build GRCh38
rs878900711 Chr.17: 62378296-62378296 on Build GRCh38
rs186566856 Chr.17: 62378002-62378002 on Build GRCh38
rs12943354 Chr.17: 62377873-62377873 on Build GRCh38
rs965599368 Chr.17: 62378123-62378123 on Build GRCh38
rs12941037 Chr.17: 62377872-62377872 on Build GRCh38
rs995627624 Chr.17: 62378280-62378280 on Build GRCh38
rs569781918 Chr.17: 62377882-62377882 on Build GRCh38
rs756272614 Chr.17: 62378072-62378072 on Build GRCh38
rs190475900 Chr.17: 62378042-62378042 on Build GRCh38
rs549492045 Chr.17: 62377878-62377878 on Build GRCh38
rs775778487 Chr.17: 62377989-62377989 on Build GRCh38
rs757676296 Chr.17: 62378165-62378165 on Build GRCh38
rs115257211 Chr.17: 62378162-62378162 on Build GRCh38
rs555602090 Chr.17: 62378205-62378205 on Build GRCh38
rs1037540740 Chr.17: 62377900-62377900 on Build GRCh38
rs982090844 Chr.17: 62378097-62378097 on Build GRCh38
rs910983532 Chr.17: 62378177-62378177 on Build GRCh38
rs934974194 Chr.17: 62378223-62378223 on Build GRCh38
rs907373379 Chr.17: 62377856-62377856 on Build GRCh38
rs541213056 Chr.17: 62378217-62378217 on Build GRCh38
rs190977243 Chr.17: 62378154-62378154 on Build GRCh38
rs777538309 Chr.17: 62377983-62377983 on Build GRCh38
rs977976811 Chr.17: 62378127-62378127 on Build GRCh38
rs1040796277 Chr.17: 62378198-62378198 on Build GRCh38
rs1048384573 Chr.17: 62378293-62378293 on Build GRCh38
rs373303781 Chr.17: 62378009-62378009 on Build GRCh38
rs543101029 Chr.17: 62378182-62378182 on Build GRCh38
rs746426454 Chr.17: 62378187-62378187 on Build GRCh38
rs182998101 Chr.17: 62378060-62378060 on Build GRCh38
rs902784430 Chr.17: 62377880-62377880 on Build GRCh38
rs777977521 Chr.17: 62378115-62378115 on Build GRCh38
rs184113033 Chr.17: 62378309-62378309 on Build GRCh38
rs928795922 Chr.17: 62378196-62378196 on Build GRCh38
rs571832239 Chr.17: 62378133-62378133 on Build GRCh38
rs1027020814 Chr.17: 62377990-62377990 on Build GRCh38
rs1006813646 Chr.17: 62378064-62378064 on Build GRCh38
rs897222653 Chr.17: 62378253-62378253 on Build GRCh38
rs187669498 Chr.17: 62378084-62378084 on Build GRCh38

View Full Product Details