Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62393435-62393946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB3
Entrez Gene ID
146779
Gene Name
EF-hand calcium binding domain 3
Gene Aliases
-
UniGene
Hs.152670
Gene Chromosome Location
Chr.17: 62370218-62416479 on Build GRCh38
Transcripts
NM_001144933.1 NM_173503.3 XM_011524380.1 XM_011524381.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710587 c.302G>A p.G101E Chr.17: 62393579-62393579 on Build GRCh38
5701962 c.489_490TC>GG p.L164V Chr.17: 62393610-62393611 on Build GRCh38
3795906 c.338C>G p.T113R Chr.17: 62393615-62393615 on Build GRCh38
6523780 c.510C>A p.F170L Chr.17: 62393631-62393631 on Build GRCh38
6523781 c.354C>A p.F118L Chr.17: 62393631-62393631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547105101 Chr.17: 62393773-62393773 on Build GRCh38
rs1017100423 Chr.17: 62393717-62393717 on Build GRCh38
rs1020793488 Chr.17: 62393582-62393582 on Build GRCh38
rs767552735 Chr.17: 62393684-62393684 on Build GRCh38
rs370363023 Chr.17: 62393569-62393569 on Build GRCh38
rs991843827 Chr.17: 62393667-62393667 on Build GRCh38
rs535079608 Chr.17: 62393899-62393902 on Build GRCh38
rs745764993 Chr.17: 62393803-62393803 on Build GRCh38
rs771336500 Chr.17: 62393589-62393589 on Build GRCh38
rs921326779 Chr.17: 62393495-62393495 on Build GRCh38
rs545597293 Chr.17: 62393549-62393549 on Build GRCh38
rs561306595 Chr.17: 62393726-62393726 on Build GRCh38
rs903714838 Chr.17: 62393861-62393861 on Build GRCh38
rs979434439 Chr.17: 62393880-62393880 on Build GRCh38
rs1013733717 Chr.17: 62393675-62393675 on Build GRCh38
rs764216450 Chr.17: 62393626-62393626 on Build GRCh38
rs762816457 Chr.17: 62393635-62393635 on Build GRCh38
rs747711546 Chr.17: 62393544-62393544 on Build GRCh38
rs764069772 Chr.17: 62393616-62393616 on Build GRCh38
rs375627224 Chr.17: 62393624-62393624 on Build GRCh38
rs945357851 Chr.17: 62393876-62393876 on Build GRCh38
rs80292970 Chr.17: 62393892-62393892 on Build GRCh38
rs772069268 Chr.17: 62393849-62393849 on Build GRCh38
rs1050987332 Chr.17: 62393520-62393520 on Build GRCh38
rs925162010 Chr.17: 62393884-62393884 on Build GRCh38
rs962941772 Chr.17: 62393763-62393763 on Build GRCh38
rs903812690 Chr.17: 62393654-62393654 on Build GRCh38
rs886096843 Chr.17: 62393696-62393696 on Build GRCh38
rs575999728 Chr.17: 62393694-62393694 on Build GRCh38
rs774721212 Chr.17: 62393590-62393590 on Build GRCh38
rs891062877 Chr.17: 62393577-62393577 on Build GRCh38
rs777470263 Chr.17: 62393483-62393483 on Build GRCh38
rs778789606 Chr.17: 62393714-62393714 on Build GRCh38
rs1051728217 Chr.17: 62393800-62393800 on Build GRCh38
rs776950824 Chr.17: 62393634-62393634 on Build GRCh38
rs749006281 Chr.17: 62393642-62393642 on Build GRCh38
rs189311418 Chr.17: 62393578-62393578 on Build GRCh38
rs934083840 Chr.17: 62393529-62393529 on Build GRCh38
rs757091340 Chr.17: 62393685-62393685 on Build GRCh38
rs777552746 Chr.17: 62393574-62393574 on Build GRCh38
rs942182652 Chr.17: 62393751-62393751 on Build GRCh38
rs987275937 Chr.17: 62393807-62393807 on Build GRCh38
rs754985498 Chr.17: 62393522-62393522 on Build GRCh38
rs896293721 Chr.17: 62393905-62393905 on Build GRCh38
rs184476573 Chr.17: 62393528-62393528 on Build GRCh38
rs1000651707 Chr.17: 62393638-62393638 on Build GRCh38
rs913886367 Chr.17: 62393573-62393573 on Build GRCh38
rs544626465 Chr.17: 62393710-62393710 on Build GRCh38
rs144845537 Chr.17: 62393618-62393618 on Build GRCh38
rs528315981 Chr.17: 62393889-62393889 on Build GRCh38
rs759475025 Chr.17: 62393665-62393665 on Build GRCh38
rs553307869 Chr.17: 62393492-62393492 on Build GRCh38
rs896212747 Chr.17: 62393674-62393674 on Build GRCh38
rs766225409 Chr.17: 62393637-62393637 on Build GRCh38
rs900542653 Chr.17: 62393460-62393460 on Build GRCh38
rs746334973 Chr.17: 62393595-62393595 on Build GRCh38
rs1043657570 Chr.17: 62393601-62393601 on Build GRCh38
rs772637360 Chr.17: 62393608-62393608 on Build GRCh38
rs867518258 Chr.17: 62393854-62393854 on Build GRCh38
rs34741297 Chr.17: 62393610-62393611 on Build GRCh38
rs202231587 Chr.17: 62393655-62393655 on Build GRCh38
rs962846978 Chr.17: 62393473-62393473 on Build GRCh38
rs975579751 Chr.17: 62393768-62393768 on Build GRCh38
rs755693731 Chr.17: 62393692-62393692 on Build GRCh38
rs551149768 Chr.17: 62393890-62393890 on Build GRCh38
rs181966404 Chr.17: 62393772-62393772 on Build GRCh38
rs933382949 Chr.17: 62393786-62393786 on Build GRCh38
rs551067066 Chr.17: 62393802-62393802 on Build GRCh38

View Full Product Details