Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63432522-63433053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561
Entrez Gene ID
1534
Gene Name
cytochrome b561
Gene Aliases
CYB561A1, FRRS2
UniGene
Hs.355264
Gene Chromosome Location
Chr.17: 63432304-63446361 on Build GRCh38
Transcripts
NM_001017916.1 NM_001017917.1 NM_001915.3 XM_005257091.1 XM_017024260.1 XM_017024261.1 XM_017024262.1

SNP Detail

SNP ID Chromosome Location
rs766952258 Chr.17: 63432939-63432949 on Build GRCh38
rs934075254 Chr.17: 63432927-63432927 on Build GRCh38
rs1057299394 Chr.17: 63432660-63432660 on Build GRCh38
rs757833714 Chr.17: 63432849-63432849 on Build GRCh38
rs578085295 Chr.17: 63432978-63432978 on Build GRCh38
rs920804374 Chr.17: 63432964-63432964 on Build GRCh38
rs888511151 Chr.17: 63432561-63432563 on Build GRCh38
rs529297199 Chr.17: 63433029-63433029 on Build GRCh38
rs569672261 Chr.17: 63432825-63432825 on Build GRCh38
rs970798196 Chr.17: 63432860-63432860 on Build GRCh38
rs763575825 Chr.17: 63432908-63432908 on Build GRCh38
rs536526108 Chr.17: 63432765-63432765 on Build GRCh38
rs57728995 Chr.17: 63432859-63432859 on Build GRCh38
rs1054089737 Chr.17: 63432845-63432845 on Build GRCh38
rs568013127 Chr.17: 63432698-63432698 on Build GRCh38
rs770018502 Chr.17: 63432855-63432855 on Build GRCh38
rs774000681 Chr.17: 63432973-63432973 on Build GRCh38
rs139891602 Chr.17: 63432972-63432972 on Build GRCh38
rs902219492 Chr.17: 63432821-63432821 on Build GRCh38
rs534727912 Chr.17: 63432730-63432730 on Build GRCh38
rs1048633223 Chr.17: 63432897-63432897 on Build GRCh38
rs548628938 Chr.17: 63432882-63432882 on Build GRCh38
rs142136314 Chr.17: 63433025-63433025 on Build GRCh38
rs927726687 Chr.17: 63432832-63432832 on Build GRCh38
rs965594527 Chr.17: 63432579-63432579 on Build GRCh38
rs927476978 Chr.17: 63432651-63432651 on Build GRCh38
rs1014920293 Chr.17: 63432903-63432903 on Build GRCh38
rs774184402 Chr.17: 63432890-63432892 on Build GRCh38
rs956341210 Chr.17: 63432595-63432595 on Build GRCh38
rs548085061 Chr.17: 63432680-63432680 on Build GRCh38
rs1012101012 Chr.17: 63432646-63432646 on Build GRCh38
rs376965007 Chr.17: 63432623-63432623 on Build GRCh38
rs768877005 Chr.17: 63432712-63432712 on Build GRCh38
rs566078528 Chr.17: 63432742-63432742 on Build GRCh38
rs549979100 Chr.17: 63432559-63432559 on Build GRCh38
rs943778332 Chr.17: 63433015-63433015 on Build GRCh38
rs780140858 Chr.17: 63432628-63432628 on Build GRCh38
rs748609201 Chr.17: 63432824-63432824 on Build GRCh38
rs918870948 Chr.17: 63432672-63432673 on Build GRCh38
rs970808190 Chr.17: 63432696-63432696 on Build GRCh38
rs184415180 Chr.17: 63432799-63432799 on Build GRCh38
rs1030962250 Chr.17: 63432593-63432593 on Build GRCh38
rs142389857 Chr.17: 63432561-63432561 on Build GRCh38
rs1048360344 Chr.17: 63432676-63432676 on Build GRCh38
rs556253232 Chr.17: 63432800-63432800 on Build GRCh38
rs145963330 Chr.17: 63432958-63432958 on Build GRCh38
rs930242463 Chr.17: 63432672-63432672 on Build GRCh38
rs976444494 Chr.17: 63433023-63433023 on Build GRCh38
rs544438693 Chr.17: 63432593-63432594 on Build GRCh38
rs943829474 Chr.17: 63432788-63432788 on Build GRCh38
rs958083415 Chr.17: 63432645-63432645 on Build GRCh38
rs77530319 Chr.17: 63432776-63432776 on Build GRCh38
rs938888285 Chr.17: 63432655-63432655 on Build GRCh38
rs980544742 Chr.17: 63432832-63432833 on Build GRCh38
rs960422697 Chr.17: 63433030-63433030 on Build GRCh38
rs942732403 Chr.17: 63432899-63432899 on Build GRCh38
rs986702594 Chr.17: 63432984-63432984 on Build GRCh38
rs576914672 Chr.17: 63432948-63432949 on Build GRCh38
rs909874907 Chr.17: 63432898-63432898 on Build GRCh38
rs973679370 Chr.17: 63432716-63432716 on Build GRCh38
rs1050625171 Chr.17: 63432777-63432777 on Build GRCh38
rs914923865 Chr.17: 63432624-63432624 on Build GRCh38
rs1040670637 Chr.17: 63432808-63432808 on Build GRCh38
rs930191469 Chr.17: 63432889-63432889 on Build GRCh38
rs543909654 Chr.17: 63433014-63433014 on Build GRCh38
rs533779034 Chr.17: 63432699-63432699 on Build GRCh38
rs910956250 Chr.17: 63432780-63432780 on Build GRCh38
rs747027383 Chr.17: 63432658-63432658 on Build GRCh38
rs1048452711 Chr.17: 63432557-63432557 on Build GRCh38
rs1018066697 Chr.17: 63432577-63432578 on Build GRCh38
rs956580135 Chr.17: 63432723-63432723 on Build GRCh38
rs888445929 Chr.17: 63432679-63432679 on Build GRCh38
rs3087776 Chr.17: 63432917-63432917 on Build GRCh38
rs912087803 Chr.17: 63432994-63432994 on Build GRCh38

View Full Product Details