Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63440595-63441099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561
Entrez Gene ID
1534
Gene Name
cytochrome b561
Gene Aliases
CYB561A1, FRRS2
UniGene
Hs.355264
Gene Chromosome Location
Chr.17: 63432304-63446361 on Build GRCh38
Transcripts
NM_001017916.1 NM_001017917.1 NM_001915.3 XM_005257091.1

SNP Detail

SNP ID Chromosome Location
rs1054161093 Chr.17: 63440860-63440860 on Build GRCh38
rs1014407713 Chr.17: 63440966-63440966 on Build GRCh38
rs529002960 Chr.17: 63440733-63440733 on Build GRCh38
rs931833781 Chr.17: 63440806-63440806 on Build GRCh38
rs944362114 Chr.17: 63440645-63440645 on Build GRCh38
rs778419890 Chr.17: 63440683-63440683 on Build GRCh38
rs983068075 Chr.17: 63440731-63440731 on Build GRCh38
rs944624285 Chr.17: 63440842-63440842 on Build GRCh38
rs574560276 Chr.17: 63441063-63441063 on Build GRCh38
rs1052055668 Chr.17: 63440809-63440809 on Build GRCh38
rs941719489 Chr.17: 63440751-63440751 on Build GRCh38
rs774799312 Chr.17: 63440885-63440885 on Build GRCh38
rs915619730 Chr.17: 63441062-63441062 on Build GRCh38
rs114549110 Chr.17: 63440617-63440617 on Build GRCh38
rs902902992 Chr.17: 63440648-63440648 on Build GRCh38
rs553384343 Chr.17: 63441039-63441039 on Build GRCh38
rs987417300 Chr.17: 63441036-63441036 on Build GRCh38
rs187323323 Chr.17: 63440986-63440986 on Build GRCh38
rs945864649 Chr.17: 63441047-63441047 on Build GRCh38
rs1051823827 Chr.17: 63440628-63440628 on Build GRCh38
rs558080583 Chr.17: 63440678-63440678 on Build GRCh38
rs577953161 Chr.17: 63440685-63440685 on Build GRCh38
rs921853691 Chr.17: 63440764-63440764 on Build GRCh38
rs1032468606 Chr.17: 63440681-63440681 on Build GRCh38
rs933164910 Chr.17: 63440807-63440807 on Build GRCh38
rs377405957 Chr.17: 63440686-63440686 on Build GRCh38
rs528044529 Chr.17: 63440883-63440883 on Build GRCh38
rs968716668 Chr.17: 63440680-63440680 on Build GRCh38
rs567811813 Chr.17: 63441018-63441018 on Build GRCh38
rs543627318 Chr.17: 63440713-63440713 on Build GRCh38
rs982093408 Chr.17: 63440767-63440767 on Build GRCh38
rs1027714082 Chr.17: 63440952-63440952 on Build GRCh38
rs962064395 Chr.17: 63440911-63440911 on Build GRCh38
rs2286571 Chr.17: 63440859-63440859 on Build GRCh38
rs879909753 Chr.17: 63440755-63440755 on Build GRCh38
rs11652994 Chr.17: 63440974-63440974 on Build GRCh38
rs2286572 Chr.17: 63440881-63440881 on Build GRCh38
rs1043128015 Chr.17: 63441059-63441059 on Build GRCh38
rs1025799777 Chr.17: 63440972-63440972 on Build GRCh38
rs780185297 Chr.17: 63440862-63440862 on Build GRCh38
rs768434833 Chr.17: 63440884-63440884 on Build GRCh38
rs960200146 Chr.17: 63440708-63440708 on Build GRCh38
rs1043179451 Chr.17: 63440846-63440846 on Build GRCh38
rs144890923 Chr.17: 63440994-63440994 on Build GRCh38
rs550781843 Chr.17: 63441013-63441013 on Build GRCh38
rs1014939979 Chr.17: 63440772-63440772 on Build GRCh38
rs904640989 Chr.17: 63441066-63441066 on Build GRCh38
rs902891871 Chr.17: 63440855-63440855 on Build GRCh38
rs950497078 Chr.17: 63440724-63440724 on Build GRCh38
rs575346098 Chr.17: 63440647-63440647 on Build GRCh38
rs1038912802 Chr.17: 63440752-63440752 on Build GRCh38
rs997763171 Chr.17: 63440620-63440620 on Build GRCh38
rs561756770 Chr.17: 63441025-63441025 on Build GRCh38
rs2286569 Chr.17: 63440644-63440644 on Build GRCh38
rs891872735 Chr.17: 63440827-63440827 on Build GRCh38
rs911723134 Chr.17: 63440993-63440993 on Build GRCh38
rs962983797 Chr.17: 63441012-63441012 on Build GRCh38
rs913044179 Chr.17: 63441046-63441046 on Build GRCh38
rs962112282 Chr.17: 63440773-63440773 on Build GRCh38
rs986385148 Chr.17: 63440823-63440823 on Build GRCh38
rs960609295 Chr.17: 63440928-63440928 on Build GRCh38
rs1039377000 Chr.17: 63440616-63440616 on Build GRCh38
rs1010296937 Chr.17: 63440835-63440835 on Build GRCh38
rs1021664271 Chr.17: 63440666-63440666 on Build GRCh38
rs1034597596 Chr.17: 63440890-63440890 on Build GRCh38
rs920409952 Chr.17: 63440802-63440802 on Build GRCh38
rs2286570 Chr.17: 63440714-63440714 on Build GRCh38
rs1041350465 Chr.17: 63440848-63440848 on Build GRCh38
rs973524493 Chr.17: 63440788-63440788 on Build GRCh38

View Full Product Details