Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46548613-46549118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL17A
Entrez Gene ID
51326
Gene Name
ADP ribosylation factor like GTPase 17A
Gene Aliases
ARF1P2, ARL17P1
UniGene
Hs.559259
Gene Chromosome Location
Chr.17: 46500213-46579781 on Build GRCh38
Transcripts
NM_001288811.1 NM_001288812.1 NM_001288813.1 NM_016632.2 XM_005257439.4 XM_011524875.2 XM_011524876.2 XM_011524877.2 XM_011524878.2 XM_017024729.1 NM_001006607.2 XM_011524841.2 XM_011524842.2 XM_011524843.2 XM_011524844.2 XM_011524846.2 XM_011524848.2

Gene Symbol
LRRC37A2
Entrez Gene ID
474170
Gene Name
leucine rich repeat containing 37 member A2
Gene Aliases
LRRC37
UniGene
Hs.646568
Gene Chromosome Location
Chr.17: 46512657-46556747 on Build GRCh38
Transcripts
NM_001288811.1 NM_001288812.1 NM_001288813.1 NM_016632.2 XM_005257439.4 XM_011524875.2 XM_011524876.2 XM_011524877.2 XM_011524878.2 XM_017024729.1 NM_001006607.2 XM_011524841.2 XM_011524842.2 XM_011524843.2 XM_011524844.2 XM_011524846.2 XM_011524848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1302934 c.3711C>T p.F1237F Chr.17: 46548850-46548850 on Build GRCh38
383906 c.3707C>T p.S1236F Chr.17: 46548846-46548846 on Build GRCh38
5164164 c.3858G>T p.K1286N Chr.17: 46548997-46548997 on Build GRCh38
1383857 c.3883T>C p.S1295P Chr.17: 46549022-46549022 on Build GRCh38
3691995 c.3583G>A p.A1195T Chr.17: 46548722-46548722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765592467 Chr.17: 46549031-46549031 on Build GRCh38
rs776229389 Chr.17: 46548959-46548959 on Build GRCh38
rs754491281 Chr.17: 46548874-46548874 on Build GRCh38
rs781341723 Chr.17: 46548832-46548832 on Build GRCh38
rs550094369 Chr.17: 46549079-46549079 on Build GRCh38
rs779253714 Chr.17: 46548947-46548947 on Build GRCh38
rs62073349 Chr.17: 46548981-46548981 on Build GRCh38
rs767008633 Chr.17: 46548979-46548979 on Build GRCh38
rs750067345 Chr.17: 46548730-46548732 on Build GRCh38
rs751144219 Chr.17: 46548974-46548974 on Build GRCh38
rs375225444 Chr.17: 46549044-46549044 on Build GRCh38
rs759790387 Chr.17: 46548793-46548793 on Build GRCh38
rs766019833 Chr.17: 46548865-46548865 on Build GRCh38
rs781253592 Chr.17: 46549004-46549004 on Build GRCh38
rs761645420 Chr.17: 46548813-46548813 on Build GRCh38
rs745768355 Chr.17: 46549055-46549055 on Build GRCh38
rs758097401 Chr.17: 46548753-46548753 on Build GRCh38
rs368129131 Chr.17: 46548970-46548970 on Build GRCh38
rs779695534 Chr.17: 46548823-46548823 on Build GRCh38
rs370112456 Chr.17: 46548827-46548827 on Build GRCh38
rs758978447 Chr.17: 46548638-46548638 on Build GRCh38
rs750778996 Chr.17: 46548655-46548655 on Build GRCh38
rs763183179 Chr.17: 46548811-46548811 on Build GRCh38
rs770123969 Chr.17: 46548835-46548835 on Build GRCh38
rs772510763 Chr.17: 46548785-46548785 on Build GRCh38
rs746245814 Chr.17: 46548833-46548833 on Build GRCh38
rs768106560 Chr.17: 46548929-46548929 on Build GRCh38
rs766989860 Chr.17: 46549021-46549021 on Build GRCh38
rs748274115 Chr.17: 46548901-46548901 on Build GRCh38
rs748802053 Chr.17: 46548782-46548782 on Build GRCh38
rs755099541 Chr.17: 46548945-46548945 on Build GRCh38
rs751355590 Chr.17: 46548758-46548758 on Build GRCh38
rs768955481 Chr.17: 46548899-46548899 on Build GRCh38
rs747102808 Chr.17: 46548998-46548998 on Build GRCh38
rs774819306 Chr.17: 46548737-46548737 on Build GRCh38
rs753897441 Chr.17: 46548676-46548676 on Build GRCh38
rs748172936 Chr.17: 46548948-46548948 on Build GRCh38
rs764088028 Chr.17: 46548812-46548812 on Build GRCh38
rs200087533 Chr.17: 46548634-46548634 on Build GRCh38
rs778152395 Chr.17: 46548991-46548991 on Build GRCh38
rs761617601 Chr.17: 46548845-46548845 on Build GRCh38
rs777793262 Chr.17: 46548997-46548997 on Build GRCh38
rs749110638 Chr.17: 46549071-46549071 on Build GRCh38
rs759430431 Chr.17: 46548961-46548961 on Build GRCh38
rs763373105 Chr.17: 46549090-46549090 on Build GRCh38
rs758551655 Chr.17: 46549034-46549034 on Build GRCh38
rs763947972 Chr.17: 46548757-46548757 on Build GRCh38
rs756624471 Chr.17: 46548975-46548975 on Build GRCh38
rs779511340 Chr.17: 46548873-46548874 on Build GRCh38
rs527794638 Chr.17: 46549018-46549018 on Build GRCh38
rs751015729 Chr.17: 46548933-46548933 on Build GRCh38
rs757644416 Chr.17: 46548881-46548881 on Build GRCh38
rs371532921 Chr.17: 46548749-46548749 on Build GRCh38
rs273529 Chr.17: 46548847-46548847 on Build GRCh38
rs776434570 Chr.17: 46548907-46548907 on Build GRCh38
rs768411441 Chr.17: 46549074-46549074 on Build GRCh38
rs778316169 Chr.17: 46548829-46548829 on Build GRCh38
rs376097433 Chr.17: 46548826-46548826 on Build GRCh38
rs779831291 Chr.17: 46549070-46549070 on Build GRCh38
rs375266817 Chr.17: 46548966-46548966 on Build GRCh38
rs765558536 Chr.17: 46548804-46548804 on Build GRCh38
rs201177598 Chr.17: 46548790-46548790 on Build GRCh38
rs577686617 Chr.17: 46548820-46548820 on Build GRCh38
rs760522683 Chr.17: 46548852-46548852 on Build GRCh38
rs773941851 Chr.17: 46549019-46549019 on Build GRCh38
rs375018792 Chr.17: 46548742-46548742 on Build GRCh38
rs372848109 Chr.17: 46549056-46549056 on Build GRCh38
rs202061223 Chr.17: 46548841-46548841 on Build GRCh38
rs273531 Chr.17: 46549094-46549094 on Build GRCh38
rs547525816 Chr.17: 46549040-46549040 on Build GRCh38
rs371932222 Chr.17: 46549087-46549087 on Build GRCh38
rs1863116 Chr.17: 46548670-46548670 on Build GRCh38
rs374053039 Chr.17: 46548771-46548771 on Build GRCh38
rs753044313 Chr.17: 46549088-46549088 on Build GRCh38
rs758340872 Chr.17: 46548755-46548755 on Build GRCh38
rs376855895 Chr.17: 46548842-46548842 on Build GRCh38
rs767920319 Chr.17: 46548654-46548654 on Build GRCh38
rs751825600 Chr.17: 46549093-46549093 on Build GRCh38
rs774509154 Chr.17: 46548844-46548844 on Build GRCh38
rs749420662 Chr.17: 46548898-46548898 on Build GRCh38
rs746784908 Chr.17: 46548706-46548706 on Build GRCh38
rs759800507 Chr.17: 46549028-46549028 on Build GRCh38
rs764265934 Chr.17: 46549092-46549092 on Build GRCh38
rs568551393 Chr.17: 46548746-46548746 on Build GRCh38
rs371285931 Chr.17: 46548971-46548971 on Build GRCh38
rs770839916 Chr.17: 46548999-46548999 on Build GRCh38
rs755438058 Chr.17: 46548983-46548983 on Build GRCh38
rs201014408 Chr.17: 46548653-46548653 on Build GRCh38
rs745616279 Chr.17: 46549008-46549008 on Build GRCh38
rs767169172 Chr.17: 46548814-46548814 on Build GRCh38
rs542986463 Chr.17: 46548908-46548908 on Build GRCh38
rs201965529 Chr.17: 46548810-46548810 on Build GRCh38
rs567718195 Chr.17: 46548736-46548736 on Build GRCh38
rs777670020 Chr.17: 46548703-46548703 on Build GRCh38
rs200419055 Chr.17: 46548902-46548902 on Build GRCh38
rs751140599 Chr.17: 46548915-46548915 on Build GRCh38
rs778975963 Chr.17: 46548991-46548991 on Build GRCh38
rs939339807 Chr.17: 46548709-46548709 on Build GRCh38
rs780966352 Chr.17: 46549053-46549053 on Build GRCh38
rs757363008 Chr.17: 46549052-46549052 on Build GRCh38
rs776140453 Chr.17: 46548637-46548637 on Build GRCh38
rs775244725 Chr.17: 46548967-46548967 on Build GRCh38
rs770521379 Chr.17: 46548636-46548636 on Build GRCh38
rs530221048 Chr.17: 46549078-46549078 on Build GRCh38
rs369469522 Chr.17: 46548754-46548754 on Build GRCh38
rs764952430 Chr.17: 46548909-46548909 on Build GRCh38
rs1863118 Chr.17: 46548759-46548759 on Build GRCh38
rs368573638 Chr.17: 46549015-46549015 on Build GRCh38
rs780399763 Chr.17: 46548895-46548895 on Build GRCh38
rs776891979 Chr.17: 46548792-46548792 on Build GRCh38
rs372793996 Chr.17: 46548819-46548819 on Build GRCh38
rs768665639 Chr.17: 46548843-46548843 on Build GRCh38
rs757066349 Chr.17: 46548766-46548766 on Build GRCh38
rs201311857 Chr.17: 46548935-46548935 on Build GRCh38
rs771228816 Chr.17: 46549062-46549062 on Build GRCh38
rs775169826 Chr.17: 46549014-46549014 on Build GRCh38
rs780524421 Chr.17: 46548936-46548936 on Build GRCh38
rs772184684 Chr.17: 46548951-46548951 on Build GRCh38
rs766214240 Chr.17: 46548906-46548906 on Build GRCh38
rs749670633 Chr.17: 46548944-46548944 on Build GRCh38
rs576535731 Chr.17: 46548925-46548925 on Build GRCh38
rs756871417 Chr.17: 46548712-46548712 on Build GRCh38
rs765263726 Chr.17: 46548681-46548681 on Build GRCh38
rs16963324 Chr.17: 46548685-46548685 on Build GRCh38
rs755167667 Chr.17: 46548677-46548677 on Build GRCh38
rs781778252 Chr.17: 46548885-46548885 on Build GRCh38
rs199829673 Chr.17: 46548743-46548743 on Build GRCh38
rs753037263 Chr.17: 46549033-46549033 on Build GRCh38
rs753533305 Chr.17: 46548871-46548871 on Build GRCh38
rs749596377 Chr.17: 46549045-46549045 on Build GRCh38
rs746622571 Chr.17: 46548635-46548635 on Build GRCh38
rs769134814 Chr.17: 46548645-46548645 on Build GRCh38
rs772623942 Chr.17: 46549081-46549081 on Build GRCh38
rs200096015 Chr.17: 46548903-46548903 on Build GRCh38
rs779500864 Chr.17: 46548779-46548779 on Build GRCh38
rs770322170 Chr.17: 46548891-46548891 on Build GRCh38
rs928538810 Chr.17: 46549057-46549057 on Build GRCh38
rs773115016 Chr.17: 46548953-46548953 on Build GRCh38
rs758644419 Chr.17: 46548996-46548996 on Build GRCh38
rs371330917 Chr.17: 46548731-46548731 on Build GRCh38
rs780864852 Chr.17: 46548767-46548767 on Build GRCh38
rs752254692 Chr.17: 46548878-46548878 on Build GRCh38
rs747521200 Chr.17: 46548830-46548830 on Build GRCh38
rs273530 Chr.17: 46548954-46548954 on Build GRCh38
rs774230393 Chr.17: 46548900-46548900 on Build GRCh38
rs761469144 Chr.17: 46549020-46549020 on Build GRCh38
rs562756647 Chr.17: 46548917-46548917 on Build GRCh38
rs756595496 Chr.17: 46548939-46548939 on Build GRCh38
rs760434987 Chr.17: 46548904-46548904 on Build GRCh38
rs553938756 Chr.17: 46548851-46548851 on Build GRCh38
rs374361415 Chr.17: 46549012-46549012 on Build GRCh38
rs745567007 Chr.17: 46548768-46548768 on Build GRCh38
rs757740581 Chr.17: 46548683-46548683 on Build GRCh38
rs748456067 Chr.17: 46548992-46548992 on Build GRCh38
rs751631615 Chr.17: 46549051-46549051 on Build GRCh38
rs771243099 Chr.17: 46548791-46548791 on Build GRCh38
rs772599065 Chr.17: 46548738-46548738 on Build GRCh38

View Full Product Details