Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48100955-48101461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX1
Entrez Gene ID
10951
Gene Name
chromobox 1
Gene Aliases
CBX, HP1-BETA, HP1Hs-beta, HP1Hsbeta, M31, MOD1, p25beta
UniGene
Hs.77254
Gene Chromosome Location
Chr.17: 48070052-48101521 on Build GRCh38
Transcripts
NM_001127228.1 NM_006807.4

SNP Detail

SNP ID Chromosome Location
rs978830581 Chr.17: 48101020-48101020 on Build GRCh38
rs998460972 Chr.17: 48101270-48101271 on Build GRCh38
rs998573087 Chr.17: 48101266-48101266 on Build GRCh38
rs1056172334 Chr.17: 48101160-48101160 on Build GRCh38
rs1018766134 Chr.17: 48101239-48101239 on Build GRCh38
rs775369726 Chr.17: 48101410-48101410 on Build GRCh38
rs945273178 Chr.17: 48101389-48101389 on Build GRCh38
rs1043514343 Chr.17: 48101193-48101193 on Build GRCh38
rs777897517 Chr.17: 48101127-48101127 on Build GRCh38
rs976811471 Chr.17: 48101038-48101038 on Build GRCh38
rs375669887 Chr.17: 48101439-48101439 on Build GRCh38
rs1056523791 Chr.17: 48101369-48101369 on Build GRCh38
rs142673229 Chr.17: 48101411-48101411 on Build GRCh38
rs138183362 Chr.17: 48101308-48101308 on Build GRCh38
rs1016015229 Chr.17: 48101360-48101360 on Build GRCh38
rs374708682 Chr.17: 48101331-48101331 on Build GRCh38
rs910213938 Chr.17: 48101342-48101342 on Build GRCh38
rs901419649 Chr.17: 48101245-48101245 on Build GRCh38
rs1043380876 Chr.17: 48101418-48101418 on Build GRCh38
rs909007056 Chr.17: 48101117-48101117 on Build GRCh38
rs983049173 Chr.17: 48101289-48101289 on Build GRCh38
rs951580478 Chr.17: 48101287-48101287 on Build GRCh38
rs1045219821 Chr.17: 48101149-48101149 on Build GRCh38
rs774279766 Chr.17: 48101155-48101155 on Build GRCh38
rs892368101 Chr.17: 48101163-48101163 on Build GRCh38
rs1022185769 Chr.17: 48101015-48101015 on Build GRCh38
rs979915449 Chr.17: 48101414-48101414 on Build GRCh38
rs998292745 Chr.17: 48101436-48101436 on Build GRCh38
rs559085084 Chr.17: 48101070-48101070 on Build GRCh38
rs910010823 Chr.17: 48101307-48101307 on Build GRCh38
rs913247890 Chr.17: 48101165-48101165 on Build GRCh38
rs964396875 Chr.17: 48101033-48101033 on Build GRCh38
rs983240355 Chr.17: 48101098-48101098 on Build GRCh38
rs910064195 Chr.17: 48101089-48101089 on Build GRCh38
rs772046584 Chr.17: 48101084-48101084 on Build GRCh38
rs963215952 Chr.17: 48101141-48101141 on Build GRCh38
rs1038888312 Chr.17: 48101424-48101424 on Build GRCh38
rs3744347 Chr.17: 48101312-48101312 on Build GRCh38
rs1000249796 Chr.17: 48101230-48101230 on Build GRCh38
rs926276603 Chr.17: 48101150-48101150 on Build GRCh38
rs922763373 Chr.17: 48101233-48101233 on Build GRCh38
rs913198289 Chr.17: 48101438-48101441 on Build GRCh38
rs539083104 Chr.17: 48101055-48101055 on Build GRCh38
rs950489907 Chr.17: 48101096-48101096 on Build GRCh38
rs997572314 Chr.17: 48101063-48101063 on Build GRCh38
rs941064136 Chr.17: 48101104-48101104 on Build GRCh38
rs939007317 Chr.17: 48101151-48101151 on Build GRCh38
rs4794445 Chr.17: 48101434-48101434 on Build GRCh38
rs891859172 Chr.17: 48100985-48100985 on Build GRCh38
rs534828544 Chr.17: 48101164-48101164 on Build GRCh38
rs1030026137 Chr.17: 48101087-48101087 on Build GRCh38
rs371360245 Chr.17: 48101241-48101241 on Build GRCh38
rs564612746 Chr.17: 48101423-48101423 on Build GRCh38
rs1009352806 Chr.17: 48101004-48101004 on Build GRCh38
rs943027791 Chr.17: 48101344-48101344 on Build GRCh38
rs570019162 Chr.17: 48101023-48101023 on Build GRCh38
rs1029692588 Chr.17: 48101278-48101278 on Build GRCh38
rs939138269 Chr.17: 48101362-48101362 on Build GRCh38
rs904648013 Chr.17: 48101207-48101208 on Build GRCh38
rs913585768 Chr.17: 48101385-48101385 on Build GRCh38
rs572536750 Chr.17: 48101143-48101143 on Build GRCh38
rs553896579 Chr.17: 48101171-48101171 on Build GRCh38
rs1022109965 Chr.17: 48101011-48101011 on Build GRCh38
rs992780032 Chr.17: 48101435-48101435 on Build GRCh38
rs968763381 Chr.17: 48101014-48101014 on Build GRCh38
rs867040246 Chr.17: 48101102-48101102 on Build GRCh38
rs372091793 Chr.17: 48101422-48101422 on Build GRCh38
rs980998057 Chr.17: 48101310-48101310 on Build GRCh38
rs1010351628 Chr.17: 48100999-48100999 on Build GRCh38
rs903716010 Chr.17: 48101216-48101216 on Build GRCh38
rs575621417 Chr.17: 48101415-48101415 on Build GRCh38
rs922839892 Chr.17: 48101041-48101041 on Build GRCh38

View Full Product Details