Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48101128-48101630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX1
Entrez Gene ID
10951
Gene Name
chromobox 1
Gene Aliases
CBX, HP1-BETA, HP1Hs-beta, HP1Hsbeta, M31, MOD1, p25beta
UniGene
Hs.77254
Gene Chromosome Location
Chr.17: 48070052-48101521 on Build GRCh38
Transcripts
NM_001127228.1 NM_006807.4

SNP Detail

SNP ID Chromosome Location
rs934029546 Chr.17: 48101547-48101547 on Build GRCh38
rs998460972 Chr.17: 48101270-48101271 on Build GRCh38
rs998573087 Chr.17: 48101266-48101266 on Build GRCh38
rs1056172334 Chr.17: 48101160-48101160 on Build GRCh38
rs1018766134 Chr.17: 48101239-48101239 on Build GRCh38
rs962994784 Chr.17: 48101473-48101473 on Build GRCh38
rs775369726 Chr.17: 48101410-48101410 on Build GRCh38
rs945273178 Chr.17: 48101389-48101389 on Build GRCh38
rs898730354 Chr.17: 48101579-48101579 on Build GRCh38
rs1043514343 Chr.17: 48101193-48101193 on Build GRCh38
rs561494861 Chr.17: 48101524-48101524 on Build GRCh38
rs375669887 Chr.17: 48101439-48101439 on Build GRCh38
rs1056523791 Chr.17: 48101369-48101369 on Build GRCh38
rs887225394 Chr.17: 48101567-48101567 on Build GRCh38
rs138183362 Chr.17: 48101308-48101308 on Build GRCh38
rs142673229 Chr.17: 48101411-48101411 on Build GRCh38
rs1016015229 Chr.17: 48101360-48101360 on Build GRCh38
rs910213938 Chr.17: 48101342-48101342 on Build GRCh38
rs374708682 Chr.17: 48101331-48101331 on Build GRCh38
rs901419649 Chr.17: 48101245-48101245 on Build GRCh38
rs1043380876 Chr.17: 48101418-48101418 on Build GRCh38
rs532299257 Chr.17: 48101530-48101530 on Build GRCh38
rs926038196 Chr.17: 48101506-48101506 on Build GRCh38
rs983049173 Chr.17: 48101289-48101289 on Build GRCh38
rs951580478 Chr.17: 48101287-48101287 on Build GRCh38
rs201387184 Chr.17: 48101521-48101522 on Build GRCh38
rs1045219821 Chr.17: 48101149-48101149 on Build GRCh38
rs892368101 Chr.17: 48101163-48101163 on Build GRCh38
rs774279766 Chr.17: 48101155-48101155 on Build GRCh38
rs1004435318 Chr.17: 48101457-48101457 on Build GRCh38
rs979915449 Chr.17: 48101414-48101414 on Build GRCh38
rs369727833 Chr.17: 48101528-48101528 on Build GRCh38
rs945033996 Chr.17: 48101500-48101500 on Build GRCh38
rs998292745 Chr.17: 48101436-48101436 on Build GRCh38
rs910010823 Chr.17: 48101307-48101307 on Build GRCh38
rs913247890 Chr.17: 48101165-48101165 on Build GRCh38
rs901693916 Chr.17: 48101546-48101546 on Build GRCh38
rs913531958 Chr.17: 48101499-48101499 on Build GRCh38
rs1051188052 Chr.17: 48101548-48101548 on Build GRCh38
rs540786833 Chr.17: 48101480-48101480 on Build GRCh38
rs570404211 Chr.17: 48101555-48101555 on Build GRCh38
rs1038888312 Chr.17: 48101424-48101424 on Build GRCh38
rs1017190857 Chr.17: 48101464-48101464 on Build GRCh38
rs3744347 Chr.17: 48101312-48101312 on Build GRCh38
rs1000249796 Chr.17: 48101230-48101230 on Build GRCh38
rs527354678 Chr.17: 48101453-48101453 on Build GRCh38
rs926276603 Chr.17: 48101150-48101150 on Build GRCh38
rs922763373 Chr.17: 48101233-48101233 on Build GRCh38
rs913198289 Chr.17: 48101438-48101441 on Build GRCh38
rs998820369 Chr.17: 48101525-48101525 on Build GRCh38
rs951361649 Chr.17: 48101456-48101456 on Build GRCh38
rs942823448 Chr.17: 48101519-48101519 on Build GRCh38
rs530236165 Chr.17: 48101526-48101526 on Build GRCh38
rs939007317 Chr.17: 48101151-48101151 on Build GRCh38
rs4794445 Chr.17: 48101434-48101434 on Build GRCh38
rs534828544 Chr.17: 48101164-48101164 on Build GRCh38
rs371360245 Chr.17: 48101241-48101241 on Build GRCh38
rs887385482 Chr.17: 48101540-48101540 on Build GRCh38
rs979104974 Chr.17: 48101522-48101522 on Build GRCh38
rs564612746 Chr.17: 48101423-48101423 on Build GRCh38
rs146015886 Chr.17: 48101599-48101599 on Build GRCh38
rs991942381 Chr.17: 48101493-48101493 on Build GRCh38
rs532535154 Chr.17: 48101569-48101569 on Build GRCh38
rs943027791 Chr.17: 48101344-48101344 on Build GRCh38
rs1029692588 Chr.17: 48101278-48101278 on Build GRCh38
rs939138269 Chr.17: 48101362-48101362 on Build GRCh38
rs904648013 Chr.17: 48101207-48101208 on Build GRCh38
rs913585768 Chr.17: 48101385-48101385 on Build GRCh38
rs1016971164 Chr.17: 48101575-48101575 on Build GRCh38
rs1039250365 Chr.17: 48101543-48101543 on Build GRCh38
rs553896579 Chr.17: 48101171-48101171 on Build GRCh38
rs981329712 Chr.17: 48101478-48101478 on Build GRCh38
rs992780032 Chr.17: 48101435-48101435 on Build GRCh38
rs768383850 Chr.17: 48101479-48101479 on Build GRCh38
rs372091793 Chr.17: 48101422-48101422 on Build GRCh38
rs980998057 Chr.17: 48101310-48101310 on Build GRCh38
rs761524852 Chr.17: 48101481-48101481 on Build GRCh38
rs903716010 Chr.17: 48101216-48101216 on Build GRCh38
rs575621417 Chr.17: 48101415-48101415 on Build GRCh38
rs1004214437 Chr.17: 48101574-48101574 on Build GRCh38

View Full Product Details