Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49141912-49142341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
UniGene
Hs.374679
Gene Chromosome Location
Chr.17: 49120347-49170722 on Build GRCh38
Transcripts
NM_001159387.1 NM_001159388.1 NM_153446.2 XM_011524314.2 XM_017024173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274721 c.404C>T p.P135L Chr.17: 49142043-49142043 on Build GRCh38
1640727 c.519A>C p.E173D Chr.17: 49142158-49142158 on Build GRCh38
6147368 c.396-1G>T p.? Chr.17: 49142034-49142034 on Build GRCh38
3958503 c.484C>G p.P162A Chr.17: 49142123-49142123 on Build GRCh38
4895277 c.443G>A p.G148E Chr.17: 49142082-49142082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757776690 Chr.17: 49142217-49142217 on Build GRCh38
rs554531643 Chr.17: 49142250-49142250 on Build GRCh38
rs751565522 Chr.17: 49142199-49142199 on Build GRCh38
rs149412440 Chr.17: 49142116-49142116 on Build GRCh38
rs749860075 Chr.17: 49142067-49142067 on Build GRCh38
rs112283922 Chr.17: 49142305-49142305 on Build GRCh38
rs1039364076 Chr.17: 49142278-49142278 on Build GRCh38
rs148596818 Chr.17: 49142169-49142169 on Build GRCh38
rs935736136 Chr.17: 49142274-49142274 on Build GRCh38
rs150222532 Chr.17: 49141971-49141971 on Build GRCh38
rs12453819 Chr.17: 49142180-49142180 on Build GRCh38
rs760387297 Chr.17: 49142264-49142264 on Build GRCh38
rs371713741 Chr.17: 49142057-49142057 on Build GRCh38
rs754271456 Chr.17: 49142216-49142216 on Build GRCh38
rs776330460 Chr.17: 49142177-49142177 on Build GRCh38
rs753693865 Chr.17: 49141999-49141999 on Build GRCh38
rs144833149 Chr.17: 49142127-49142127 on Build GRCh38
rs940308211 Chr.17: 49142281-49142281 on Build GRCh38
rs919951290 Chr.17: 49142297-49142297 on Build GRCh38
rs767544957 Chr.17: 49142203-49142203 on Build GRCh38
rs768197366 Chr.17: 49141987-49141987 on Build GRCh38
rs765600676 Chr.17: 49142131-49142131 on Build GRCh38
rs750751647 Chr.17: 49142217-49142218 on Build GRCh38
rs771679760 Chr.17: 49142071-49142071 on Build GRCh38
rs186837742 Chr.17: 49142002-49142002 on Build GRCh38
rs201550101 Chr.17: 49142100-49142100 on Build GRCh38
rs374155308 Chr.17: 49142166-49142166 on Build GRCh38
rs183217497 Chr.17: 49141970-49141970 on Build GRCh38
rs552933724 Chr.17: 49142128-49142128 on Build GRCh38
rs368001777 Chr.17: 49141994-49141994 on Build GRCh38
rs750335597 Chr.17: 49142121-49142121 on Build GRCh38
rs755097459 Chr.17: 49142135-49142135 on Build GRCh38
rs765171809 Chr.17: 49142075-49142075 on Build GRCh38
rs957045074 Chr.17: 49142214-49142214 on Build GRCh38
rs763824485 Chr.17: 49142109-49142109 on Build GRCh38
rs986897510 Chr.17: 49142265-49142265 on Build GRCh38
rs780155086 Chr.17: 49142031-49142031 on Build GRCh38
rs760358504 Chr.17: 49142105-49142105 on Build GRCh38
rs749775812 Chr.17: 49142144-49142144 on Build GRCh38
rs1039333025 Chr.17: 49141978-49141978 on Build GRCh38
rs759370660 Chr.17: 49142010-49142010 on Build GRCh38
rs954216628 Chr.17: 49142259-49142259 on Build GRCh38
rs748550373 Chr.17: 49142044-49142044 on Build GRCh38
rs773758575 Chr.17: 49142063-49142063 on Build GRCh38
rs781473343 Chr.17: 49142136-49142136 on Build GRCh38
rs760446465 Chr.17: 49141992-49141992 on Build GRCh38
rs543838419 Chr.17: 49142263-49142263 on Build GRCh38
rs754315435 Chr.17: 49142041-49142041 on Build GRCh38
rs925734495 Chr.17: 49142273-49142273 on Build GRCh38
rs779554542 Chr.17: 49142221-49142221 on Build GRCh38
rs757732113 Chr.17: 49142044-49142045 on Build GRCh38
rs756324945 Chr.17: 49142209-49142209 on Build GRCh38
rs761417339 Chr.17: 49142179-49142179 on Build GRCh38
rs771591485 Chr.17: 49142165-49142165 on Build GRCh38
rs532174062 Chr.17: 49141966-49141966 on Build GRCh38
rs775174162 Chr.17: 49142086-49142086 on Build GRCh38
rs766316761 Chr.17: 49142185-49142185 on Build GRCh38
rs759114156 Chr.17: 49142020-49142026 on Build GRCh38
rs201539211 Chr.17: 49142043-49142043 on Build GRCh38
rs752920813 Chr.17: 49142137-49142137 on Build GRCh38
rs750457476 Chr.17: 49142286-49142286 on Build GRCh38
rs200147969 Chr.17: 49142172-49142172 on Build GRCh38
rs778237209 Chr.17: 49142213-49142213 on Build GRCh38
rs565993034 Chr.17: 49142142-49142142 on Build GRCh38
rs755223890 Chr.17: 49142041-49142041 on Build GRCh38
rs765222341 Chr.17: 49142013-49142013 on Build GRCh38
rs1037475950 Chr.17: 49142101-49142101 on Build GRCh38
rs766944146 Chr.17: 49142025-49142025 on Build GRCh38
rs775261715 Chr.17: 49141990-49141990 on Build GRCh38
rs752935494 Chr.17: 49142206-49142206 on Build GRCh38
rs762827940 Chr.17: 49142183-49142183 on Build GRCh38
rs747199055 Chr.17: 49142036-49142036 on Build GRCh38
rs759649122 Chr.17: 49142200-49142200 on Build GRCh38
rs764997159 Chr.17: 49142117-49142117 on Build GRCh38
rs945909262 Chr.17: 49142229-49142229 on Build GRCh38
rs758457399 Chr.17: 49142030-49142030 on Build GRCh38
rs957387550 Chr.17: 49141969-49141969 on Build GRCh38
rs932791089 Chr.17: 49141997-49141997 on Build GRCh38
rs750390315 Chr.17: 49142029-49142029 on Build GRCh38
rs776219183 Chr.17: 49142115-49142115 on Build GRCh38

View Full Product Details