Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49156459-49156951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
UniGene
Hs.374679
Gene Chromosome Location
Chr.17: 49120347-49170722 on Build GRCh38
Transcripts
NM_001159387.1 NM_001159388.1 NM_153446.2 XM_011524314.2 XM_017024173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016693 c.641-3C>T p.? Chr.17: 49156563-49156563 on Build GRCh38
3518880 c.661G>A p.D221N Chr.17: 49156586-49156586 on Build GRCh38
3518879 c.645C>T p.L215L Chr.17: 49156570-49156570 on Build GRCh38
4787927 c.666C>T p.A222A Chr.17: 49156591-49156591 on Build GRCh38
4567302 c.655_656GG>AA p.G219K Chr.17: 49156580-49156581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776879040 Chr.17: 49156586-49156586 on Build GRCh38
rs979490130 Chr.17: 49156746-49156746 on Build GRCh38
rs148015203 Chr.17: 49156854-49156854 on Build GRCh38
rs971801530 Chr.17: 49156509-49156509 on Build GRCh38
rs780239716 Chr.17: 49156546-49156547 on Build GRCh38
rs968011985 Chr.17: 49156926-49156926 on Build GRCh38
rs758880608 Chr.17: 49156519-49156519 on Build GRCh38
rs1042111328 Chr.17: 49156782-49156782 on Build GRCh38
rs989991890 Chr.17: 49156793-49156793 on Build GRCh38
rs147804520 Chr.17: 49156585-49156585 on Build GRCh38
rs775280574 Chr.17: 49156564-49156564 on Build GRCh38
rs753361854 Chr.17: 49156632-49156632 on Build GRCh38
rs751980377 Chr.17: 49156611-49156611 on Build GRCh38
rs1010020276 Chr.17: 49156689-49156689 on Build GRCh38
rs140280086 Chr.17: 49156592-49156592 on Build GRCh38
rs560373909 Chr.17: 49156590-49156590 on Build GRCh38
rs971451159 Chr.17: 49156730-49156730 on Build GRCh38
rs760068943 Chr.17: 49156621-49156621 on Build GRCh38
rs747011773 Chr.17: 49156571-49156571 on Build GRCh38
rs756839940 Chr.17: 49156634-49156634 on Build GRCh38
rs371909190 Chr.17: 49156518-49156518 on Build GRCh38
rs1009784443 Chr.17: 49156913-49156913 on Build GRCh38
rs2293217 Chr.17: 49156755-49156755 on Build GRCh38
rs2293216 Chr.17: 49156860-49156860 on Build GRCh38
rs762075860 Chr.17: 49156591-49156591 on Build GRCh38
rs914371360 Chr.17: 49156802-49156802 on Build GRCh38
rs1002473206 Chr.17: 49156655-49156655 on Build GRCh38
rs1044509659 Chr.17: 49156925-49156925 on Build GRCh38
rs533702167 Chr.17: 49156683-49156683 on Build GRCh38
rs887979974 Chr.17: 49156868-49156868 on Build GRCh38
rs1003268680 Chr.17: 49156734-49156734 on Build GRCh38
rs765395555 Chr.17: 49156839-49156839 on Build GRCh38
rs371182343 Chr.17: 49156895-49156895 on Build GRCh38
rs955881666 Chr.17: 49156533-49156533 on Build GRCh38
rs548211983 Chr.17: 49156530-49156530 on Build GRCh38
rs778714924 Chr.17: 49156552-49156552 on Build GRCh38
rs755599431 Chr.17: 49156534-49156534 on Build GRCh38
rs766800055 Chr.17: 49156521-49156521 on Build GRCh38
rs1049184035 Chr.17: 49156645-49156645 on Build GRCh38
rs1049865857 Chr.17: 49156581-49156581 on Build GRCh38
rs182085500 Chr.17: 49156890-49156890 on Build GRCh38
rs748900433 Chr.17: 49156545-49156545 on Build GRCh38
rs147824605 Chr.17: 49156562-49156562 on Build GRCh38
rs199988212 Chr.17: 49156576-49156576 on Build GRCh38
rs887863892 Chr.17: 49156647-49156647 on Build GRCh38
rs200397463 Chr.17: 49156560-49156560 on Build GRCh38
rs964249919 Chr.17: 49156772-49156772 on Build GRCh38
rs756927926 Chr.17: 49156551-49156551 on Build GRCh38
rs759740665 Chr.17: 49156816-49156816 on Build GRCh38
rs373391531 Chr.17: 49156649-49156649 on Build GRCh38
rs148865659 Chr.17: 49156626-49156626 on Build GRCh38
rs542811196 Chr.17: 49156843-49156843 on Build GRCh38
rs777305349 Chr.17: 49156541-49156541 on Build GRCh38
rs572128158 Chr.17: 49156741-49156741 on Build GRCh38
rs960967774 Chr.17: 49156676-49156676 on Build GRCh38
rs922550458 Chr.17: 49156834-49156834 on Build GRCh38
rs747295227 Chr.17: 49156591-49156609 on Build GRCh38
rs985264553 Chr.17: 49156836-49156836 on Build GRCh38
rs1056631016 Chr.17: 49156879-49156879 on Build GRCh38
rs764667014 Chr.17: 49156891-49156891 on Build GRCh38
rs1013040484 Chr.17: 49156670-49156670 on Build GRCh38
rs768574863 Chr.17: 49156503-49156503 on Build GRCh38
rs376911707 Chr.17: 49156594-49156594 on Build GRCh38
rs1040000778 Chr.17: 49156546-49156546 on Build GRCh38
rs149941656 Chr.17: 49156595-49156595 on Build GRCh38
rs143510096 Chr.17: 49156852-49156852 on Build GRCh38
rs1031612210 Chr.17: 49156754-49156754 on Build GRCh38
rs975256261 Chr.17: 49156806-49156806 on Build GRCh38
rs989959367 Chr.17: 49156557-49156557 on Build GRCh38
rs929519346 Chr.17: 49156859-49156859 on Build GRCh38
rs1005224255 Chr.17: 49156622-49156622 on Build GRCh38
rs946779451 Chr.17: 49156494-49156494 on Build GRCh38
rs368743153 Chr.17: 49156643-49156643 on Build GRCh38

View Full Product Details