Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49206427-49206937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNGT2
Entrez Gene ID
2793
Gene Name
G protein subunit gamma transducin 2
Gene Aliases
G-GAMMA-8, G-GAMMA-C, GNG8, GNG9, GNGT8
UniGene
Hs.181781
Gene Chromosome Location
Chr.17: 49206234-49210574 on Build GRCh38
Transcripts
NM_001198754.1 NM_001198755.1 NM_001198756.1 NM_031498.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067466 c.138C>T p.N46N Chr.17: 49206829-49206829 on Build GRCh38
980762 c.136A>C p.N46H Chr.17: 49206831-49206831 on Build GRCh38
4067467 c.130G>T p.A44S Chr.17: 49206837-49206837 on Build GRCh38
1208421 c.95C>T p.A32V Chr.17: 49206872-49206872 on Build GRCh38
141508 c.148C>T p.L50F Chr.17: 49206819-49206819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017285919 Chr.17: 49206746-49206746 on Build GRCh38
rs552013035 Chr.17: 49206614-49206614 on Build GRCh38
rs1040268556 Chr.17: 49206868-49206868 on Build GRCh38
rs1005987343 Chr.17: 49206914-49206914 on Build GRCh38
rs759764285 Chr.17: 49206863-49206863 on Build GRCh38
rs934119896 Chr.17: 49206908-49206908 on Build GRCh38
rs757812179 Chr.17: 49206801-49206801 on Build GRCh38
rs73338390 Chr.17: 49206698-49206698 on Build GRCh38
rs377007222 Chr.17: 49206850-49206850 on Build GRCh38
rs774408069 Chr.17: 49206756-49206756 on Build GRCh38
rs751167025 Chr.17: 49206828-49206828 on Build GRCh38
rs773268505 Chr.17: 49206749-49206749 on Build GRCh38
rs1050250985 Chr.17: 49206716-49206716 on Build GRCh38
rs771436509 Chr.17: 49206511-49206511 on Build GRCh38
rs369600299 Chr.17: 49206751-49206751 on Build GRCh38
rs201646997 Chr.17: 49206777-49206777 on Build GRCh38
rs534482384 Chr.17: 49206775-49206775 on Build GRCh38
rs776436299 Chr.17: 49206722-49206722 on Build GRCh38
rs111300830 Chr.17: 49206900-49206900 on Build GRCh38
rs562012275 Chr.17: 49206512-49206512 on Build GRCh38
rs1020682709 Chr.17: 49206832-49206832 on Build GRCh38
rs926196583 Chr.17: 49206489-49206489 on Build GRCh38
rs777130659 Chr.17: 49206567-49206567 on Build GRCh38
rs991911017 Chr.17: 49206805-49206805 on Build GRCh38
rs771092606 Chr.17: 49206752-49206752 on Build GRCh38
rs770885737 Chr.17: 49206515-49206515 on Build GRCh38
rs748023523 Chr.17: 49206723-49206723 on Build GRCh38
rs745801317 Chr.17: 49206776-49206776 on Build GRCh38
rs556681723 Chr.17: 49206887-49206887 on Build GRCh38
rs370999150 Chr.17: 49206871-49206871 on Build GRCh38
rs993083390 Chr.17: 49206466-49206466 on Build GRCh38
rs924921281 Chr.17: 49206886-49206886 on Build GRCh38
rs764557160 Chr.17: 49206906-49206906 on Build GRCh38
rs776497107 Chr.17: 49206693-49206693 on Build GRCh38
rs901763365 Chr.17: 49206869-49206869 on Build GRCh38
rs943171161 Chr.17: 49206829-49206829 on Build GRCh38
rs1023114725 Chr.17: 49206525-49206525 on Build GRCh38
rs533159623 Chr.17: 49206575-49206575 on Build GRCh38
rs769636427 Chr.17: 49206849-49206849 on Build GRCh38
rs747933291 Chr.17: 49206847-49206847 on Build GRCh38
rs914673272 Chr.17: 49206689-49206689 on Build GRCh38
rs189501677 Chr.17: 49206907-49206907 on Build GRCh38
rs976061226 Chr.17: 49206892-49206892 on Build GRCh38
rs145685499 Chr.17: 49206516-49206516 on Build GRCh38
rs988806425 Chr.17: 49206612-49206612 on Build GRCh38
rs568050345 Chr.17: 49206792-49206792 on Build GRCh38
rs541567570 Chr.17: 49206840-49206840 on Build GRCh38
rs564544093 Chr.17: 49206549-49206549 on Build GRCh38
rs759857617 Chr.17: 49206763-49206763 on Build GRCh38
rs537193118 Chr.17: 49206859-49206859 on Build GRCh38
rs56070338 Chr.17: 49206738-49206739 on Build GRCh38
rs376218315 Chr.17: 49206902-49206902 on Build GRCh38
rs545201539 Chr.17: 49206574-49206574 on Build GRCh38
rs55634267 Chr.17: 49206453-49206453 on Build GRCh38
rs761188599 Chr.17: 49206784-49206784 on Build GRCh38
rs764490517 Chr.17: 49206787-49206787 on Build GRCh38
rs147752514 Chr.17: 49206548-49206548 on Build GRCh38
rs888966383 Chr.17: 49206813-49206813 on Build GRCh38
rs779644009 Chr.17: 49206824-49206824 on Build GRCh38
rs769732022 Chr.17: 49206736-49206736 on Build GRCh38
rs960525700 Chr.17: 49206797-49206797 on Build GRCh38
rs768402494 Chr.17: 49206713-49206713 on Build GRCh38

View Full Product Details