Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49210135-49210643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI3
Entrez Gene ID
51225
Gene Name
ABI family member 3
Gene Aliases
NESH, SSH3BP3
UniGene
Hs.130719
Gene Chromosome Location
Chr.17: 49210227-49223225 on Build GRCh38
Transcripts
NM_001135186.1 NM_016428.2 XM_005257429.2 XM_011524873.1 XM_017024721.1 NM_001198754.1

Gene Symbol
GNGT2
Entrez Gene ID
2793
Gene Name
G protein subunit gamma transducin 2
Gene Aliases
G-GAMMA-8, G-GAMMA-C, GNG8, GNG9, GNGT8
UniGene
Hs.181781
Gene Chromosome Location
Chr.17: 49206234-49210574 on Build GRCh38
Transcripts
NM_001135186.1 NM_016428.2 XM_005257429.2 XM_011524873.1 XM_017024721.1 NM_001198754.1

SNP Detail

SNP ID Chromosome Location
rs564577594 Chr.17: 49210246-49210246 on Build GRCh38
rs930927064 Chr.17: 49210536-49210536 on Build GRCh38
rs12948271 Chr.17: 49210333-49210333 on Build GRCh38
rs1021741495 Chr.17: 49210540-49210540 on Build GRCh38
rs550378316 Chr.17: 49210534-49210534 on Build GRCh38
rs188837768 Chr.17: 49210613-49210613 on Build GRCh38
rs938484619 Chr.17: 49210489-49210489 on Build GRCh38
rs2233364 Chr.17: 49210289-49210289 on Build GRCh38
rs73338392 Chr.17: 49210204-49210204 on Build GRCh38
rs948657219 Chr.17: 49210432-49210432 on Build GRCh38
rs368713398 Chr.17: 49210159-49210159 on Build GRCh38
rs1052891510 Chr.17: 49210497-49210497 on Build GRCh38
rs1011309432 Chr.17: 49210516-49210516 on Build GRCh38
rs912508512 Chr.17: 49210411-49210411 on Build GRCh38
rs891560760 Chr.17: 49210504-49210504 on Build GRCh38
rs1028993993 Chr.17: 49210311-49210311 on Build GRCh38
rs73338394 Chr.17: 49210570-49210570 on Build GRCh38
rs1046984314 Chr.17: 49210594-49210594 on Build GRCh38
rs889445724 Chr.17: 49210549-49210549 on Build GRCh38
rs1025102005 Chr.17: 49210235-49210235 on Build GRCh38
rs1017766205 Chr.17: 49210312-49210312 on Build GRCh38
rs532694748 Chr.17: 49210571-49210571 on Build GRCh38
rs985225891 Chr.17: 49210389-49210389 on Build GRCh38
rs939913662 Chr.17: 49210456-49210456 on Build GRCh38
rs1017784453 Chr.17: 49210569-49210569 on Build GRCh38
rs530198239 Chr.17: 49210519-49210519 on Build GRCh38
rs919313807 Chr.17: 49210380-49210380 on Build GRCh38
rs137881034 Chr.17: 49210169-49210170 on Build GRCh38
rs907042294 Chr.17: 49210486-49210486 on Build GRCh38
rs759513872 Chr.17: 49210167-49210167 on Build GRCh38
rs995269125 Chr.17: 49210188-49210188 on Build GRCh38
rs377689573 Chr.17: 49210614-49210614 on Build GRCh38
rs967353806 Chr.17: 49210241-49210241 on Build GRCh38
rs929390550 Chr.17: 49210385-49210385 on Build GRCh38
rs764545556 Chr.17: 49210580-49210580 on Build GRCh38
rs1026469916 Chr.17: 49210586-49210586 on Build GRCh38
rs140527653 Chr.17: 49210476-49210487 on Build GRCh38
rs979839937 Chr.17: 49210545-49210545 on Build GRCh38
rs895320474 Chr.17: 49210199-49210199 on Build GRCh38
rs1044298351 Chr.17: 49210441-49210441 on Build GRCh38
rs1032772642 Chr.17: 49210257-49210257 on Build GRCh38
rs560322766 Chr.17: 49210388-49210388 on Build GRCh38
rs544585985 Chr.17: 49210211-49210211 on Build GRCh38
rs976207681 Chr.17: 49210344-49210344 on Build GRCh38
rs552602148 Chr.17: 49210612-49210612 on Build GRCh38
rs909204680 Chr.17: 49210399-49210399 on Build GRCh38
rs1021270908 Chr.17: 49210232-49210232 on Build GRCh38
rs986588007 Chr.17: 49210341-49210341 on Build GRCh38
rs149838793 Chr.17: 49210320-49210320 on Build GRCh38
rs1054847449 Chr.17: 49210198-49210198 on Build GRCh38
rs761739974 Chr.17: 49210559-49210559 on Build GRCh38
rs1054921935 Chr.17: 49210555-49210555 on Build GRCh38
rs894890421 Chr.17: 49210556-49210556 on Build GRCh38
rs561635226 Chr.17: 49210396-49210396 on Build GRCh38
rs796773379 Chr.17: 49210388-49210390 on Build GRCh38
rs1011634878 Chr.17: 49210215-49210215 on Build GRCh38
rs1025260890 Chr.17: 49210209-49210209 on Build GRCh38
rs898268178 Chr.17: 49210176-49210176 on Build GRCh38
rs1007774589 Chr.17: 49210550-49210550 on Build GRCh38
rs958001026 Chr.17: 49210261-49210261 on Build GRCh38

View Full Product Details