Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50373046-50373525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME1
Entrez Gene ID
146956
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
MMS4L, SLX2A
UniGene
Hs.514330
Gene Chromosome Location
Chr.17: 50373220-50381479 on Build GRCh38
Transcripts
NM_001166131.1 NM_152463.2 XM_005257081.3 XM_011524395.2 XM_017024236.1 NM_016504.2

Gene Symbol
MRPL27
Entrez Gene ID
51264
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
L27mt
UniGene
Hs.7736
Gene Chromosome Location
Chr.17: 50367867-50373201 on Build GRCh38
Transcripts
NM_001166131.1 NM_152463.2 XM_005257081.3 XM_011524395.2 XM_017024236.1 NM_016504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5015493 c.37G>A p.A13T Chr.17: 50373134-50373134 on Build GRCh38
4703206 c.38C>T p.A13V Chr.17: 50373133-50373133 on Build GRCh38
3937454 c.10G>T p.V4L Chr.17: 50373161-50373161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs925681979 Chr.17: 50373104-50373104 on Build GRCh38
rs773780458 Chr.17: 50373215-50373215 on Build GRCh38
rs184623385 Chr.17: 50373255-50373255 on Build GRCh38
rs908732811 Chr.17: 50373301-50373301 on Build GRCh38
rs192961184 Chr.17: 50373183-50373183 on Build GRCh38
rs752386559 Chr.17: 50373120-50373120 on Build GRCh38
rs926853249 Chr.17: 50373380-50373380 on Build GRCh38
rs891776095 Chr.17: 50373134-50373134 on Build GRCh38
rs16948975 Chr.17: 50373295-50373295 on Build GRCh38
rs367597280 Chr.17: 50373112-50373112 on Build GRCh38
rs758702613 Chr.17: 50373497-50373497 on Build GRCh38
rs1002130400 Chr.17: 50373177-50373177 on Build GRCh38
rs767188409 Chr.17: 50373119-50373119 on Build GRCh38
rs974364414 Chr.17: 50373337-50373337 on Build GRCh38
rs372549459 Chr.17: 50373166-50373166 on Build GRCh38
rs777116631 Chr.17: 50373085-50373085 on Build GRCh38
rs771249999 Chr.17: 50373220-50373221 on Build GRCh38
rs896534968 Chr.17: 50373186-50373186 on Build GRCh38
rs369873290 Chr.17: 50373453-50373453 on Build GRCh38
rs776704453 Chr.17: 50373225-50373226 on Build GRCh38
rs769733454 Chr.17: 50373439-50373439 on Build GRCh38
rs149938290 Chr.17: 50373098-50373098 on Build GRCh38
rs369778119 Chr.17: 50373428-50373428 on Build GRCh38
rs1020622824 Chr.17: 50373086-50373086 on Build GRCh38
rs201524116 Chr.17: 50373155-50373155 on Build GRCh38
rs773312574 Chr.17: 50373219-50373220 on Build GRCh38
rs757023737 Chr.17: 50373149-50373149 on Build GRCh38
rs572460785 Chr.17: 50373286-50373286 on Build GRCh38
rs763754044 Chr.17: 50373174-50373174 on Build GRCh38
rs779967420 Chr.17: 50373189-50373189 on Build GRCh38
rs753612911 Chr.17: 50373145-50373145 on Build GRCh38
rs980576694 Chr.17: 50373249-50373249 on Build GRCh38
rs963690567 Chr.17: 50373111-50373111 on Build GRCh38
rs16948970 Chr.17: 50373175-50373175 on Build GRCh38
rs374096806 Chr.17: 50373474-50373474 on Build GRCh38
rs199804651 Chr.17: 50373180-50373180 on Build GRCh38
rs768040080 Chr.17: 50373139-50373139 on Build GRCh38
rs768852960 Chr.17: 50373157-50373157 on Build GRCh38
rs938699833 Chr.17: 50373107-50373107 on Build GRCh38
rs750930134 Chr.17: 50373106-50373106 on Build GRCh38
rs1032917434 Chr.17: 50373253-50373253 on Build GRCh38
rs767611295 Chr.17: 50373167-50373167 on Build GRCh38
rs547685245 Chr.17: 50373223-50373223 on Build GRCh38
rs913269304 Chr.17: 50373422-50373422 on Build GRCh38
rs768886355 Chr.17: 50373082-50373082 on Build GRCh38
rs116081337 Chr.17: 50373243-50373243 on Build GRCh38
rs138005173 Chr.17: 50373153-50373153 on Build GRCh38
rs770184814 Chr.17: 50373209-50373209 on Build GRCh38
rs769190156 Chr.17: 50373178-50373181 on Build GRCh38
rs867117967 Chr.17: 50373420-50373420 on Build GRCh38
rs566829737 Chr.17: 50373184-50373185 on Build GRCh38
rs556823345 Chr.17: 50373227-50373227 on Build GRCh38
rs938805718 Chr.17: 50373278-50373278 on Build GRCh38
rs767132980 Chr.17: 50373170-50373170 on Build GRCh38
rs762085471 Chr.17: 50373087-50373087 on Build GRCh38
rs938015793 Chr.17: 50373412-50373412 on Build GRCh38
rs941509972 Chr.17: 50373302-50373302 on Build GRCh38
rs550246717 Chr.17: 50373443-50373443 on Build GRCh38
rs374090666 Chr.17: 50373152-50373152 on Build GRCh38
rs148670613 Chr.17: 50373171-50373171 on Build GRCh38
rs752873547 Chr.17: 50373390-50373390 on Build GRCh38
rs781448962 Chr.17: 50373205-50373205 on Build GRCh38
rs755048396 Chr.17: 50373203-50373203 on Build GRCh38
rs115623988 Chr.17: 50373178-50373178 on Build GRCh38
rs144070750 Chr.17: 50373163-50373163 on Build GRCh38
rs534336515 Chr.17: 50373237-50373237 on Build GRCh38
rs755899688 Chr.17: 50373140-50373140 on Build GRCh38
rs191349575 Chr.17: 50373482-50373482 on Build GRCh38
rs905917193 Chr.17: 50373099-50373099 on Build GRCh38
rs770052158 Chr.17: 50373214-50373215 on Build GRCh38
rs374744412 Chr.17: 50373198-50373198 on Build GRCh38
rs749677689 Chr.17: 50373217-50373217 on Build GRCh38
rs113983688 Chr.17: 50373162-50373162 on Build GRCh38
rs963721480 Chr.17: 50373257-50373257 on Build GRCh38
rs979078334 Chr.17: 50373103-50373103 on Build GRCh38
rs147365593 Chr.17: 50373142-50373142 on Build GRCh38
rs934168326 Chr.17: 50373141-50373141 on Build GRCh38
rs776728568 Chr.17: 50373160-50373160 on Build GRCh38
rs959877327 Chr.17: 50373081-50373081 on Build GRCh38
rs922351361 Chr.17: 50373274-50373274 on Build GRCh38
rs186932275 Chr.17: 50373260-50373260 on Build GRCh38
rs778620389 Chr.17: 50373179-50373179 on Build GRCh38

View Full Product Details