Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50575203-50575740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1G
Entrez Gene ID
8913
Gene Name
calcium voltage-gated channel subunit alpha1 G
Gene Aliases
Ca(V)T.1, Cav3.1, NBR13, SCA42
UniGene
Hs.591169
Gene Chromosome Location
Chr.17: 50560929-50629625 on Build GRCh38
Transcripts
NM_001256324.1 NM_001256325.1 NM_001256326.1 NM_001256327.1 NM_001256328.1 NM_001256329.1 NM_001256330.1 NM_001256331.1 NM_001256332.1 NM_001256333.1 NM_001256334.1 NM_001256359.1 NM_001256360.1 NM_001256361.1 NM_018896.4 NM_198376.2 NM_198377.2 NM_198378.2 NM_198379.2 NM_198380.2 NM_198382.2 NM_198383.2 NM_198384.2 NM_198385.2 NM_198386.2 NM_198387.2 NM_198388.2 NM_198396.2 NR_046054.1 NR_046055.1 NR_046056.1 NR_046057.1 NR_046058.1 XM_006722160.3 XM_006722161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3742312 c.1189C>G p.Q397E Chr.17: 50575591-50575591 on Build GRCh38
5386710 c.1222C>T p.L408L Chr.17: 50575624-50575624 on Build GRCh38
6532498 c.1303G>C p.E435Q Chr.17: 50575705-50575705 on Build GRCh38
5386702 c.1148C>T p.S383F Chr.17: 50575550-50575550 on Build GRCh38
473015 c.1185C>T p.A395A Chr.17: 50575587-50575587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs198550 Chr.17: 50575514-50575514 on Build GRCh38
rs16949166 Chr.17: 50575472-50575472 on Build GRCh38
rs749686862 Chr.17: 50575665-50575665 on Build GRCh38
rs182751909 Chr.17: 50575449-50575449 on Build GRCh38
rs925702083 Chr.17: 50575594-50575594 on Build GRCh38
rs77688102 Chr.17: 50575384-50575384 on Build GRCh38
rs757955804 Chr.17: 50575674-50575674 on Build GRCh38
rs765000164 Chr.17: 50575695-50575695 on Build GRCh38
rs373526821 Chr.17: 50575626-50575626 on Build GRCh38
rs756530578 Chr.17: 50575659-50575659 on Build GRCh38
rs779381563 Chr.17: 50575680-50575680 on Build GRCh38
rs746504110 Chr.17: 50575689-50575689 on Build GRCh38
rs1016981892 Chr.17: 50575444-50575444 on Build GRCh38
rs981184617 Chr.17: 50575244-50575244 on Build GRCh38
rs756728670 Chr.17: 50575533-50575533 on Build GRCh38
rs766648722 Chr.17: 50575450-50575450 on Build GRCh38
rs987308687 Chr.17: 50575535-50575535 on Build GRCh38
rs1024953961 Chr.17: 50575258-50575258 on Build GRCh38
rs925430131 Chr.17: 50575300-50575300 on Build GRCh38
rs753175840 Chr.17: 50575658-50575658 on Build GRCh38
rs562719792 Chr.17: 50575492-50575492 on Build GRCh38
rs1037852621 Chr.17: 50575421-50575421 on Build GRCh38
rs547658380 Chr.17: 50575566-50575566 on Build GRCh38
rs1013865211 Chr.17: 50575256-50575256 on Build GRCh38
rs763098527 Chr.17: 50575640-50575640 on Build GRCh38
rs755405289 Chr.17: 50575509-50575509 on Build GRCh38
rs781651848 Chr.17: 50575513-50575513 on Build GRCh38
rs767819939 Chr.17: 50575648-50575648 on Build GRCh38
rs1025923118 Chr.17: 50575372-50575372 on Build GRCh38
rs573685174 Chr.17: 50575338-50575338 on Build GRCh38
rs945924165 Chr.17: 50575312-50575312 on Build GRCh38
rs577828187 Chr.17: 50575266-50575266 on Build GRCh38
rs933214480 Chr.17: 50575357-50575357 on Build GRCh38
rs958104495 Chr.17: 50575314-50575314 on Build GRCh38
rs1042778715 Chr.17: 50575315-50575315 on Build GRCh38
rs960763193 Chr.17: 50575469-50575469 on Build GRCh38
rs938610604 Chr.17: 50575255-50575255 on Build GRCh38
rs370667544 Chr.17: 50575646-50575646 on Build GRCh38
rs746601255 Chr.17: 50575624-50575624 on Build GRCh38
rs761521704 Chr.17: 50575639-50575639 on Build GRCh38
rs748260403 Chr.17: 50575318-50575318 on Build GRCh38
rs376631282 Chr.17: 50575589-50575589 on Build GRCh38
rs768937046 Chr.17: 50575249-50575249 on Build GRCh38
rs368735882 Chr.17: 50575636-50575636 on Build GRCh38
rs778433453 Chr.17: 50575664-50575664 on Build GRCh38
rs954552135 Chr.17: 50575534-50575534 on Build GRCh38
rs984185002 Chr.17: 50575227-50575227 on Build GRCh38
rs958171091 Chr.17: 50575473-50575473 on Build GRCh38
rs540468820 Chr.17: 50575320-50575320 on Build GRCh38
rs557884190 Chr.17: 50575237-50575237 on Build GRCh38
rs562357237 Chr.17: 50575431-50575431 on Build GRCh38
rs766638827 Chr.17: 50575501-50575501 on Build GRCh38
rs751667201 Chr.17: 50575508-50575508 on Build GRCh38
rs774365845 Chr.17: 50575309-50575309 on Build GRCh38
rs775209826 Chr.17: 50575613-50575613 on Build GRCh38
rs375495099 Chr.17: 50575645-50575645 on Build GRCh38
rs1033922148 Chr.17: 50575245-50575245 on Build GRCh38
rs1057006786 Chr.17: 50575260-50575260 on Build GRCh38
rs776457023 Chr.17: 50575630-50575630 on Build GRCh38
rs999568214 Chr.17: 50575470-50575470 on Build GRCh38
rs766333339 Chr.17: 50575642-50575642 on Build GRCh38
rs761563364 Chr.17: 50575631-50575631 on Build GRCh38
rs910004021 Chr.17: 50575236-50575236 on Build GRCh38
rs978543203 Chr.17: 50575287-50575287 on Build GRCh38
rs198549 Chr.17: 50575445-50575445 on Build GRCh38
rs1025233886 Chr.17: 50575456-50575456 on Build GRCh38
rs765219241 Chr.17: 50575632-50575632 on Build GRCh38
rs901394334 Chr.17: 50575331-50575331 on Build GRCh38
rs138120428 Chr.17: 50575321-50575321 on Build GRCh38
rs907498143 Chr.17: 50575379-50575379 on Build GRCh38
rs201089692 Chr.17: 50575590-50575590 on Build GRCh38

View Full Product Details