Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 51630174-51630674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
UniGene
Hs.463466
Gene Chromosome Location
Chr.17: 51630313-52160017 on Build GRCh38
Transcripts
NM_001082533.1 NM_001082534.1 NM_020178.4 XM_017024878.1

SNP Detail

SNP ID Chromosome Location
rs1023691265 Chr.17: 51630234-51630234 on Build GRCh38
rs1016905900 Chr.17: 51630335-51630336 on Build GRCh38
rs535850353 Chr.17: 51630485-51630485 on Build GRCh38
rs918045538 Chr.17: 51630502-51630502 on Build GRCh38
rs770301118 Chr.17: 51630547-51630547 on Build GRCh38
rs79281434 Chr.17: 51630246-51630246 on Build GRCh38
rs528632222 Chr.17: 51630433-51630433 on Build GRCh38
rs1048987217 Chr.17: 51630505-51630505 on Build GRCh38
rs989729252 Chr.17: 51630289-51630289 on Build GRCh38
rs1050549908 Chr.17: 51630388-51630388 on Build GRCh38
rs897776297 Chr.17: 51630207-51630207 on Build GRCh38
rs894369542 Chr.17: 51630511-51630511 on Build GRCh38
rs527580188 Chr.17: 51630328-51630329 on Build GRCh38
rs549401307 Chr.17: 51630222-51630222 on Build GRCh38
rs994227103 Chr.17: 51630386-51630390 on Build GRCh38
rs1018068308 Chr.17: 51630431-51630431 on Build GRCh38
rs150103381 Chr.17: 51630208-51630209 on Build GRCh38
rs552826339 Chr.17: 51630462-51630463 on Build GRCh38
rs994758329 Chr.17: 51630219-51630219 on Build GRCh38
rs551929370 Chr.17: 51630273-51630273 on Build GRCh38
rs918979376 Chr.17: 51630608-51630608 on Build GRCh38
rs1008987146 Chr.17: 51630202-51630202 on Build GRCh38
rs879281028 Chr.17: 51630578-51630585 on Build GRCh38
rs955124346 Chr.17: 51630404-51630404 on Build GRCh38
rs1052554469 Chr.17: 51630279-51630279 on Build GRCh38
rs986337486 Chr.17: 51630566-51630566 on Build GRCh38
rs768787669 Chr.17: 51630353-51630354 on Build GRCh38
rs1012714140 Chr.17: 51630526-51630526 on Build GRCh38
rs935609505 Chr.17: 51630281-51630281 on Build GRCh38
rs558521286 Chr.17: 51630617-51630617 on Build GRCh38
rs771211640 Chr.17: 51630567-51630567 on Build GRCh38
rs375511228 Chr.17: 51630296-51630296 on Build GRCh38
rs982558211 Chr.17: 51630265-51630265 on Build GRCh38
rs1025330931 Chr.17: 51630220-51630220 on Build GRCh38
rs1024055226 Chr.17: 51630532-51630532 on Build GRCh38
rs912057295 Chr.17: 51630588-51630588 on Build GRCh38
rs1014552725 Chr.17: 51630206-51630206 on Build GRCh38
rs989856725 Chr.17: 51630542-51630542 on Build GRCh38
rs911179878 Chr.17: 51630233-51630233 on Build GRCh38
rs1045255488 Chr.17: 51630322-51630322 on Build GRCh38
rs925173129 Chr.17: 51630473-51630474 on Build GRCh38
rs182895107 Chr.17: 51630308-51630308 on Build GRCh38
rs769527349 Chr.17: 51630394-51630394 on Build GRCh38
rs778805610 Chr.17: 51630537-51630537 on Build GRCh38
rs879809297 Chr.17: 51630606-51630606 on Build GRCh38
rs896558624 Chr.17: 51630287-51630287 on Build GRCh38
rs3744664 Chr.17: 51630247-51630247 on Build GRCh38
rs988132102 Chr.17: 51630500-51630500 on Build GRCh38
rs1017953688 Chr.17: 51630648-51630648 on Build GRCh38
rs905328047 Chr.17: 51630326-51630326 on Build GRCh38
rs953570504 Chr.17: 51630561-51630561 on Build GRCh38
rs1048677859 Chr.17: 51630636-51630636 on Build GRCh38
rs187567345 Chr.17: 51630348-51630348 on Build GRCh38
rs3744663 Chr.17: 51630232-51630232 on Build GRCh38
rs890211644 Chr.17: 51630424-51630424 on Build GRCh38
rs1045144937 Chr.17: 51630519-51630519 on Build GRCh38
rs958515590 Chr.17: 51630231-51630231 on Build GRCh38
rs970818328 Chr.17: 51630536-51630536 on Build GRCh38
rs1036095508 Chr.17: 51630595-51630595 on Build GRCh38
rs144362878 Chr.17: 51630539-51630539 on Build GRCh38
rs184870993 Chr.17: 51630645-51630645 on Build GRCh38
rs142076004 Chr.17: 51630490-51630490 on Build GRCh38
rs531175862 Chr.17: 51630502-51630503 on Build GRCh38
rs192457236 Chr.17: 51630540-51630540 on Build GRCh38
rs72830531 Chr.17: 51630461-51630461 on Build GRCh38
rs944483634 Chr.17: 51630591-51630591 on Build GRCh38
rs554101260 Chr.17: 51630403-51630403 on Build GRCh38
rs779848451 Chr.17: 51630452-51630452 on Build GRCh38
rs369638091 Chr.17: 51630525-51630525 on Build GRCh38
rs1007943006 Chr.17: 51630639-51630640 on Build GRCh38

View Full Product Details