Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 51630573-51631080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
UniGene
Hs.463466
Gene Chromosome Location
Chr.17: 51630313-52160017 on Build GRCh38
Transcripts
NM_001082533.1 NM_001082534.1 NM_020178.4 XM_017024878.1

SNP Detail

SNP ID Chromosome Location
rs915562540 Chr.17: 51630936-51630936 on Build GRCh38
rs899572569 Chr.17: 51630943-51630943 on Build GRCh38
rs770566179 Chr.17: 51630936-51630937 on Build GRCh38
rs972614096 Chr.17: 51630855-51630855 on Build GRCh38
rs775836605 Chr.17: 51630974-51630974 on Build GRCh38
rs986368610 Chr.17: 51630784-51630784 on Build GRCh38
rs369549755 Chr.17: 51630771-51630771 on Build GRCh38
rs762119374 Chr.17: 51630699-51630700 on Build GRCh38
rs1032592890 Chr.17: 51630972-51630972 on Build GRCh38
rs1045639765 Chr.17: 51630950-51630950 on Build GRCh38
rs571812560 Chr.17: 51631054-51631054 on Build GRCh38
rs999070314 Chr.17: 51630712-51630712 on Build GRCh38
rs189945280 Chr.17: 51630780-51630780 on Build GRCh38
rs748980064 Chr.17: 51630934-51630935 on Build GRCh38
rs776307891 Chr.17: 51631050-51631050 on Build GRCh38
rs918979376 Chr.17: 51630608-51630608 on Build GRCh38
rs191981919 Chr.17: 51630816-51630816 on Build GRCh38
rs970693797 Chr.17: 51630740-51630740 on Build GRCh38
rs1051933431 Chr.17: 51630846-51630846 on Build GRCh38
rs1000744250 Chr.17: 51630966-51630966 on Build GRCh38
rs770399667 Chr.17: 51630949-51630949 on Build GRCh38
rs141500560 Chr.17: 51631040-51631054 on Build GRCh38
rs956971540 Chr.17: 51630735-51630735 on Build GRCh38
rs776864581 Chr.17: 51630705-51630705 on Build GRCh38
rs894402110 Chr.17: 51630664-51630664 on Build GRCh38
rs560617450 Chr.17: 51630714-51630714 on Build GRCh38
rs558521286 Chr.17: 51630617-51630617 on Build GRCh38
rs890807530 Chr.17: 51630854-51630854 on Build GRCh38
rs77188516 Chr.17: 51630706-51630706 on Build GRCh38
rs912088618 Chr.17: 51630801-51630801 on Build GRCh38
rs966257073 Chr.17: 51630829-51630829 on Build GRCh38
rs879809297 Chr.17: 51630606-51630606 on Build GRCh38
rs184487706 Chr.17: 51630823-51630823 on Build GRCh38
rs963933928 Chr.17: 51630665-51630665 on Build GRCh38
rs956831706 Chr.17: 51630708-51630708 on Build GRCh38
rs1007828515 Chr.17: 51630903-51630903 on Build GRCh38
rs745923351 Chr.17: 51630872-51630872 on Build GRCh38
rs1017953688 Chr.17: 51630648-51630648 on Build GRCh38
rs867126774 Chr.17: 51630827-51630827 on Build GRCh38
rs1048677859 Chr.17: 51630636-51630636 on Build GRCh38
rs1048756773 Chr.17: 51630918-51630918 on Build GRCh38
rs934514913 Chr.17: 51631027-51631027 on Build GRCh38
rs1031097209 Chr.17: 51630719-51630719 on Build GRCh38
rs1039456690 Chr.17: 51630935-51630935 on Build GRCh38
rs192787528 Chr.17: 51630760-51630760 on Build GRCh38
rs907092228 Chr.17: 51630991-51630991 on Build GRCh38
rs187533967 Chr.17: 51630698-51630698 on Build GRCh38
rs923943035 Chr.17: 51630779-51630780 on Build GRCh38
rs184618937 Chr.17: 51630779-51630779 on Build GRCh38
rs184870993 Chr.17: 51630645-51630645 on Build GRCh38
rs948660175 Chr.17: 51630941-51630941 on Build GRCh38
rs930352306 Chr.17: 51630875-51630875 on Build GRCh38
rs980876517 Chr.17: 51630753-51630753 on Build GRCh38
rs1007943006 Chr.17: 51630639-51630640 on Build GRCh38
rs919022799 Chr.17: 51630870-51630870 on Build GRCh38

View Full Product Details