Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56835030-56835537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKE
Entrez Gene ID
8526
Gene Name
diacylglycerol kinase epsilon
Gene Aliases
AHUS7, DAGK5, DAGK6, DGK, NPHS7
UniGene
Hs.239514
Gene Chromosome Location
Chr.17: 56834099-56869565 on Build GRCh38
Transcripts
NM_003647.2 XM_011525394.2 XM_011525395.2 XM_011525396.2 XM_017025243.1 XM_017025244.1 XM_017025245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3194614 c.456C>T p.C152C Chr.17: 56835251-56835251 on Build GRCh38
3194610 c.254C>T p.A85V Chr.17: 56835049-56835049 on Build GRCh38
3519899 c.305C>T p.A102V Chr.17: 56835100-56835100 on Build GRCh38
3519898 c.284A>C p.D95A Chr.17: 56835079-56835079 on Build GRCh38
417437 c.307G>C p.D103H Chr.17: 56835102-56835102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761622363 Chr.17: 56835318-56835318 on Build GRCh38
rs369611861 Chr.17: 56835107-56835107 on Build GRCh38
rs767609629 Chr.17: 56835071-56835071 on Build GRCh38
rs977825208 Chr.17: 56835388-56835388 on Build GRCh38
rs763932703 Chr.17: 56835187-56835187 on Build GRCh38
rs984097927 Chr.17: 56835491-56835491 on Build GRCh38
rs753647098 Chr.17: 56835316-56835316 on Build GRCh38
rs972843644 Chr.17: 56835173-56835173 on Build GRCh38
rs181891887 Chr.17: 56835383-56835383 on Build GRCh38
rs752488975 Chr.17: 56835077-56835077 on Build GRCh38
rs938904565 Chr.17: 56835171-56835171 on Build GRCh38
rs7225724 Chr.17: 56835376-56835376 on Build GRCh38
rs755966050 Chr.17: 56835083-56835083 on Build GRCh38
rs955650821 Chr.17: 56835505-56835505 on Build GRCh38
rs143220117 Chr.17: 56835233-56835233 on Build GRCh38
rs201433428 Chr.17: 56835287-56835287 on Build GRCh38
rs747298055 Chr.17: 56835138-56835138 on Build GRCh38
rs745902152 Chr.17: 56835220-56835220 on Build GRCh38
rs772683069 Chr.17: 56835149-56835149 on Build GRCh38
rs755487226 Chr.17: 56835351-56835351 on Build GRCh38
rs754101695 Chr.17: 56835386-56835386 on Build GRCh38
rs751180799 Chr.17: 56835065-56835065 on Build GRCh38
rs752681978 Chr.17: 56835180-56835180 on Build GRCh38
rs948987299 Chr.17: 56835457-56835457 on Build GRCh38
rs781570437 Chr.17: 56835355-56835355 on Build GRCh38
rs1021920540 Chr.17: 56835379-56835379 on Build GRCh38
rs758506570 Chr.17: 56835229-56835229 on Build GRCh38
rs766174427 Chr.17: 56835153-56835153 on Build GRCh38
rs747565175 Chr.17: 56835240-56835240 on Build GRCh38
rs1023029162 Chr.17: 56835080-56835080 on Build GRCh38
rs776855682 Chr.17: 56835431-56835433 on Build GRCh38
rs960259165 Chr.17: 56835398-56835398 on Build GRCh38
rs148302081 Chr.17: 56835257-56835257 on Build GRCh38
rs373468576 Chr.17: 56835195-56835195 on Build GRCh38
rs763982859 Chr.17: 56835302-56835302 on Build GRCh38
rs760515851 Chr.17: 56835337-56835337 on Build GRCh38
rs759138367 Chr.17: 56835158-56835158 on Build GRCh38
rs780169841 Chr.17: 56835341-56835341 on Build GRCh38
rs751630337 Chr.17: 56835347-56835347 on Build GRCh38
rs905525688 Chr.17: 56835375-56835375 on Build GRCh38
rs201209316 Chr.17: 56835098-56835098 on Build GRCh38
rs778963327 Chr.17: 56835110-56835110 on Build GRCh38
rs755804606 Chr.17: 56835181-56835181 on Build GRCh38
rs370071276 Chr.17: 56835396-56835396 on Build GRCh38
rs534521111 Chr.17: 56835445-56835445 on Build GRCh38
rs922860341 Chr.17: 56835204-56835204 on Build GRCh38
rs946844153 Chr.17: 56835250-56835250 on Build GRCh38
rs754934356 Chr.17: 56835066-56835066 on Build GRCh38
rs762662843 Chr.17: 56835150-56835150 on Build GRCh38
rs201997595 Chr.17: 56835113-56835113 on Build GRCh38
rs915029037 Chr.17: 56835123-56835123 on Build GRCh38
rs1004317526 Chr.17: 56835271-56835271 on Build GRCh38
rs777022657 Chr.17: 56835252-56835252 on Build GRCh38
rs762304023 Chr.17: 56835049-56835049 on Build GRCh38
rs775618307 Chr.17: 56835334-56835334 on Build GRCh38
rs200324699 Chr.17: 56835106-56835106 on Build GRCh38
rs564611390 Chr.17: 56835136-56835136 on Build GRCh38
rs770283642 Chr.17: 56835365-56835365 on Build GRCh38
rs748482164 Chr.17: 56835361-56835361 on Build GRCh38
rs779158220 Chr.17: 56835207-56835207 on Build GRCh38
rs757444386 Chr.17: 56835198-56835198 on Build GRCh38
rs758893686 Chr.17: 56835164-56835165 on Build GRCh38
rs972894256 Chr.17: 56835418-56835418 on Build GRCh38
rs771718146 Chr.17: 56835281-56835281 on Build GRCh38
rs748508357 Chr.17: 56835256-56835256 on Build GRCh38
rs777787526 Chr.17: 56835096-56835096 on Build GRCh38
rs757482556 Chr.17: 56835104-56835104 on Build GRCh38
rs551649354 Chr.17: 56835155-56835155 on Build GRCh38
rs878925112 Chr.17: 56835167-56835167 on Build GRCh38
rs769109284 Chr.17: 56835244-56835244 on Build GRCh38
rs750649560 Chr.17: 56835330-56835330 on Build GRCh38
rs774176060 Chr.17: 56835262-56835262 on Build GRCh38
rs148908859 Chr.17: 56835145-56835145 on Build GRCh38
rs377695357 Chr.17: 56835301-56835301 on Build GRCh38
rs934221669 Chr.17: 56835209-56835209 on Build GRCh38
rs146765015 Chr.17: 56835050-56835050 on Build GRCh38
rs765405428 Chr.17: 56835324-56835324 on Build GRCh38
rs1029757442 Chr.17: 56835474-56835474 on Build GRCh38
rs1022656591 Chr.17: 56835344-56835344 on Build GRCh38
rs969844522 Chr.17: 56835095-56835095 on Build GRCh38
rs745990961 Chr.17: 56835111-56835111 on Build GRCh38
rs776001127 Chr.17: 56835135-56835135 on Build GRCh38
rs767517741 Chr.17: 56835254-56835254 on Build GRCh38
rs769070028 Chr.17: 56835146-56835146 on Build GRCh38
rs371972116 Chr.17: 56835282-56835282 on Build GRCh38
rs759046836 Chr.17: 56835263-56835263 on Build GRCh38
rs767519633 Chr.17: 56835162-56835162 on Build GRCh38
rs916130603 Chr.17: 56835411-56835411 on Build GRCh38
rs993485497 Chr.17: 56835399-56835399 on Build GRCh38

View Full Product Details