Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58083252-58083755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL2
Entrez Gene ID
140735
Gene Name
dynein light chain LC8-type 2
Gene Aliases
DNCL1B, Dlc2, RSPH22
UniGene
Hs.720595
Gene Chromosome Location
Chr.17: 58083419-58090257 on Build GRCh38
Transcripts
NM_080677.2

SNP Detail

SNP ID Chromosome Location
rs946543887 Chr.17: 58083475-58083475 on Build GRCh38
rs530108220 Chr.17: 58083303-58083303 on Build GRCh38
rs1027784432 Chr.17: 58083411-58083411 on Build GRCh38
rs867070294 Chr.17: 58083490-58083490 on Build GRCh38
rs376668096 Chr.17: 58083459-58083462 on Build GRCh38
rs946576490 Chr.17: 58083710-58083710 on Build GRCh38
rs935187900 Chr.17: 58083310-58083310 on Build GRCh38
rs919861811 Chr.17: 58083467-58083491 on Build GRCh38
rs994091605 Chr.17: 58083407-58083407 on Build GRCh38
rs1035708846 Chr.17: 58083377-58083377 on Build GRCh38
rs898075026 Chr.17: 58083549-58083549 on Build GRCh38
rs1056446210 Chr.17: 58083545-58083545 on Build GRCh38
rs1028316006 Chr.17: 58083559-58083559 on Build GRCh38
rs548340955 Chr.17: 58083321-58083321 on Build GRCh38
rs376962847 Chr.17: 58083672-58083672 on Build GRCh38
rs556028195 Chr.17: 58083446-58083446 on Build GRCh38
rs541402228 Chr.17: 58083571-58083571 on Build GRCh38
rs980544352 Chr.17: 58083729-58083729 on Build GRCh38
rs2533003 Chr.17: 58083481-58083481 on Build GRCh38
rs905268272 Chr.17: 58083500-58083500 on Build GRCh38
rs976430099 Chr.17: 58083452-58083452 on Build GRCh38
rs1029290560 Chr.17: 58083606-58083606 on Build GRCh38
rs1003496690 Chr.17: 58083371-58083371 on Build GRCh38
rs537706036 Chr.17: 58083437-58083437 on Build GRCh38
rs572074764 Chr.17: 58083646-58083646 on Build GRCh38
rs910974596 Chr.17: 58083422-58083422 on Build GRCh38
rs60600936 Chr.17: 58083478-58083479 on Build GRCh38
rs962292329 Chr.17: 58083380-58083380 on Build GRCh38
rs939368025 Chr.17: 58083508-58083508 on Build GRCh38
rs553520549 Chr.17: 58083575-58083575 on Build GRCh38
rs370082329 Chr.17: 58083706-58083706 on Build GRCh38
rs549244103 Chr.17: 58083421-58083421 on Build GRCh38
rs929876042 Chr.17: 58083468-58083479 on Build GRCh38
rs993731532 Chr.17: 58083550-58083550 on Build GRCh38
rs905235711 Chr.17: 58083353-58083353 on Build GRCh38
rs558688714 Chr.17: 58083418-58083418 on Build GRCh38
rs59204964 Chr.17: 58083344-58083344 on Build GRCh38
rs963758759 Chr.17: 58083578-58083578 on Build GRCh38
rs915156511 Chr.17: 58083694-58083694 on Build GRCh38
rs1052244547 Chr.17: 58083320-58083320 on Build GRCh38
rs770639111 Chr.17: 58083669-58083669 on Build GRCh38
rs976840193 Chr.17: 58083604-58083604 on Build GRCh38
rs935237513 Chr.17: 58083457-58083464 on Build GRCh38
rs1045052030 Chr.17: 58083349-58083349 on Build GRCh38
rs956844999 Chr.17: 58083607-58083607 on Build GRCh38
rs963732922 Chr.17: 58083425-58083426 on Build GRCh38
rs570913538 Chr.17: 58083458-58083459 on Build GRCh38
rs887987561 Chr.17: 58083566-58083567 on Build GRCh38
rs566936965 Chr.17: 58083329-58083329 on Build GRCh38
rs988110129 Chr.17: 58083661-58083661 on Build GRCh38
rs922522397 Chr.17: 58083454-58083454 on Build GRCh38

View Full Product Details