Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58089058-58089561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL2
Entrez Gene ID
140735
Gene Name
dynein light chain LC8-type 2
Gene Aliases
DNCL1B, Dlc2, RSPH22
UniGene
Hs.720595
Gene Chromosome Location
Chr.17: 58083419-58090257 on Build GRCh38
Transcripts
NM_080677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4402955 c.180A>C p.R60R Chr.17: 58089189-58089189 on Build GRCh38
4188643 c.270G>C p.*90Y Chr.17: 58089279-58089279 on Build GRCh38
4912587 c.219C>T p.F73F Chr.17: 58089228-58089228 on Build GRCh38
4800809 c.188G>T p.G63V Chr.17: 58089197-58089197 on Build GRCh38
981717 c.205G>A p.E69K Chr.17: 58089214-58089214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780990885 Chr.17: 58089094-58089094 on Build GRCh38
rs374144805 Chr.17: 58089299-58089299 on Build GRCh38
rs892392359 Chr.17: 58089369-58089369 on Build GRCh38
rs760693436 Chr.17: 58089305-58089305 on Build GRCh38
rs765952435 Chr.17: 58089178-58089178 on Build GRCh38
rs1006029116 Chr.17: 58089374-58089374 on Build GRCh38
rs201010065 Chr.17: 58089134-58089134 on Build GRCh38
rs369484056 Chr.17: 58089321-58089321 on Build GRCh38
rs763089795 Chr.17: 58089302-58089303 on Build GRCh38
rs1014766893 Chr.17: 58089410-58089410 on Build GRCh38
rs752199536 Chr.17: 58089326-58089326 on Build GRCh38
rs1050682461 Chr.17: 58089345-58089346 on Build GRCh38
rs953198842 Chr.17: 58089450-58089450 on Build GRCh38
rs746158448 Chr.17: 58089294-58089294 on Build GRCh38
rs371611185 Chr.17: 58089317-58089317 on Build GRCh38
rs749033895 Chr.17: 58089261-58089261 on Build GRCh38
rs756035674 Chr.17: 58089225-58089225 on Build GRCh38
rs190895072 Chr.17: 58089489-58089489 on Build GRCh38
rs16942662 Chr.17: 58089138-58089138 on Build GRCh38
rs997554375 Chr.17: 58089415-58089415 on Build GRCh38
rs372402569 Chr.17: 58089412-58089412 on Build GRCh38
rs763296342 Chr.17: 58089131-58089131 on Build GRCh38
rs546944674 Chr.17: 58089411-58089411 on Build GRCh38
rs372037785 Chr.17: 58089358-58089358 on Build GRCh38
rs139580700 Chr.17: 58089219-58089219 on Build GRCh38
rs769450807 Chr.17: 58089102-58089102 on Build GRCh38
rs1021406474 Chr.17: 58089471-58089471 on Build GRCh38
rs777131527 Chr.17: 58089314-58089314 on Build GRCh38
rs571670186 Chr.17: 58089425-58089425 on Build GRCh38
rs1002012086 Chr.17: 58089494-58089494 on Build GRCh38
rs924205898 Chr.17: 58089280-58089280 on Build GRCh38
rs754525149 Chr.17: 58089091-58089091 on Build GRCh38
rs1034197980 Chr.17: 58089506-58089506 on Build GRCh38
rs187859298 Chr.17: 58089468-58089468 on Build GRCh38
rs981285563 Chr.17: 58089097-58089097 on Build GRCh38
rs773178570 Chr.17: 58089104-58089104 on Build GRCh38
rs754436188 Chr.17: 58089208-58089208 on Build GRCh38
rs535023430 Chr.17: 58089328-58089328 on Build GRCh38
rs754153288 Chr.17: 58089168-58089168 on Build GRCh38
rs777872108 Chr.17: 58089256-58089256 on Build GRCh38
rs370390753 Chr.17: 58089106-58089106 on Build GRCh38
rs762183088 Chr.17: 58089174-58089174 on Build GRCh38
rs879100665 Chr.17: 58089112-58089112 on Build GRCh38
rs760617745 Chr.17: 58089401-58089401 on Build GRCh38
rs564557407 Chr.17: 58089088-58089088 on Build GRCh38
rs111433106 Chr.17: 58089101-58089101 on Build GRCh38
rs777786474 Chr.17: 58089331-58089331 on Build GRCh38
rs748177953 Chr.17: 58089098-58089098 on Build GRCh38
rs200481800 Chr.17: 58089286-58089286 on Build GRCh38
rs753864633 Chr.17: 58089445-58089445 on Build GRCh38
rs775840589 Chr.17: 58089129-58089129 on Build GRCh38
rs146307541 Chr.17: 58089213-58089213 on Build GRCh38
rs751151868 Chr.17: 58089319-58089319 on Build GRCh38
rs140644850 Chr.17: 58089090-58089090 on Build GRCh38
rs774726730 Chr.17: 58089110-58089110 on Build GRCh38
rs749122805 Chr.17: 58089105-58089105 on Build GRCh38
rs769093945 Chr.17: 58089308-58089308 on Build GRCh38
rs145820835 Chr.17: 58089448-58089448 on Build GRCh38
rs985883923 Chr.17: 58089465-58089465 on Build GRCh38
rs767103775 Chr.17: 58089324-58089324 on Build GRCh38
rs909411221 Chr.17: 58089306-58089306 on Build GRCh38
rs368513542 Chr.17: 58089303-58089303 on Build GRCh38
rs201029806 Chr.17: 58089115-58089115 on Build GRCh38
rs767701702 Chr.17: 58089126-58089126 on Build GRCh38
rs764117085 Chr.17: 58089429-58089429 on Build GRCh38
rs751283300 Chr.17: 58089180-58089180 on Build GRCh38
rs762164785 Chr.17: 58089315-58089315 on Build GRCh38
rs377760636 Chr.17: 58089285-58089285 on Build GRCh38

View Full Product Details