Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76076893-76077428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALR2
Entrez Gene ID
8811
Gene Name
galanin receptor 2
Gene Aliases
GAL2-R, GALNR2, GALR-2
UniGene
Hs.666366
Gene Chromosome Location
Chr.17: 76070822-76077541 on Build GRCh38
Transcripts
NM_003857.3 XM_011525427.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4885256 c.919T>C p.C307R Chr.17: 76077186-76077186 on Build GRCh38
6669173 c.1052C>A p.A351D Chr.17: 76077319-76077319 on Build GRCh38
4845479 c.765C>T p.L255L Chr.17: 76077032-76077032 on Build GRCh38
4757488 c.937C>T p.R313C Chr.17: 76077204-76077204 on Build GRCh38
4686359 c.949C>T p.R317* Chr.17: 76077216-76077216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055594226 Chr.17: 76076976-76076976 on Build GRCh38
rs763493769 Chr.17: 76077167-76077167 on Build GRCh38
rs750365802 Chr.17: 76077138-76077138 on Build GRCh38
rs752254609 Chr.17: 76077041-76077041 on Build GRCh38
rs896449074 Chr.17: 76077296-76077296 on Build GRCh38
rs140309278 Chr.17: 76077080-76077080 on Build GRCh38
rs772601523 Chr.17: 76077316-76077316 on Build GRCh38
rs772365230 Chr.17: 76077105-76077105 on Build GRCh38
rs984991461 Chr.17: 76077406-76077406 on Build GRCh38
rs759979593 Chr.17: 76077100-76077100 on Build GRCh38
rs778173749 Chr.17: 76077260-76077260 on Build GRCh38
rs565014665 Chr.17: 76077003-76077003 on Build GRCh38
rs765436798 Chr.17: 76077349-76077349 on Build GRCh38
rs761738206 Chr.17: 76076928-76076928 on Build GRCh38
rs373366919 Chr.17: 76076974-76076974 on Build GRCh38
rs752529873 Chr.17: 76077345-76077345 on Build GRCh38
rs1056489857 Chr.17: 76077292-76077292 on Build GRCh38
rs76890663 Chr.17: 76076943-76076943 on Build GRCh38
rs550086254 Chr.17: 76077061-76077061 on Build GRCh38
rs944960201 Chr.17: 76077191-76077191 on Build GRCh38
rs66906990 Chr.17: 76077375-76077384 on Build GRCh38
rs756409862 Chr.17: 76077154-76077154 on Build GRCh38
rs113884061 Chr.17: 76077143-76077143 on Build GRCh38
rs554624828 Chr.17: 76077347-76077347 on Build GRCh38
rs1011340458 Chr.17: 76077030-76077030 on Build GRCh38
rs892771472 Chr.17: 76077021-76077021 on Build GRCh38
rs964472392 Chr.17: 76077062-76077062 on Build GRCh38
rs751413774 Chr.17: 76077259-76077259 on Build GRCh38
rs752265361 Chr.17: 76076964-76076964 on Build GRCh38
rs868811539 Chr.17: 76077249-76077249 on Build GRCh38
rs769482734 Chr.17: 76077310-76077310 on Build GRCh38
rs145000591 Chr.17: 76077149-76077149 on Build GRCh38
rs190734697 Chr.17: 76076988-76076988 on Build GRCh38
rs767470690 Chr.17: 76077339-76077339 on Build GRCh38
rs974891508 Chr.17: 76076927-76076936 on Build GRCh38
rs67693166 Chr.17: 76077367-76077376 on Build GRCh38
rs774782865 Chr.17: 76077025-76077025 on Build GRCh38
rs746414961 Chr.17: 76077204-76077204 on Build GRCh38
rs778760826 Chr.17: 76077060-76077060 on Build GRCh38
rs780073845 Chr.17: 76077386-76077386 on Build GRCh38
rs778084055 Chr.17: 76077165-76077165 on Build GRCh38
rs935605222 Chr.17: 76077317-76077317 on Build GRCh38
rs919531875 Chr.17: 76077373-76077373 on Build GRCh38
rs140278862 Chr.17: 76076947-76076947 on Build GRCh38
rs374970890 Chr.17: 76077335-76077335 on Build GRCh38
rs997231236 Chr.17: 76077408-76077408 on Build GRCh38
rs748288647 Chr.17: 76077006-76077006 on Build GRCh38
rs61745847 Chr.17: 76077013-76077013 on Build GRCh38
rs768753307 Chr.17: 76076924-76076924 on Build GRCh38
rs760968262 Chr.17: 76077219-76077219 on Build GRCh38
rs766898366 Chr.17: 76076946-76076946 on Build GRCh38
rs1038690900 Chr.17: 76077359-76077359 on Build GRCh38
rs771325151 Chr.17: 76077199-76077199 on Build GRCh38
rs566999194 Chr.17: 76077238-76077238 on Build GRCh38
rs371320366 Chr.17: 76077251-76077251 on Build GRCh38
rs749314723 Chr.17: 76077083-76077083 on Build GRCh38
rs771579313 Chr.17: 76077094-76077094 on Build GRCh38
rs538882315 Chr.17: 76077229-76077229 on Build GRCh38
rs377747977 Chr.17: 76077190-76077190 on Build GRCh38
rs766251513 Chr.17: 76077324-76077324 on Build GRCh38
rs755734479 Chr.17: 76077043-76077043 on Build GRCh38
rs924074444 Chr.17: 76077315-76077315 on Build GRCh38
rs768316504 Chr.17: 76077223-76077223 on Build GRCh38
rs767223522 Chr.17: 76077033-76077033 on Build GRCh38
rs759786129 Chr.17: 76077359-76077359 on Build GRCh38
rs574791995 Chr.17: 76077355-76077355 on Build GRCh38
rs774673040 Chr.17: 76077098-76077098 on Build GRCh38
rs765248886 Chr.17: 76077136-76077136 on Build GRCh38
rs758460927 Chr.17: 76077375-76077375 on Build GRCh38
rs766272954 Chr.17: 76077245-76077245 on Build GRCh38
rs564505715 Chr.17: 76077306-76077306 on Build GRCh38
rs753355530 Chr.17: 76077129-76077129 on Build GRCh38
rs1011684168 Chr.17: 76077357-76077357 on Build GRCh38
rs757482821 Chr.17: 76077270-76077270 on Build GRCh38
rs755533463 Chr.17: 76076965-76076965 on Build GRCh38
rs764700367 Chr.17: 76077234-76077234 on Build GRCh38
rs201878382 Chr.17: 76077015-76077015 on Build GRCh38
rs199983830 Chr.17: 76076971-76076971 on Build GRCh38
rs747561393 Chr.17: 76077389-76077389 on Build GRCh38
rs769046796 Chr.17: 76077294-76077294 on Build GRCh38
rs949464130 Chr.17: 76077302-76077302 on Build GRCh38
rs750995213 Chr.17: 76077241-76077241 on Build GRCh38
rs758085909 Chr.17: 76077065-76077065 on Build GRCh38
rs569895534 Chr.17: 76077145-76077145 on Build GRCh38
rs774106063 Chr.17: 76076921-76076921 on Build GRCh38
rs375951258 Chr.17: 76077266-76077266 on Build GRCh38
rs763666379 Chr.17: 76077125-76077125 on Build GRCh38
rs763611858 Chr.17: 76077019-76077019 on Build GRCh38
rs749267257 Chr.17: 76076973-76076973 on Build GRCh38
rs753337103 Chr.17: 76077051-76077051 on Build GRCh38
rs201137943 Chr.17: 76077044-76077044 on Build GRCh38
rs916768054 Chr.17: 76076978-76076978 on Build GRCh38
rs779021741 Chr.17: 76077202-76077202 on Build GRCh38
rs961060355 Chr.17: 76077364-76077364 on Build GRCh38
rs990822351 Chr.17: 76077276-76077276 on Build GRCh38
rs776955872 Chr.17: 76077300-76077300 on Build GRCh38
rs762468562 Chr.17: 76077320-76077320 on Build GRCh38
rs767013439 Chr.17: 76076963-76076963 on Build GRCh38
rs369479330 Chr.17: 76076962-76076962 on Build GRCh38
rs774844174 Chr.17: 76077333-76077342 on Build GRCh38
rs773855880 Chr.17: 76077096-76077097 on Build GRCh38
rs747308090 Chr.17: 76076922-76076922 on Build GRCh38
rs780558089 Chr.17: 76077279-76077279 on Build GRCh38
rs762093548 Chr.17: 76076913-76076913 on Build GRCh38
rs8192514 Chr.17: 76077305-76077305 on Build GRCh38
rs774391668 Chr.17: 76077331-76077331 on Build GRCh38
rs375198936 Chr.17: 76076942-76076942 on Build GRCh38
rs902808127 Chr.17: 76076994-76076994 on Build GRCh38
rs761421630 Chr.17: 76077230-76077230 on Build GRCh38
rs545102045 Chr.17: 76076993-76076993 on Build GRCh38
rs758838164 Chr.17: 76077277-76077277 on Build GRCh38
rs776772676 Chr.17: 76076927-76076927 on Build GRCh38
rs775789398 Chr.17: 76077205-76077205 on Build GRCh38
rs749105845 Chr.17: 76076915-76076915 on Build GRCh38
rs766989983 Chr.17: 76077020-76077020 on Build GRCh38
rs761260942 Chr.17: 76077132-76077132 on Build GRCh38
rs750487678 Chr.17: 76077351-76077351 on Build GRCh38
rs778742716 Chr.17: 76076986-76076986 on Build GRCh38
rs760381828 Chr.17: 76077114-76077114 on Build GRCh38
rs938067672 Chr.17: 76077275-76077275 on Build GRCh38
rs770674605 Chr.17: 76076918-76076918 on Build GRCh38
rs747403278 Chr.17: 76077281-76077281 on Build GRCh38
rs751959289 Chr.17: 76076955-76076955 on Build GRCh38
rs779866373 Chr.17: 76077070-76077070 on Build GRCh38
rs759763601 Chr.17: 76077028-76077028 on Build GRCh38
rs866770990 Chr.17: 76077151-76077151 on Build GRCh38
rs77852923 Chr.17: 76077274-76077274 on Build GRCh38
rs775905475 Chr.17: 76077111-76077111 on Build GRCh38
rs930867812 Chr.17: 76077401-76077401 on Build GRCh38
rs778921138 Chr.17: 76077272-76077272 on Build GRCh38
rs867849473 Chr.17: 76077374-76077384 on Build GRCh38
rs749403037 Chr.17: 76077189-76077189 on Build GRCh38
rs903757787 Chr.17: 76077325-76077325 on Build GRCh38
rs112772908 Chr.17: 76077056-76077056 on Build GRCh38

View Full Product Details