Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76077287-76077779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALR2
Entrez Gene ID
8811
Gene Name
galanin receptor 2
Gene Aliases
GAL2-R, GALNR2, GALR-2
UniGene
Hs.666366
Gene Chromosome Location
Chr.17: 76070822-76077541 on Build GRCh38
Transcripts
NM_003857.3 XM_011525427.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669173 c.1052C>A p.A351D Chr.17: 76077319-76077319 on Build GRCh38
5972423 c.1092C>G p.L364L Chr.17: 76077359-76077359 on Build GRCh38
4069798 c.1033A>G p.M345V Chr.17: 76077300-76077300 on Build GRCh38
4069797 c.1021G>A p.D341N Chr.17: 76077288-76077288 on Build GRCh38
7235490 c.1123C>A p.P375T Chr.17: 76077390-76077390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541139574 Chr.17: 76077753-76077753 on Build GRCh38
rs1008005289 Chr.17: 76077588-76077588 on Build GRCh38
rs867008969 Chr.17: 76077699-76077699 on Build GRCh38
rs772601523 Chr.17: 76077316-76077316 on Build GRCh38
rs182022064 Chr.17: 76077492-76077492 on Build GRCh38
rs73361860 Chr.17: 76077633-76077633 on Build GRCh38
rs984991461 Chr.17: 76077406-76077406 on Build GRCh38
rs747561393 Chr.17: 76077389-76077389 on Build GRCh38
rs569282575 Chr.17: 76077488-76077488 on Build GRCh38
rs765436798 Chr.17: 76077349-76077349 on Build GRCh38
rs1027039834 Chr.17: 76077695-76077695 on Build GRCh38
rs775283793 Chr.17: 76077737-76077737 on Build GRCh38
rs755462916 Chr.17: 76077412-76077412 on Build GRCh38
rs752529873 Chr.17: 76077345-76077345 on Build GRCh38
rs1039117193 Chr.17: 76077749-76077749 on Build GRCh38
rs950505891 Chr.17: 76077487-76077487 on Build GRCh38
rs66906990 Chr.17: 76077375-76077384 on Build GRCh38
rs141120606 Chr.17: 76077565-76077565 on Build GRCh38
rs576295277 Chr.17: 76077546-76077546 on Build GRCh38
rs554624828 Chr.17: 76077347-76077347 on Build GRCh38
rs961060355 Chr.17: 76077364-76077364 on Build GRCh38
rs972392856 Chr.17: 76077658-76077658 on Build GRCh38
rs762468562 Chr.17: 76077320-76077320 on Build GRCh38
rs769482734 Chr.17: 76077310-76077310 on Build GRCh38
rs774844174 Chr.17: 76077333-76077342 on Build GRCh38
rs982139731 Chr.17: 76077562-76077562 on Build GRCh38
rs752336513 Chr.17: 76077717-76077717 on Build GRCh38
rs748478431 Chr.17: 76077430-76077430 on Build GRCh38
rs915520778 Chr.17: 76077730-76077730 on Build GRCh38
rs970789151 Chr.17: 76077561-76077561 on Build GRCh38
rs572005385 Chr.17: 76077722-76077722 on Build GRCh38
rs767470690 Chr.17: 76077339-76077339 on Build GRCh38
rs67693166 Chr.17: 76077367-76077376 on Build GRCh38
rs745808377 Chr.17: 76077541-76077541 on Build GRCh38
rs764800258 Chr.17: 76077680-76077680 on Build GRCh38
rs774391668 Chr.17: 76077331-76077331 on Build GRCh38
rs780073845 Chr.17: 76077386-76077386 on Build GRCh38
rs935605222 Chr.17: 76077317-76077317 on Build GRCh38
rs919531875 Chr.17: 76077373-76077373 on Build GRCh38
rs374970890 Chr.17: 76077335-76077335 on Build GRCh38
rs35980071 Chr.17: 76077604-76077605 on Build GRCh38
rs997231236 Chr.17: 76077408-76077408 on Build GRCh38
rs991262961 Chr.17: 76077554-76077554 on Build GRCh38
rs926893262 Chr.17: 76077750-76077750 on Build GRCh38
rs989647633 Chr.17: 76077721-76077721 on Build GRCh38
rs1038690900 Chr.17: 76077359-76077359 on Build GRCh38
rs961092681 Chr.17: 76077653-76077653 on Build GRCh38
rs903781888 Chr.17: 76077510-76077510 on Build GRCh38
rs766251513 Chr.17: 76077324-76077324 on Build GRCh38
rs558185853 Chr.17: 76077719-76077719 on Build GRCh38
rs1001156691 Chr.17: 76077516-76077516 on Build GRCh38
rs750487678 Chr.17: 76077351-76077351 on Build GRCh38
rs924074444 Chr.17: 76077315-76077315 on Build GRCh38
rs781602615 Chr.17: 76077415-76077415 on Build GRCh38
rs963306796 Chr.17: 76077553-76077553 on Build GRCh38
rs766032684 Chr.17: 76077439-76077439 on Build GRCh38
rs916552219 Chr.17: 76077556-76077556 on Build GRCh38
rs759786129 Chr.17: 76077359-76077359 on Build GRCh38
rs574791995 Chr.17: 76077355-76077355 on Build GRCh38
rs758460927 Chr.17: 76077375-76077375 on Build GRCh38
rs76669670 Chr.17: 76077490-76077490 on Build GRCh38
rs930867812 Chr.17: 76077401-76077401 on Build GRCh38
rs867849473 Chr.17: 76077374-76077384 on Build GRCh38
rs1011684168 Chr.17: 76077357-76077357 on Build GRCh38
rs769344371 Chr.17: 76077446-76077446 on Build GRCh38
rs903757787 Chr.17: 76077325-76077325 on Build GRCh38

View Full Product Details