Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76083282-76083780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC7
Entrez Gene ID
23265
Gene Name
exocyst complex component 7
Gene Aliases
2-5-3p, EX070, EXO70, EXOC1, Exo70p, YJL085W
UniGene
Hs.514496
Gene Chromosome Location
Chr.17: 76080993-76104221 on Build GRCh38
Transcripts
NM_001013839.3 NM_001145297.3 NM_001145298.3 NM_001145299.3 NM_001282313.1 NM_015219.4 XM_006721786.3 XM_006721787.3 XM_006721788.3 XM_006721789.3 XM_011524571.2 XM_017024397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069807 c.1963G>A p.V655M Chr.17: 76083740-76083740 on Build GRCh38
3522474 c.2032C>T p.R678C Chr.17: 76083671-76083671 on Build GRCh38
4069805 c.2019C>T p.G673G Chr.17: 76083684-76083684 on Build GRCh38
5440048 c.2021A>T p.D674V Chr.17: 76083682-76083682 on Build GRCh38
1480034 c.2054G>A p.*685* Chr.17: 76083649-76083649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775593717 Chr.17: 76083343-76083343 on Build GRCh38
rs1008985503 Chr.17: 76083336-76083336 on Build GRCh38
rs555388505 Chr.17: 76083629-76083629 on Build GRCh38
rs201181528 Chr.17: 76083677-76083677 on Build GRCh38
rs376255016 Chr.17: 76083674-76083674 on Build GRCh38
rs773148479 Chr.17: 76083663-76083663 on Build GRCh38
rs770640366 Chr.17: 76083734-76083734 on Build GRCh38
rs765710310 Chr.17: 76083683-76083683 on Build GRCh38
rs554118658 Chr.17: 76083348-76083348 on Build GRCh38
rs924213537 Chr.17: 76083392-76083392 on Build GRCh38
rs568906232 Chr.17: 76083374-76083374 on Build GRCh38
rs748167217 Chr.17: 76083636-76083636 on Build GRCh38
rs754676259 Chr.17: 76083701-76083701 on Build GRCh38
rs922695386 Chr.17: 76083483-76083483 on Build GRCh38
rs187050978 Chr.17: 76083759-76083759 on Build GRCh38
rs946798864 Chr.17: 76083569-76083569 on Build GRCh38
rs181224243 Chr.17: 76083741-76083741 on Build GRCh38
rs752235400 Chr.17: 76083706-76083706 on Build GRCh38
rs377309793 Chr.17: 76083632-76083632 on Build GRCh38
rs11539426 Chr.17: 76083478-76083478 on Build GRCh38
rs761020236 Chr.17: 76083702-76083702 on Build GRCh38
rs559705384 Chr.17: 76083557-76083557 on Build GRCh38
rs373384194 Chr.17: 76083666-76083666 on Build GRCh38
rs942878710 Chr.17: 76083438-76083438 on Build GRCh38
rs756162747 Chr.17: 76083710-76083710 on Build GRCh38
rs765657201 Chr.17: 76083601-76083601 on Build GRCh38
rs537837865 Chr.17: 76083387-76083387 on Build GRCh38
rs1038292097 Chr.17: 76083527-76083527 on Build GRCh38
rs1027345860 Chr.17: 76083412-76083412 on Build GRCh38
rs146250593 Chr.17: 76083721-76083721 on Build GRCh38
rs751176361 Chr.17: 76083699-76083699 on Build GRCh38
rs763250769 Chr.17: 76083691-76083691 on Build GRCh38
rs1012740720 Chr.17: 76083543-76083543 on Build GRCh38
rs200397075 Chr.17: 76083669-76083669 on Build GRCh38
rs980628255 Chr.17: 76083466-76083466 on Build GRCh38
rs887034571 Chr.17: 76083502-76083502 on Build GRCh38
rs373865233 Chr.17: 76083501-76083501 on Build GRCh38
rs778698782 Chr.17: 76083740-76083740 on Build GRCh38
rs557427223 Chr.17: 76083409-76083409 on Build GRCh38
rs188787726 Chr.17: 76083327-76083327 on Build GRCh38
rs927905024 Chr.17: 76083487-76083487 on Build GRCh38
rs200713268 Chr.17: 76083720-76083720 on Build GRCh38
rs915443076 Chr.17: 76083437-76083437 on Build GRCh38
rs766708575 Chr.17: 76083693-76083693 on Build GRCh38
rs147177976 Chr.17: 76083403-76083403 on Build GRCh38
rs2665986 Chr.17: 76083757-76083757 on Build GRCh38
rs960622585 Chr.17: 76083500-76083500 on Build GRCh38
rs563676881 Chr.17: 76083465-76083465 on Build GRCh38
rs776687293 Chr.17: 76083680-76083680 on Build GRCh38
rs969652286 Chr.17: 76083430-76083430 on Build GRCh38
rs997013220 Chr.17: 76083319-76083319 on Build GRCh38
rs543689345 Chr.17: 76083453-76083453 on Build GRCh38
rs368736156 Chr.17: 76083651-76083651 on Build GRCh38
rs1002454688 Chr.17: 76083729-76083729 on Build GRCh38
rs1024488231 Chr.17: 76083587-76083587 on Build GRCh38
rs528551097 Chr.17: 76083670-76083670 on Build GRCh38
rs994517333 Chr.17: 76083407-76083407 on Build GRCh38
rs577361022 Chr.17: 76083496-76083496 on Build GRCh38
rs758117130 Chr.17: 76083610-76083610 on Build GRCh38
rs779604859 Chr.17: 76083611-76083611 on Build GRCh38
rs961787413 Chr.17: 76083378-76083378 on Build GRCh38
rs754549320 Chr.17: 76083628-76083628 on Build GRCh38
rs746624910 Chr.17: 76083627-76083627 on Build GRCh38
rs111929820 Chr.17: 76083709-76083709 on Build GRCh38
rs570901069 Chr.17: 76083521-76083521 on Build GRCh38
rs1005507723 Chr.17: 76083520-76083520 on Build GRCh38
rs1051820807 Chr.17: 76083322-76083322 on Build GRCh38
rs750903396 Chr.17: 76083684-76083684 on Build GRCh38
rs750738193 Chr.17: 76083604-76083604 on Build GRCh38
rs977085276 Chr.17: 76083388-76083388 on Build GRCh38
rs760890089 Chr.17: 76083671-76083671 on Build GRCh38
rs73996111 Chr.17: 76083532-76083532 on Build GRCh38

View Full Product Details