Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76465800-76466296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AANAT
Entrez Gene ID
15
Gene Name
aralkylamine N-acetyltransferase
Gene Aliases
DSPS, SNAT
UniGene
Hs.431417
Gene Chromosome Location
Chr.17: 76453351-76470797 on Build GRCh38
Transcripts
NM_001166579.1 XM_017024259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139846 c.27G>A p.Q9Q Chr.17: 76466199-76466199 on Build GRCh38
6371817 c.31A>G p.R11G Chr.17: 76466203-76466203 on Build GRCh38
3937584 c.60+1G>A p.? Chr.17: 76466233-76466233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044023755 Chr.17: 76465845-76465845 on Build GRCh38
rs557225975 Chr.17: 76465889-76465889 on Build GRCh38
rs979384549 Chr.17: 76465919-76465919 on Build GRCh38
rs751265786 Chr.17: 76466117-76466117 on Build GRCh38
rs868108781 Chr.17: 76466222-76466222 on Build GRCh38
rs995801228 Chr.17: 76465918-76465918 on Build GRCh38
rs879737352 Chr.17: 76465845-76465845 on Build GRCh38
rs931302908 Chr.17: 76466239-76466239 on Build GRCh38
rs979414991 Chr.17: 76466106-76466106 on Build GRCh38
rs750686267 Chr.17: 76466175-76466175 on Build GRCh38
rs568027872 Chr.17: 76465833-76465833 on Build GRCh38
rs896262640 Chr.17: 76465944-76465944 on Build GRCh38
rs368031088 Chr.17: 76466135-76466135 on Build GRCh38
rs995436462 Chr.17: 76465993-76465993 on Build GRCh38
rs1004647274 Chr.17: 76466064-76466064 on Build GRCh38
rs766433496 Chr.17: 76466195-76466195 on Build GRCh38
rs747870485 Chr.17: 76466021-76466021 on Build GRCh38
rs946680542 Chr.17: 76465843-76465843 on Build GRCh38
rs887020317 Chr.17: 76466038-76466038 on Build GRCh38
rs968112739 Chr.17: 76466080-76466080 on Build GRCh38
rs562569116 Chr.17: 76465924-76465926 on Build GRCh38
rs904998462 Chr.17: 76465971-76465971 on Build GRCh38
rs749649675 Chr.17: 76465849-76465849 on Build GRCh38
rs142414187 Chr.17: 76465888-76465888 on Build GRCh38
rs949135655 Chr.17: 76465962-76465962 on Build GRCh38
rs145973909 Chr.17: 76465934-76465934 on Build GRCh38
rs951417525 Chr.17: 76465852-76465852 on Build GRCh38
rs982903769 Chr.17: 76465863-76465863 on Build GRCh38
rs138364794 Chr.17: 76466250-76466250 on Build GRCh38
rs960421346 Chr.17: 76465994-76465994 on Build GRCh38
rs770801692 Chr.17: 76466134-76466134 on Build GRCh38
rs1034745792 Chr.17: 76465961-76465961 on Build GRCh38
rs1037009205 Chr.17: 76465896-76465896 on Build GRCh38
rs530979426 Chr.17: 76465943-76465943 on Build GRCh38
rs934962629 Chr.17: 76465927-76465928 on Build GRCh38
rs1051189019 Chr.17: 76465860-76465860 on Build GRCh38
rs1004227570 Chr.17: 76465892-76465892 on Build GRCh38
rs1044909125 Chr.17: 76465963-76465963 on Build GRCh38
rs755961969 Chr.17: 76466168-76466169 on Build GRCh38
rs777433231 Chr.17: 76466104-76466104 on Build GRCh38
rs776669923 Chr.17: 76466171-76466171 on Build GRCh38
rs1020149382 Chr.17: 76465865-76465865 on Build GRCh38
rs921143706 Chr.17: 76465848-76465848 on Build GRCh38
rs774286433 Chr.17: 76466170-76466171 on Build GRCh38
rs975619322 Chr.17: 76465897-76465897 on Build GRCh38
rs537731492 Chr.17: 76465907-76465907 on Build GRCh38
rs868659660 Chr.17: 76465928-76465928 on Build GRCh38
rs573015333 Chr.17: 76466144-76466144 on Build GRCh38
rs913839545 Chr.17: 76465842-76465842 on Build GRCh38
rs965489443 Chr.17: 76466120-76466120 on Build GRCh38
rs58453332 Chr.17: 76466153-76466153 on Build GRCh38
rs912667955 Chr.17: 76466122-76466122 on Build GRCh38
rs763840229 Chr.17: 76466166-76466166 on Build GRCh38
rs905157109 Chr.17: 76465840-76465840 on Build GRCh38
rs906070384 Chr.17: 76465935-76465935 on Build GRCh38
rs771638625 Chr.17: 76466063-76466063 on Build GRCh38
rs906079996 Chr.17: 76466270-76466270 on Build GRCh38
rs746911016 Chr.17: 76466109-76466109 on Build GRCh38
rs114793165 Chr.17: 76466108-76466108 on Build GRCh38
rs932506609 Chr.17: 76466115-76466115 on Build GRCh38
rs1028499588 Chr.17: 76466272-76466275 on Build GRCh38
rs751660538 Chr.17: 76466203-76466203 on Build GRCh38

View Full Product Details