Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76469051-76469544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AANAT
Entrez Gene ID
15
Gene Name
aralkylamine N-acetyltransferase
Gene Aliases
DSPS, SNAT
UniGene
Hs.431417
Gene Chromosome Location
Chr.17: 76453351-76470797 on Build GRCh38
Transcripts
NM_001088.2 NM_001166579.1 NR_110548.1 XM_017024259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589272 c.255C>T p.F85F Chr.17: 76469264-76469264 on Build GRCh38
4547854 c.297G>A p.W99* Chr.17: 76469306-76469306 on Build GRCh38
6589274 c.217T>C p.F73L Chr.17: 76469226-76469226 on Build GRCh38
4271563 c.375G>A p.L125L Chr.17: 76469249-76469249 on Build GRCh38
4517495 c.219_220CC>TT p.(=) Chr.17: 76469228-76469229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763629424 Chr.17: 76469186-76469186 on Build GRCh38
rs769809410 Chr.17: 76469220-76469220 on Build GRCh38
rs745733534 Chr.17: 76469261-76469261 on Build GRCh38
rs763427905 Chr.17: 76469165-76469165 on Build GRCh38
rs749859006 Chr.17: 76469233-76469233 on Build GRCh38
rs868453416 Chr.17: 76469269-76469269 on Build GRCh38
rs753769409 Chr.17: 76469119-76469119 on Build GRCh38
rs201200092 Chr.17: 76469193-76469193 on Build GRCh38
rs375531754 Chr.17: 76469346-76469346 on Build GRCh38
rs892859574 Chr.17: 76469392-76469392 on Build GRCh38
rs577400794 Chr.17: 76469299-76469299 on Build GRCh38
rs781298498 Chr.17: 76469202-76469202 on Build GRCh38
rs753396157 Chr.17: 76469279-76469279 on Build GRCh38
rs199774032 Chr.17: 76469343-76469343 on Build GRCh38
rs761343877 Chr.17: 76469291-76469291 on Build GRCh38
rs974805191 Chr.17: 76469415-76469415 on Build GRCh38
rs924123735 Chr.17: 76469521-76469521 on Build GRCh38
rs1053289970 Chr.17: 76469391-76469391 on Build GRCh38
rs751260992 Chr.17: 76469075-76469075 on Build GRCh38
rs371535966 Chr.17: 76469265-76469265 on Build GRCh38
rs72466445 Chr.17: 76469161-76469161 on Build GRCh38
rs375853679 Chr.17: 76469294-76469294 on Build GRCh38
rs72466446 Chr.17: 76469321-76469321 on Build GRCh38
rs754020581 Chr.17: 76469365-76469365 on Build GRCh38
rs754611615 Chr.17: 76469083-76469083 on Build GRCh38
rs368578697 Chr.17: 76469204-76469204 on Build GRCh38
rs373114830 Chr.17: 76469184-76469184 on Build GRCh38
rs765284543 Chr.17: 76469292-76469292 on Build GRCh38
rs565511651 Chr.17: 76469408-76469408 on Build GRCh38
rs200643384 Chr.17: 76469104-76469104 on Build GRCh38
rs4646261 Chr.17: 76469366-76469366 on Build GRCh38
rs997468169 Chr.17: 76469431-76469431 on Build GRCh38
rs146430405 Chr.17: 76469239-76469239 on Build GRCh38
rs116395846 Chr.17: 76469393-76469393 on Build GRCh38
rs201893744 Chr.17: 76469128-76469128 on Build GRCh38
rs767079405 Chr.17: 76469092-76469092 on Build GRCh38
rs777595448 Chr.17: 76469105-76469105 on Build GRCh38
rs758216041 Chr.17: 76469201-76469201 on Build GRCh38
rs867255223 Chr.17: 76469307-76469307 on Build GRCh38
rs763395121 Chr.17: 76469264-76469264 on Build GRCh38
rs562435256 Chr.17: 76469347-76469347 on Build GRCh38
rs774692254 Chr.17: 76469174-76469174 on Build GRCh38
rs368679164 Chr.17: 76469183-76469183 on Build GRCh38
rs963722237 Chr.17: 76469339-76469339 on Build GRCh38
rs756927358 Chr.17: 76469360-76469375 on Build GRCh38
rs900157368 Chr.17: 76469425-76469425 on Build GRCh38
rs371141066 Chr.17: 76469516-76469516 on Build GRCh38
rs756948871 Chr.17: 76469300-76469300 on Build GRCh38
rs1017547104 Chr.17: 76469238-76469238 on Build GRCh38
rs755086775 Chr.17: 76469335-76469335 on Build GRCh38
rs779947252 Chr.17: 76469502-76469502 on Build GRCh38
rs776791006 Chr.17: 76469125-76469125 on Build GRCh38
rs747296425 Chr.17: 76469134-76469134 on Build GRCh38
rs374297718 Chr.17: 76469377-76469377 on Build GRCh38
rs140754096 Chr.17: 76469270-76469270 on Build GRCh38
rs777893542 Chr.17: 76469221-76469221 on Build GRCh38
rs761436653 Chr.17: 76469344-76469344 on Build GRCh38
rs372670134 Chr.17: 76469192-76469192 on Build GRCh38
rs902805003 Chr.17: 76469080-76469080 on Build GRCh38
rs746421234 Chr.17: 76469376-76469376 on Build GRCh38
rs780363316 Chr.17: 76469379-76469379 on Build GRCh38
rs201434563 Chr.17: 76469137-76469137 on Build GRCh38
rs369158061 Chr.17: 76469312-76469312 on Build GRCh38
rs58495210 Chr.17: 76469456-76469456 on Build GRCh38
rs1030246742 Chr.17: 76469439-76469439 on Build GRCh38
rs752152016 Chr.17: 76469103-76469103 on Build GRCh38
rs963457310 Chr.17: 76469127-76469127 on Build GRCh38
rs950888345 Chr.17: 76469124-76469124 on Build GRCh38
rs751373995 Chr.17: 76469351-76469351 on Build GRCh38
rs200201199 Chr.17: 76469272-76469272 on Build GRCh38
rs199858169 Chr.17: 76469167-76469167 on Build GRCh38
rs766153570 Chr.17: 76469348-76469348 on Build GRCh38
rs771430665 Chr.17: 76469237-76469237 on Build GRCh38
rs939546733 Chr.17: 76469522-76469522 on Build GRCh38
rs943410668 Chr.17: 76469162-76469162 on Build GRCh38
rs759827642 Chr.17: 76469177-76469177 on Build GRCh38
rs200603566 Chr.17: 76469336-76469336 on Build GRCh38
rs550099296 Chr.17: 76469198-76469198 on Build GRCh38
rs527869077 Chr.17: 76469427-76469427 on Build GRCh38
rs754669497 Chr.17: 76469360-76469360 on Build GRCh38
rs554635215 Chr.17: 76469289-76469289 on Build GRCh38
rs778942728 Chr.17: 76469369-76469369 on Build GRCh38
rs935471500 Chr.17: 76469389-76469389 on Build GRCh38
rs373459354 Chr.17: 76469328-76469328 on Build GRCh38
rs201479458 Chr.17: 76469325-76469325 on Build GRCh38
rs370776679 Chr.17: 76469138-76469138 on Build GRCh38
rs772443673 Chr.17: 76469256-76469256 on Build GRCh38
rs1038743904 Chr.17: 76469395-76469395 on Build GRCh38
rs749382858 Chr.17: 76469311-76469311 on Build GRCh38
rs557551257 Chr.17: 76469295-76469295 on Build GRCh38
rs1016778198 Chr.17: 76469505-76469505 on Build GRCh38
rs1052868789 Chr.17: 76469218-76469218 on Build GRCh38
rs753635704 Chr.17: 76469270-76469270 on Build GRCh38
rs767091245 Chr.17: 76469178-76469178 on Build GRCh38
rs201338517 Chr.17: 76469350-76469350 on Build GRCh38
rs759917807 Chr.17: 76469253-76469253 on Build GRCh38

View Full Product Details