Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76528202-76528724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYGB
Entrez Gene ID
114757
Gene Name
cytoglobin
Gene Aliases
HGB, STAP
UniGene
Hs.95120
Gene Chromosome Location
Chr.17: 76527348-76557692 on Build GRCh38
Transcripts
NM_134268.4 XM_005257005.2 XM_017024116.1 NR_033357.1

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
UniGene
Hs.634380
Gene Chromosome Location
Chr.17: 76527586-76555339 on Build GRCh38
Transcripts
NM_134268.4 XM_005257005.2 XM_017024116.1 NR_033357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984723 c.570G>A p.P190P Chr.17: 76528581-76528581 on Build GRCh38
6538836 c.572A>C p.*191S Chr.17: 76528579-76528579 on Build GRCh38
5665811 c.569C>T p.P190L Chr.17: 76528582-76528582 on Build GRCh38
3717610 c.567G>T p.G189G Chr.17: 76528584-76528584 on Build GRCh38
4552656 c.565G>A p.G189R Chr.17: 76528586-76528586 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1033467068 Chr.17: 76528249-76528249 on Build GRCh38
rs751979950 Chr.17: 76528573-76528573 on Build GRCh38
rs868605420 Chr.17: 76528604-76528604 on Build GRCh38
rs376439431 Chr.17: 76528619-76528619 on Build GRCh38
rs533594657 Chr.17: 76528561-76528561 on Build GRCh38
rs751049284 Chr.17: 76528419-76528419 on Build GRCh38
rs564484579 Chr.17: 76528236-76528236 on Build GRCh38
rs999922961 Chr.17: 76528368-76528368 on Build GRCh38
rs376871878 Chr.17: 76528587-76528587 on Build GRCh38
rs1013628539 Chr.17: 76528252-76528252 on Build GRCh38
rs533419813 Chr.17: 76528251-76528251 on Build GRCh38
rs973009695 Chr.17: 76528377-76528377 on Build GRCh38
rs376307503 Chr.17: 76528569-76528570 on Build GRCh38
rs754447986 Chr.17: 76528557-76528558 on Build GRCh38
rs867492091 Chr.17: 76528289-76528289 on Build GRCh38
rs550914688 Chr.17: 76528448-76528448 on Build GRCh38
rs764541248 Chr.17: 76528558-76528558 on Build GRCh38
rs748173141 Chr.17: 76528501-76528501 on Build GRCh38
rs4238995 Chr.17: 76528569-76528569 on Build GRCh38
rs982098971 Chr.17: 76528277-76528277 on Build GRCh38
rs757473237 Chr.17: 76528610-76528610 on Build GRCh38
rs755463734 Chr.17: 76528579-76528579 on Build GRCh38
rs866655560 Chr.17: 76528602-76528602 on Build GRCh38
rs774830735 Chr.17: 76528539-76528539 on Build GRCh38
rs777836442 Chr.17: 76528507-76528507 on Build GRCh38
rs1024992635 Chr.17: 76528273-76528273 on Build GRCh38
rs73365381 Chr.17: 76528686-76528686 on Build GRCh38
rs200614504 Chr.17: 76528653-76528653 on Build GRCh38
rs990913722 Chr.17: 76528316-76528316 on Build GRCh38
rs982844157 Chr.17: 76528226-76528226 on Build GRCh38
rs781268131 Chr.17: 76528453-76528453 on Build GRCh38
rs8077736 Chr.17: 76528570-76528570 on Build GRCh38
rs1032747762 Chr.17: 76528668-76528668 on Build GRCh38
rs755703427 Chr.17: 76528692-76528692 on Build GRCh38
rs200763264 Chr.17: 76528541-76528541 on Build GRCh38
rs560413801 Chr.17: 76528290-76528290 on Build GRCh38
rs368357218 Chr.17: 76528506-76528506 on Build GRCh38
rs531892493 Chr.17: 76528435-76528435 on Build GRCh38
rs934207459 Chr.17: 76528412-76528412 on Build GRCh38
rs201985001 Chr.17: 76528605-76528605 on Build GRCh38
rs371993406 Chr.17: 76528580-76528580 on Build GRCh38
rs749452700 Chr.17: 76528606-76528606 on Build GRCh38
rs1003383757 Chr.17: 76528487-76528487 on Build GRCh38
rs767827601 Chr.17: 76528611-76528611 on Build GRCh38
rs908242743 Chr.17: 76528455-76528455 on Build GRCh38
rs762947995 Chr.17: 76528373-76528373 on Build GRCh38
rs890093517 Chr.17: 76528419-76528420 on Build GRCh38
rs540711421 Chr.17: 76528262-76528262 on Build GRCh38
rs529465191 Chr.17: 76528359-76528359 on Build GRCh38
rs549330907 Chr.17: 76528367-76528367 on Build GRCh38
rs536419150 Chr.17: 76528576-76528576 on Build GRCh38
rs776112356 Chr.17: 76528565-76528565 on Build GRCh38
rs186050618 Chr.17: 76528642-76528642 on Build GRCh38
rs190465457 Chr.17: 76528697-76528697 on Build GRCh38
rs181118054 Chr.17: 76528235-76528235 on Build GRCh38
rs1047652080 Chr.17: 76528225-76528225 on Build GRCh38
rs879232301 Chr.17: 76528436-76528436 on Build GRCh38
rs879523719 Chr.17: 76528569-76528570 on Build GRCh38
rs1010679166 Chr.17: 76528449-76528449 on Build GRCh38
rs764657629 Chr.17: 76528422-76528422 on Build GRCh38
rs142721826 Chr.17: 76528584-76528584 on Build GRCh38
rs958028642 Chr.17: 76528669-76528669 on Build GRCh38
rs370895072 Chr.17: 76528529-76528529 on Build GRCh38
rs1032616748 Chr.17: 76528369-76528369 on Build GRCh38
rs1054928513 Chr.17: 76528595-76528595 on Build GRCh38
rs779170393 Chr.17: 76528620-76528620 on Build GRCh38
rs922987063 Chr.17: 76528473-76528473 on Build GRCh38
rs774144863 Chr.17: 76528559-76528559 on Build GRCh38
rs775255584 Chr.17: 76528657-76528657 on Build GRCh38
rs775552141 Chr.17: 76528564-76528564 on Build GRCh38
rs145129132 Chr.17: 76528698-76528698 on Build GRCh38
rs894948635 Chr.17: 76528250-76528250 on Build GRCh38
rs918855458 Chr.17: 76528399-76528399 on Build GRCh38
rs1020713425 Chr.17: 76528458-76528458 on Build GRCh38
rs1007150582 Chr.17: 76528680-76528680 on Build GRCh38
rs201496863 Chr.17: 76528624-76528624 on Build GRCh38
rs866011378 Chr.17: 76528643-76528643 on Build GRCh38
rs985678787 Chr.17: 76528421-76528421 on Build GRCh38
rs767897368 Chr.17: 76528581-76528581 on Build GRCh38
rs1052143 Chr.17: 76528280-76528280 on Build GRCh38
rs201303782 Chr.17: 76528424-76528424 on Build GRCh38
rs745736716 Chr.17: 76528560-76528560 on Build GRCh38
rs902444184 Chr.17: 76528461-76528461 on Build GRCh38
rs752067750 Chr.17: 76528447-76528447 on Build GRCh38

View Full Product Details