Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76539955-76540457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYGB
Entrez Gene ID
114757
Gene Name
cytoglobin
Gene Aliases
HGB, STAP
UniGene
Hs.95120
Gene Chromosome Location
Chr.17: 76527348-76557692 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
UniGene
Hs.634380
Gene Chromosome Location
Chr.17: 76527586-76555339 on Build GRCh38
Transcripts
XM_017024116.1 NM_001077620.2 NR_033357.1 XM_017025013.1 XM_017025014.1 XM_017025015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6043133 c.46C>T p.R16C Chr.17: 76540187-76540187 on Build GRCh38
4505718 c.69C>T p.V23V Chr.17: 76540210-76540210 on Build GRCh38
4717425 c.42C>A p.L14L Chr.17: 76540183-76540183 on Build GRCh38
5364062 c.74C>T p.P25L Chr.17: 76540215-76540215 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs190352626 Chr.17: 76540381-76540381 on Build GRCh38
rs752819238 Chr.17: 76540110-76540110 on Build GRCh38
rs756245857 Chr.17: 76540114-76540114 on Build GRCh38
rs765956791 Chr.17: 76540247-76540248 on Build GRCh38
rs8176326 Chr.17: 76540251-76540251 on Build GRCh38
rs200398429 Chr.17: 76540357-76540357 on Build GRCh38
rs763381044 Chr.17: 76540099-76540099 on Build GRCh38
rs760704583 Chr.17: 76540206-76540206 on Build GRCh38
rs771143178 Chr.17: 76540194-76540194 on Build GRCh38
rs577810284 Chr.17: 76540250-76540250 on Build GRCh38
rs796417986 Chr.17: 76540251-76540252 on Build GRCh38
rs997138343 Chr.17: 76540416-76540416 on Build GRCh38
rs767104874 Chr.17: 76540251-76540253 on Build GRCh38
rs200682867 Chr.17: 76540138-76540138 on Build GRCh38
rs796839278 Chr.17: 76540250-76540251 on Build GRCh38
rs550914395 Chr.17: 76539992-76539992 on Build GRCh38
rs924180679 Chr.17: 76540308-76540308 on Build GRCh38
rs900212175 Chr.17: 76540092-76540092 on Build GRCh38
rs780865955 Chr.17: 76540232-76540232 on Build GRCh38
rs752618408 Chr.17: 76540267-76540267 on Build GRCh38
rs1011242073 Chr.17: 76540060-76540060 on Build GRCh38
rs758173779 Chr.17: 76540228-76540228 on Build GRCh38
rs556136946 Chr.17: 76540353-76540353 on Build GRCh38
rs900150433 Chr.17: 76540120-76540120 on Build GRCh38
rs763273515 Chr.17: 76540426-76540426 on Build GRCh38
rs564510990 Chr.17: 76540364-76540364 on Build GRCh38
rs768323513 Chr.17: 76540191-76540191 on Build GRCh38
rs754000197 Chr.17: 76540213-76540213 on Build GRCh38
rs377612170 Chr.17: 76540104-76540104 on Build GRCh38
rs948245799 Chr.17: 76540014-76540014 on Build GRCh38
rs562136087 Chr.17: 76540307-76540307 on Build GRCh38
rs1052907848 Chr.17: 76540047-76540047 on Build GRCh38
rs121918369 Chr.17: 76540146-76540146 on Build GRCh38
rs201090759 Chr.17: 76540236-76540236 on Build GRCh38
rs1043608064 Chr.17: 76540383-76540383 on Build GRCh38
rs199693977 Chr.17: 76540196-76540196 on Build GRCh38
rs760102518 Chr.17: 76540102-76540102 on Build GRCh38
rs387907268 Chr.17: 76540205-76540205 on Build GRCh38
rs755130399 Chr.17: 76540252-76540253 on Build GRCh38
rs765266825 Chr.17: 76540262-76540262 on Build GRCh38
rs111326198 Chr.17: 76540253-76540253 on Build GRCh38
rs892886625 Chr.17: 76540048-76540048 on Build GRCh38
rs769820518 Chr.17: 76540137-76540138 on Build GRCh38
rs779911519 Chr.17: 76540175-76540175 on Build GRCh38
rs759275085 Chr.17: 76540256-76540257 on Build GRCh38
rs4648338 Chr.17: 76540254-76540254 on Build GRCh38
rs112889580 Chr.17: 76540398-76540398 on Build GRCh38
rs200954045 Chr.17: 76540154-76540154 on Build GRCh38
rs769483001 Chr.17: 76540241-76540241 on Build GRCh38
rs527236092 Chr.17: 76540143-76540143 on Build GRCh38
rs112831645 Chr.17: 76540346-76540346 on Build GRCh38
rs151090701 Chr.17: 76540300-76540300 on Build GRCh38
rs757471313 Chr.17: 76540215-76540215 on Build GRCh38
rs78529481 Chr.17: 76540257-76540257 on Build GRCh38
rs8176324 Chr.17: 76540255-76540255 on Build GRCh38
rs772423009 Chr.17: 76540260-76540260 on Build GRCh38
rs766812624 Chr.17: 76540261-76540262 on Build GRCh38
rs201363694 Chr.17: 76540197-76540197 on Build GRCh38
rs370328118 Chr.17: 76540139-76540139 on Build GRCh38
rs754878521 Chr.17: 76540265-76540265 on Build GRCh38
rs776386516 Chr.17: 76540192-76540192 on Build GRCh38
rs557680109 Chr.17: 76540243-76540243 on Build GRCh38
rs758721254 Chr.17: 76540252-76540252 on Build GRCh38
rs754088083 Chr.17: 76540133-76540133 on Build GRCh38
rs751836791 Chr.17: 76540263-76540264 on Build GRCh38
rs575651772 Chr.17: 76540328-76540328 on Build GRCh38
rs746223894 Chr.17: 76540244-76540244 on Build GRCh38
rs765737108 Chr.17: 76540258-76540259 on Build GRCh38
rs763101674 Chr.17: 76540264-76540264 on Build GRCh38
rs1040410887 Chr.17: 76540358-76540359 on Build GRCh38
rs747650092 Chr.17: 76540258-76540258 on Build GRCh38
rs527451635 Chr.17: 76540193-76540193 on Build GRCh38
rs77817682 Chr.17: 76540251-76540251 on Build GRCh38
rs766885858 Chr.17: 76540100-76540100 on Build GRCh38
rs754493922 Chr.17: 76540230-76540230 on Build GRCh38
rs547310148 Chr.17: 76540036-76540036 on Build GRCh38
rs757827110 Chr.17: 76540251-76540252 on Build GRCh38
rs773201535 Chr.17: 76540200-76540203 on Build GRCh38
rs751966024 Chr.17: 76540101-76540101 on Build GRCh38
rs773995352 Chr.17: 76540255-76540256 on Build GRCh38
rs200645008 Chr.17: 76540229-76540229 on Build GRCh38
rs947152822 Chr.17: 76540375-76540375 on Build GRCh38
rs1022135434 Chr.17: 76540141-76540142 on Build GRCh38
rs778170546 Chr.17: 76539980-76539980 on Build GRCh38
rs963473792 Chr.17: 76540287-76540287 on Build GRCh38
rs560994325 Chr.17: 76540263-76540263 on Build GRCh38
rs201716822 Chr.17: 76540253-76540254 on Build GRCh38
rs575918636 Chr.17: 76540254-76540255 on Build GRCh38
rs1008952622 Chr.17: 76540066-76540066 on Build GRCh38
rs548816444 Chr.17: 76540223-76540223 on Build GRCh38
rs566126436 Chr.17: 76540090-76540090 on Build GRCh38
rs935563092 Chr.17: 76540334-76540334 on Build GRCh38
rs375181336 Chr.17: 76540190-76540190 on Build GRCh38
rs139779268 Chr.17: 76539995-76539995 on Build GRCh38
rs774019278 Chr.17: 76540250-76540251 on Build GRCh38
rs762780716 Chr.17: 76540210-76540210 on Build GRCh38
rs772352854 Chr.17: 76540252-76540252 on Build GRCh38
rs777938956 Chr.17: 76540115-76540115 on Build GRCh38
rs142920556 Chr.17: 76539998-76539998 on Build GRCh38
rs766112913 Chr.17: 76540256-76540256 on Build GRCh38
rs970698723 Chr.17: 76540305-76540305 on Build GRCh38
rs755292579 Chr.17: 76540264-76540266 on Build GRCh38
rs779066277 Chr.17: 76540216-76540216 on Build GRCh38
rs756997414 Chr.17: 76540134-76540134 on Build GRCh38
rs758991108 Chr.17: 76540250-76540250 on Build GRCh38
rs999334657 Chr.17: 76540413-76540413 on Build GRCh38
rs767541846 Chr.17: 76540266-76540266 on Build GRCh38
rs748914780 Chr.17: 76540259-76540259 on Build GRCh38
rs769134763 Chr.17: 76540261-76540261 on Build GRCh38
rs990873381 Chr.17: 76540289-76540289 on Build GRCh38
rs775037822 Chr.17: 76540257-76540258 on Build GRCh38

View Full Product Details