Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78165884-78166380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf99
Entrez Gene ID
100141515
Gene Name
chromosome 17 open reading frame 99
Gene Aliases
UNQ464
UniGene
Hs.633034
Gene Chromosome Location
Chr.17: 78146035-78166283 on Build GRCh38
Transcripts
NM_001163075.1 XM_011524152.2 XM_017023999.1 XM_017024000.1 XM_017024001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985038 c.719G>A p.R240H Chr.17: 78165967-78165967 on Build GRCh38
6532571 c.711G>C p.R237S Chr.17: 78165959-78165959 on Build GRCh38
5387449 c.654G>A p.K218K Chr.17: 78165902-78165902 on Build GRCh38
985039 c.721C>T p.R241C Chr.17: 78165969-78165969 on Build GRCh38
3820762 c.651G>A p.Q217Q Chr.17: 78165899-78165899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371895105 Chr.17: 78165991-78165991 on Build GRCh38
rs983665036 Chr.17: 78166306-78166307 on Build GRCh38
rs915812933 Chr.17: 78165990-78165990 on Build GRCh38
rs770106906 Chr.17: 78166026-78166026 on Build GRCh38
rs962547205 Chr.17: 78166126-78166126 on Build GRCh38
rs535562967 Chr.17: 78165999-78165999 on Build GRCh38
rs970481491 Chr.17: 78166199-78166199 on Build GRCh38
rs900194249 Chr.17: 78166055-78166055 on Build GRCh38
rs867675890 Chr.17: 78165914-78165914 on Build GRCh38
rs1011099297 Chr.17: 78166217-78166217 on Build GRCh38
rs34402400 Chr.17: 78166154-78166154 on Build GRCh38
rs61377640 Chr.17: 78166177-78166177 on Build GRCh38
rs921676521 Chr.17: 78166345-78166345 on Build GRCh38
rs1052885689 Chr.17: 78166203-78166203 on Build GRCh38
rs781226810 Chr.17: 78165984-78165984 on Build GRCh38
rs999437847 Chr.17: 78166124-78166124 on Build GRCh38
rs759256009 Chr.17: 78165989-78165990 on Build GRCh38
rs71360608 Chr.17: 78166155-78166156 on Build GRCh38
rs3642 Chr.17: 78166097-78166097 on Build GRCh38
rs922729853 Chr.17: 78165949-78165949 on Build GRCh38
rs555132847 Chr.17: 78166027-78166027 on Build GRCh38
rs142807501 Chr.17: 78166216-78166216 on Build GRCh38
rs997539607 Chr.17: 78166270-78166270 on Build GRCh38
rs542794121 Chr.17: 78166307-78166307 on Build GRCh38
rs1022567817 Chr.17: 78166068-78166068 on Build GRCh38
rs374986464 Chr.17: 78166012-78166012 on Build GRCh38
rs774290112 Chr.17: 78166017-78166017 on Build GRCh38
rs148532300 Chr.17: 78166041-78166041 on Build GRCh38
rs34177315 Chr.17: 78166037-78166038 on Build GRCh38
rs930808913 Chr.17: 78165994-78165994 on Build GRCh38
rs973848576 Chr.17: 78166136-78166136 on Build GRCh38
rs868659118 Chr.17: 78165919-78165919 on Build GRCh38
rs982642714 Chr.17: 78165961-78165961 on Build GRCh38
rs920851825 Chr.17: 78166246-78166246 on Build GRCh38
rs375303408 Chr.17: 78166072-78166072 on Build GRCh38
rs934326594 Chr.17: 78166189-78166189 on Build GRCh38
rs964068815 Chr.17: 78165908-78165908 on Build GRCh38
rs750873362 Chr.17: 78165920-78165920 on Build GRCh38
rs766926568 Chr.17: 78166070-78166070 on Build GRCh38
rs3644 Chr.17: 78166053-78166053 on Build GRCh38
rs948603483 Chr.17: 78166157-78166157 on Build GRCh38
rs74360190 Chr.17: 78166181-78166181 on Build GRCh38
rs1030041876 Chr.17: 78166087-78166087 on Build GRCh38
rs758983769 Chr.17: 78166034-78166034 on Build GRCh38
rs34243262 Chr.17: 78165972-78165972 on Build GRCh38
rs886600615 Chr.17: 78166021-78166021 on Build GRCh38
rs59369521 Chr.17: 78166313-78166313 on Build GRCh38
rs908612131 Chr.17: 78166107-78166107 on Build GRCh38
rs1043879436 Chr.17: 78166220-78166220 on Build GRCh38
rs915847002 Chr.17: 78166138-78166138 on Build GRCh38
rs1011066391 Chr.17: 78166062-78166062 on Build GRCh38
rs906960191 Chr.17: 78166050-78166050 on Build GRCh38
rs759038653 Chr.17: 78165938-78165938 on Build GRCh38
rs781377423 Chr.17: 78166080-78166080 on Build GRCh38
rs549659332 Chr.17: 78165995-78165995 on Build GRCh38
rs769297604 Chr.17: 78166096-78166096 on Build GRCh38
rs200942539 Chr.17: 78165948-78165948 on Build GRCh38
rs372937776 Chr.17: 78165966-78165966 on Build GRCh38
rs762012201 Chr.17: 78166245-78166245 on Build GRCh38
rs1045644231 Chr.17: 78166178-78166178 on Build GRCh38
rs771602535 Chr.17: 78166036-78166036 on Build GRCh38
rs1038274309 Chr.17: 78165977-78165977 on Build GRCh38
rs560 Chr.17: 78166054-78166054 on Build GRCh38
rs900090992 Chr.17: 78166237-78166237 on Build GRCh38
rs1040110377 Chr.17: 78166325-78166325 on Build GRCh38
rs1024366601 Chr.17: 78166172-78166172 on Build GRCh38
rs928330511 Chr.17: 78166184-78166184 on Build GRCh38
rs540940003 Chr.17: 78165970-78165970 on Build GRCh38
rs770915682 Chr.17: 78166013-78166013 on Build GRCh38
rs1014378396 Chr.17: 78166159-78166160 on Build GRCh38
rs577882614 Chr.17: 78166058-78166058 on Build GRCh38
rs553113603 Chr.17: 78165950-78165950 on Build GRCh38
rs955605505 Chr.17: 78165956-78165956 on Build GRCh38
rs560203521 Chr.17: 78166143-78166143 on Build GRCh38
rs755057023 Chr.17: 78166073-78166073 on Build GRCh38

View Full Product Details