Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78423609-78424112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH17
Entrez Gene ID
8632
Gene Name
dynein axonemal heavy chain 17
Gene Aliases
DNAHL1, DNEL2
UniGene
Hs.375975
Gene Chromosome Location
Chr.17: 78423697-78577430 on Build GRCh38
Transcripts
NM_173628.3 XM_011525416.2 XM_017025261.1 NM_024419.4 NR_110601.1 NR_110602.1 NR_111989.1 XM_011525487.2 XM_011525488.2 XM_011525489.2 XM_017025357.1 XM_017025358.1 XM_017025359.1 XM_017025360.1 XM_017025361.1 XM_017025362.1 XM_017025363.1 XM_017025364.1 XM_017025365.1 XM_017025366.1

Gene Symbol
PGS1
Entrez Gene ID
9489
Gene Name
phosphatidylglycerophosphate synthase 1
Gene Aliases
-
UniGene
Hs.654671
Gene Chromosome Location
Chr.17: 78378618-78424659 on Build GRCh38
Transcripts
NM_173628.3 XM_011525416.2 XM_017025261.1 NM_024419.4 NR_110601.1 NR_110602.1 NR_111989.1 XM_011525487.2 XM_011525488.2 XM_011525489.2 XM_017025357.1 XM_017025358.1 XM_017025359.1 XM_017025360.1 XM_017025361.1 XM_017025362.1 XM_017025363.1 XM_017025364.1 XM_017025365.1 XM_017025366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244075 c.13189G>A p.A4397T Chr.17: 78424091-78424091 on Build GRCh38
148353 c.13276A>G p.I4426V Chr.17: 78424004-78424004 on Build GRCh38
4872346 c.13434C>T p.A4478A Chr.17: 78423930-78423930 on Build GRCh38
4985750 c.13187C>T p.P4396L Chr.17: 78424093-78424093 on Build GRCh38
6389642 c.13436T>A p.V4479E Chr.17: 78423928-78423928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367916838 Chr.17: 78423858-78423858 on Build GRCh38
rs768324369 Chr.17: 78424042-78424042 on Build GRCh38
rs866320078 Chr.17: 78423662-78423662 on Build GRCh38
rs761373066 Chr.17: 78423998-78423998 on Build GRCh38
rs375297222 Chr.17: 78424019-78424019 on Build GRCh38
rs761355727 Chr.17: 78423892-78423893 on Build GRCh38
rs1034543625 Chr.17: 78423660-78423660 on Build GRCh38
rs768575116 Chr.17: 78423990-78423990 on Build GRCh38
rs143472261 Chr.17: 78423640-78423640 on Build GRCh38
rs1009343510 Chr.17: 78423992-78423992 on Build GRCh38
rs190586281 Chr.17: 78423993-78423993 on Build GRCh38
rs956448686 Chr.17: 78423815-78423815 on Build GRCh38
rs751856381 Chr.17: 78423886-78423886 on Build GRCh38
rs930506200 Chr.17: 78423785-78423785 on Build GRCh38
rs781747756 Chr.17: 78424029-78424030 on Build GRCh38
rs777727996 Chr.17: 78423963-78423963 on Build GRCh38
rs762364121 Chr.17: 78424079-78424079 on Build GRCh38
rs377016460 Chr.17: 78423924-78423924 on Build GRCh38
rs761156949 Chr.17: 78424060-78424060 on Build GRCh38
rs763633164 Chr.17: 78423879-78423879 on Build GRCh38
rs779721438 Chr.17: 78424029-78424029 on Build GRCh38
rs570803591 Chr.17: 78423925-78423925 on Build GRCh38
rs143246806 Chr.17: 78423949-78423949 on Build GRCh38
rs903350683 Chr.17: 78423865-78423865 on Build GRCh38
rs904849133 Chr.17: 78423641-78423641 on Build GRCh38
rs765424936 Chr.17: 78424092-78424092 on Build GRCh38
rs764526288 Chr.17: 78424061-78424061 on Build GRCh38
rs774289227 Chr.17: 78423921-78423921 on Build GRCh38
rs984753075 Chr.17: 78424032-78424032 on Build GRCh38
rs142983650 Chr.17: 78424052-78424052 on Build GRCh38
rs749838869 Chr.17: 78423965-78423967 on Build GRCh38
rs755041875 Chr.17: 78423763-78423763 on Build GRCh38
rs1020371413 Chr.17: 78423693-78423693 on Build GRCh38
rs755577173 Chr.17: 78423987-78423988 on Build GRCh38
rs116446606 Chr.17: 78423887-78423887 on Build GRCh38
rs78023288 Chr.17: 78424004-78424004 on Build GRCh38
rs952026712 Chr.17: 78424073-78424073 on Build GRCh38
rs746767108 Chr.17: 78423845-78423845 on Build GRCh38
rs548751432 Chr.17: 78423646-78423646 on Build GRCh38
rs769803281 Chr.17: 78423902-78423902 on Build GRCh38
rs979108910 Chr.17: 78423787-78423790 on Build GRCh38
rs781607423 Chr.17: 78423894-78423894 on Build GRCh38
rs761462566 Chr.17: 78423938-78423938 on Build GRCh38
rs149800703 Chr.17: 78424001-78424001 on Build GRCh38
rs773868447 Chr.17: 78424058-78424058 on Build GRCh38
rs909240251 Chr.17: 78424049-78424049 on Build GRCh38
rs984728174 Chr.17: 78424082-78424082 on Build GRCh38
rs953388197 Chr.17: 78423750-78423750 on Build GRCh38
rs753237288 Chr.17: 78424032-78424037 on Build GRCh38
rs760156744 Chr.17: 78423875-78423875 on Build GRCh38
rs748047706 Chr.17: 78424048-78424048 on Build GRCh38
rs764725957 Chr.17: 78424007-78424007 on Build GRCh38
rs542698237 Chr.17: 78423760-78423760 on Build GRCh38
rs112488587 Chr.17: 78423761-78423761 on Build GRCh38
rs769326697 Chr.17: 78424051-78424051 on Build GRCh38
rs113538058 Chr.17: 78423951-78423951 on Build GRCh38
rs376706067 Chr.17: 78423825-78423827 on Build GRCh38
rs767732868 Chr.17: 78423639-78423639 on Build GRCh38
rs566746938 Chr.17: 78423936-78423936 on Build GRCh38
rs370782545 Chr.17: 78423720-78423720 on Build GRCh38
rs1056034513 Chr.17: 78423825-78423825 on Build GRCh38
rs776601455 Chr.17: 78424047-78424047 on Build GRCh38
rs141424232 Chr.17: 78423930-78423930 on Build GRCh38
rs779330949 Chr.17: 78423972-78423972 on Build GRCh38
rs764815092 Chr.17: 78423959-78423960 on Build GRCh38
rs761221802 Chr.17: 78423681-78423681 on Build GRCh38
rs753468668 Chr.17: 78423882-78423882 on Build GRCh38
rs756856607 Chr.17: 78423883-78423883 on Build GRCh38
rs1033553098 Chr.17: 78423742-78423742 on Build GRCh38
rs753590432 Chr.17: 78423862-78423863 on Build GRCh38
rs779212708 Chr.17: 78423764-78423764 on Build GRCh38
rs553471949 Chr.17: 78423670-78423670 on Build GRCh38
rs762524778 Chr.17: 78424053-78424053 on Build GRCh38
rs150783880 Chr.17: 78423704-78423704 on Build GRCh38
rs753334032 Chr.17: 78423934-78423934 on Build GRCh38
rs754432776 Chr.17: 78424068-78424068 on Build GRCh38
rs199669660 Chr.17: 78423965-78423965 on Build GRCh38
rs533842240 Chr.17: 78424020-78424020 on Build GRCh38
rs1000797316 Chr.17: 78423869-78423869 on Build GRCh38
rs544945593 Chr.17: 78423800-78423800 on Build GRCh38
rs775310662 Chr.17: 78423983-78423983 on Build GRCh38
rs553733191 Chr.17: 78424027-78424027 on Build GRCh38
rs780413522 Chr.17: 78423884-78423884 on Build GRCh38
rs562278569 Chr.17: 78423767-78423767 on Build GRCh38
rs927551330 Chr.17: 78423861-78423861 on Build GRCh38
rs775238626 Chr.17: 78424000-78424000 on Build GRCh38
rs746936275 Chr.17: 78423988-78423988 on Build GRCh38
rs764239849 Chr.17: 78423834-78423835 on Build GRCh38
rs765988102 Chr.17: 78424055-78424055 on Build GRCh38
rs866168240 Chr.17: 78423912-78423912 on Build GRCh38
rs568171189 Chr.17: 78424002-78424002 on Build GRCh38
rs777476607 Chr.17: 78423843-78423843 on Build GRCh38
rs748666814 Chr.17: 78423897-78423897 on Build GRCh38
rs1000467241 Chr.17: 78424003-78424003 on Build GRCh38
rs117340568 Chr.17: 78423999-78423999 on Build GRCh38
rs753237874 Chr.17: 78423679-78423679 on Build GRCh38
rs757406187 Chr.17: 78423969-78423969 on Build GRCh38
rs372004613 Chr.17: 78423948-78423948 on Build GRCh38
rs771797237 Chr.17: 78423979-78423979 on Build GRCh38
rs746062866 Chr.17: 78423976-78423976 on Build GRCh38
rs138312957 Chr.17: 78424017-78424017 on Build GRCh38
rs72903323 Chr.17: 78423801-78423801 on Build GRCh38
rs752671887 Chr.17: 78423868-78423868 on Build GRCh38
rs531093474 Chr.17: 78423791-78423791 on Build GRCh38
rs945086397 Chr.17: 78423984-78423984 on Build GRCh38
rs951893698 Chr.17: 78423947-78423947 on Build GRCh38
rs771217788 Chr.17: 78423911-78423911 on Build GRCh38
rs568715759 Chr.17: 78423674-78423674 on Build GRCh38
rs937678019 Chr.17: 78423819-78423819 on Build GRCh38
rs919056185 Chr.17: 78423784-78423784 on Build GRCh38
rs971975444 Chr.17: 78423775-78423775 on Build GRCh38
rs775335572 Chr.17: 78423929-78423929 on Build GRCh38
rs199723688 Chr.17: 78424093-78424093 on Build GRCh38
rs746118933 Chr.17: 78424022-78424022 on Build GRCh38
rs72914863 Chr.17: 78423903-78423903 on Build GRCh38
rs746892324 Chr.17: 78424035-78424035 on Build GRCh38
rs150439390 Chr.17: 78424006-78424006 on Build GRCh38
rs754163022 Chr.17: 78423890-78423890 on Build GRCh38
rs1029061933 Chr.17: 78423765-78423765 on Build GRCh38

View Full Product Details