Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36095551-36096062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A2
Entrez Gene ID
1296
Gene Name
collagen type VIII alpha 2
Gene Aliases
FECD, FECD1, PPCD, PPCD2
UniGene
Hs.353001
Gene Chromosome Location
Chr.1: 36095236-36126207 on Build GRCh38
Transcripts
NM_001294347.1 NM_005202.3 XM_005270477.3

SNP Detail

SNP ID Chromosome Location
rs1013829386 Chr.1: 36095745-36095745 on Build GRCh38
rs1032847618 Chr.1: 36095798-36095798 on Build GRCh38
rs958138373 Chr.1: 36095816-36095816 on Build GRCh38
rs777449161 Chr.1: 36095783-36095783 on Build GRCh38
rs898118822 Chr.1: 36095744-36095744 on Build GRCh38
rs943953509 Chr.1: 36095585-36095585 on Build GRCh38
rs147779302 Chr.1: 36095975-36095975 on Build GRCh38
rs952645674 Chr.1: 36095655-36095656 on Build GRCh38
rs1025178326 Chr.1: 36095767-36095767 on Build GRCh38
rs932472614 Chr.1: 36095666-36095666 on Build GRCh38
rs578129822 Chr.1: 36095649-36095649 on Build GRCh38
rs895195213 Chr.1: 36096011-36096011 on Build GRCh38
rs577411019 Chr.1: 36095610-36095610 on Build GRCh38
rs1036724646 Chr.1: 36096006-36096006 on Build GRCh38
rs929661140 Chr.1: 36095946-36095949 on Build GRCh38
rs563693175 Chr.1: 36095933-36095933 on Build GRCh38
rs534760285 Chr.1: 36095658-36095658 on Build GRCh38
rs1555652 Chr.1: 36095988-36095988 on Build GRCh38
rs139543144 Chr.1: 36095982-36095982 on Build GRCh38
rs909658203 Chr.1: 36095918-36095918 on Build GRCh38
rs545393346 Chr.1: 36095842-36095842 on Build GRCh38
rs1034198489 Chr.1: 36096008-36096008 on Build GRCh38
rs879467223 Chr.1: 36095972-36095972 on Build GRCh38
rs115602831 Chr.1: 36095784-36095784 on Build GRCh38
rs978323463 Chr.1: 36095803-36095803 on Build GRCh38
rs563830315 Chr.1: 36095719-36095719 on Build GRCh38
rs888393117 Chr.1: 36096003-36096003 on Build GRCh38
rs888165512 Chr.1: 36095696-36095696 on Build GRCh38
rs921155555 Chr.1: 36095851-36095851 on Build GRCh38
rs557409077 Chr.1: 36095872-36095872 on Build GRCh38
rs913852663 Chr.1: 36095584-36095584 on Build GRCh38
rs775350483 Chr.1: 36095820-36095823 on Build GRCh38
rs900846130 Chr.1: 36095939-36095939 on Build GRCh38
rs983937414 Chr.1: 36095905-36095905 on Build GRCh38
rs976649283 Chr.1: 36095846-36095846 on Build GRCh38
rs910949863 Chr.1: 36095908-36095910 on Build GRCh38
rs9803882 Chr.1: 36095820-36095820 on Build GRCh38
rs76680564 Chr.1: 36095830-36095830 on Build GRCh38
rs1040710752 Chr.1: 36095930-36095930 on Build GRCh38
rs976578650 Chr.1: 36095650-36095650 on Build GRCh38
rs553537757 Chr.1: 36095731-36095731 on Build GRCh38
rs959510800 Chr.1: 36095707-36095707 on Build GRCh38
rs939691022 Chr.1: 36095700-36095700 on Build GRCh38
rs991197671 Chr.1: 36095717-36095717 on Build GRCh38
rs773119451 Chr.1: 36095673-36095673 on Build GRCh38
rs144198843 Chr.1: 36095836-36095836 on Build GRCh38

View Full Product Details