Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37557504-37557903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNALI1
Entrez Gene ID
7802
Gene Name
dynein axonemal light intermediate chain 1
Gene Aliases
P28, dJ423B22.5, hp28
UniGene
Hs.406050
Gene Chromosome Location
Chr.1: 37556919-37566857 on Build GRCh38
Transcripts
NM_003462.3 XM_005271172.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4539211 c.265G>A p.E89K Chr.1: 37557720-37557720 on Build GRCh38
5583455 c.213G>A p.K71K Chr.1: 37557668-37557668 on Build GRCh38
3671748 c.225C>A p.P75P Chr.1: 37557680-37557680 on Build GRCh38
1748398 c.271T>A p.L91M Chr.1: 37557726-37557726 on Build GRCh38
5846578 c.151C>T p.R51W Chr.1: 37557606-37557606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773394668 Chr.1: 37557604-37557604 on Build GRCh38
rs765928213 Chr.1: 37557528-37557528 on Build GRCh38
rs753509254 Chr.1: 37557657-37557657 on Build GRCh38
rs771643431 Chr.1: 37557786-37557786 on Build GRCh38
rs1801956 Chr.1: 37557688-37557688 on Build GRCh38
rs770308101 Chr.1: 37557600-37557600 on Build GRCh38
rs373665703 Chr.1: 37557766-37557766 on Build GRCh38
rs144000156 Chr.1: 37557776-37557776 on Build GRCh38
rs773768864 Chr.1: 37557699-37557699 on Build GRCh38
rs759145155 Chr.1: 37557725-37557725 on Build GRCh38
rs368402927 Chr.1: 37557606-37557606 on Build GRCh38
rs530145221 Chr.1: 37557689-37557689 on Build GRCh38
rs11749 Chr.1: 37557715-37557715 on Build GRCh38
rs556359043 Chr.1: 37557811-37557811 on Build GRCh38
rs111272874 Chr.1: 37557792-37557792 on Build GRCh38
rs771302872 Chr.1: 37557718-37557718 on Build GRCh38
rs1051087171 Chr.1: 37557723-37557723 on Build GRCh38
rs751208218 Chr.1: 37557618-37557618 on Build GRCh38
rs781430099 Chr.1: 37557753-37557753 on Build GRCh38
rs371640349 Chr.1: 37557607-37557607 on Build GRCh38
rs747202885 Chr.1: 37557676-37557676 on Build GRCh38
rs539568664 Chr.1: 37557870-37557870 on Build GRCh38
rs747170907 Chr.1: 37557814-37557814 on Build GRCh38
rs140782337 Chr.1: 37557881-37557881 on Build GRCh38
rs552385945 Chr.1: 37557741-37557741 on Build GRCh38
rs755196470 Chr.1: 37557750-37557750 on Build GRCh38
rs1016110375 Chr.1: 37557765-37557765 on Build GRCh38
rs760154244 Chr.1: 37557800-37557800 on Build GRCh38
rs777636528 Chr.1: 37557656-37557656 on Build GRCh38
rs752574069 Chr.1: 37557581-37557581 on Build GRCh38
rs577512383 Chr.1: 37557647-37557647 on Build GRCh38
rs559111896 Chr.1: 37557640-37557640 on Build GRCh38
rs146412745 Chr.1: 37557840-37557840 on Build GRCh38
rs182790694 Chr.1: 37557859-37557859 on Build GRCh38
rs765042592 Chr.1: 37557745-37557745 on Build GRCh38
rs200574126 Chr.1: 37557686-37557686 on Build GRCh38
rs757063771 Chr.1: 37557664-37557664 on Build GRCh38
rs745532362 Chr.1: 37557780-37557780 on Build GRCh38
rs981682262 Chr.1: 37557813-37557814 on Build GRCh38
rs777318339 Chr.1: 37557575-37557575 on Build GRCh38
rs116565398 Chr.1: 37557876-37557876 on Build GRCh38
rs770135365 Chr.1: 37557690-37557690 on Build GRCh38
rs963213111 Chr.1: 37557531-37557531 on Build GRCh38
rs199834277 Chr.1: 37557687-37557687 on Build GRCh38
rs199883096 Chr.1: 37557639-37557639 on Build GRCh38
rs191581730 Chr.1: 37557815-37557815 on Build GRCh38
rs974243361 Chr.1: 37557533-37557533 on Build GRCh38
rs767552781 Chr.1: 37557579-37557579 on Build GRCh38
rs907299847 Chr.1: 37557783-37557783 on Build GRCh38
rs72657795 Chr.1: 37557547-37557547 on Build GRCh38
rs762323760 Chr.1: 37557714-37557716 on Build GRCh38
rs780185974 Chr.1: 37557554-37557554 on Build GRCh38
rs144577882 Chr.1: 37557737-37557737 on Build GRCh38
rs751853936 Chr.1: 37557747-37557747 on Build GRCh38
rs377298896 Chr.1: 37557574-37557574 on Build GRCh38
rs768561327 Chr.1: 37557869-37557869 on Build GRCh38
rs140822664 Chr.1: 37557722-37557722 on Build GRCh38
rs780190818 Chr.1: 37557670-37557670 on Build GRCh38
rs182397050 Chr.1: 37557566-37557566 on Build GRCh38
rs913612898 Chr.1: 37557669-37557669 on Build GRCh38
rs758658526 Chr.1: 37557759-37557759 on Build GRCh38
rs1023095115 Chr.1: 37557774-37557774 on Build GRCh38
rs755996510 Chr.1: 37557653-37557653 on Build GRCh38
rs570351290 Chr.1: 37557744-37557744 on Build GRCh38
rs748308242 Chr.1: 37557763-37557763 on Build GRCh38
rs766979427 Chr.1: 37557646-37557646 on Build GRCh38
rs1039529294 Chr.1: 37557626-37557626 on Build GRCh38
rs771227020 Chr.1: 37557779-37557779 on Build GRCh38
rs1022666209 Chr.1: 37557816-37557816 on Build GRCh38

View Full Product Details