Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37561477-37561985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNALI1
Entrez Gene ID
7802
Gene Name
dynein axonemal light intermediate chain 1
Gene Aliases
P28, dJ423B22.5, hp28
UniGene
Hs.406050
Gene Chromosome Location
Chr.1: 37556919-37566857 on Build GRCh38
Transcripts
NM_003462.3 XM_005271172.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667421 c.586T>C p.F196L Chr.1: 37561679-37561679 on Build GRCh38
3489208 c.475C>T p.R159W Chr.1: 37561568-37561568 on Build GRCh38
1667422 c.593T>C p.M198T Chr.1: 37561686-37561686 on Build GRCh38
4553653 c.597G>A p.R199R Chr.1: 37561690-37561690 on Build GRCh38
4566979 c.527_528GG>AA p.R176Q Chr.1: 37561620-37561621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190675614 Chr.1: 37561618-37561618 on Build GRCh38
rs752031981 Chr.1: 37561587-37561587 on Build GRCh38
rs771061760 Chr.1: 37561757-37561757 on Build GRCh38
rs574421086 Chr.1: 37561941-37561941 on Build GRCh38
rs1054155860 Chr.1: 37561866-37561866 on Build GRCh38
rs776125437 Chr.1: 37561773-37561773 on Build GRCh38
rs368021115 Chr.1: 37561760-37561760 on Build GRCh38
rs139139227 Chr.1: 37561682-37561682 on Build GRCh38
rs150256817 Chr.1: 37561556-37561556 on Build GRCh38
rs547905165 Chr.1: 37561680-37561680 on Build GRCh38
rs749249059 Chr.1: 37561620-37561620 on Build GRCh38
rs990905854 Chr.1: 37561933-37561933 on Build GRCh38
rs140823500 Chr.1: 37561664-37561664 on Build GRCh38
rs558543758 Chr.1: 37561921-37561922 on Build GRCh38
rs759907358 Chr.1: 37561693-37561693 on Build GRCh38
rs778238207 Chr.1: 37561725-37561725 on Build GRCh38
rs17559127 Chr.1: 37561547-37561547 on Build GRCh38
rs758204522 Chr.1: 37561731-37561731 on Build GRCh38
rs762940257 Chr.1: 37561520-37561520 on Build GRCh38
rs201185676 Chr.1: 37561751-37561751 on Build GRCh38
rs752351857 Chr.1: 37561648-37561649 on Build GRCh38
rs756830560 Chr.1: 37561724-37561724 on Build GRCh38
rs1042513377 Chr.1: 37561784-37561784 on Build GRCh38
rs375606011 Chr.1: 37561735-37561735 on Build GRCh38
rs774148914 Chr.1: 37561624-37561624 on Build GRCh38
rs370651927 Chr.1: 37561614-37561614 on Build GRCh38
rs200794930 Chr.1: 37561685-37561685 on Build GRCh38
rs753575785 Chr.1: 37561776-37561776 on Build GRCh38
rs755287513 Chr.1: 37561591-37561591 on Build GRCh38
rs769685939 Chr.1: 37561636-37561636 on Build GRCh38
rs768925360 Chr.1: 37561568-37561568 on Build GRCh38
rs909203115 Chr.1: 37561873-37561873 on Build GRCh38
rs561955832 Chr.1: 37561619-37561619 on Build GRCh38
rs754489684 Chr.1: 37561600-37561602 on Build GRCh38
rs369054456 Chr.1: 37561521-37561521 on Build GRCh38
rs1042482618 Chr.1: 37561557-37561557 on Build GRCh38
rs778958257 Chr.1: 37561668-37561668 on Build GRCh38
rs767933489 Chr.1: 37561704-37561704 on Build GRCh38
rs758846821 Chr.1: 37561573-37561573 on Build GRCh38
rs751079730 Chr.1: 37561599-37561600 on Build GRCh38
rs1049360294 Chr.1: 37561509-37561509 on Build GRCh38
rs760809781 Chr.1: 37561533-37561533 on Build GRCh38
rs756540426 Chr.1: 37561711-37561711 on Build GRCh38
rs1055280790 Chr.1: 37561503-37561503 on Build GRCh38
rs764442553 Chr.1: 37561534-37561534 on Build GRCh38
rs373304393 Chr.1: 37561527-37561527 on Build GRCh38
rs893067533 Chr.1: 37561883-37561883 on Build GRCh38
rs544621984 Chr.1: 37561822-37561822 on Build GRCh38
rs565918744 Chr.1: 37561684-37561684 on Build GRCh38
rs749475247 Chr.1: 37561752-37561752 on Build GRCh38
rs746620956 Chr.1: 37561517-37561517 on Build GRCh38
rs916629790 Chr.1: 37561942-37561942 on Build GRCh38
rs374132227 Chr.1: 37561639-37561639 on Build GRCh38
rs760090156 Chr.1: 37561762-37561762 on Build GRCh38
rs771924444 Chr.1: 37561673-37561673 on Build GRCh38
rs752417147 Chr.1: 37561653-37561653 on Build GRCh38
rs776053232 Chr.1: 37561569-37561569 on Build GRCh38
rs540760477 Chr.1: 37561863-37561863 on Build GRCh38
rs202055051 Chr.1: 37561677-37561677 on Build GRCh38
rs763018839 Chr.1: 37561640-37561640 on Build GRCh38
rs138230125 Chr.1: 37561847-37561847 on Build GRCh38
rs780403251 Chr.1: 37561578-37561578 on Build GRCh38
rs772517808 Chr.1: 37561768-37561768 on Build GRCh38
rs764628424 Chr.1: 37561524-37561525 on Build GRCh38
rs571619096 Chr.1: 37561651-37561651 on Build GRCh38
rs141419180 Chr.1: 37561714-37561714 on Build GRCh38
rs976289855 Chr.1: 37561854-37561854 on Build GRCh38
rs774956779 Chr.1: 37561686-37561686 on Build GRCh38
rs532698995 Chr.1: 37561678-37561678 on Build GRCh38
rs1030276196 Chr.1: 37561855-37561855 on Build GRCh38
rs552628321 Chr.1: 37561880-37561880 on Build GRCh38
rs113025994 Chr.1: 37561793-37561793 on Build GRCh38
rs16824234 Chr.1: 37561774-37561774 on Build GRCh38
rs745733545 Chr.1: 37561631-37561631 on Build GRCh38
rs983490382 Chr.1: 37561870-37561870 on Build GRCh38
rs909447290 Chr.1: 37561515-37561515 on Build GRCh38
rs377679738 Chr.1: 37561642-37561642 on Build GRCh38
rs755415221 Chr.1: 37561649-37561649 on Build GRCh38
rs146521352 Chr.1: 37561716-37561716 on Build GRCh38
rs767259764 Chr.1: 37561640-37561641 on Build GRCh38
rs896589802 Chr.1: 37561539-37561539 on Build GRCh38
rs147235049 Chr.1: 37561663-37561663 on Build GRCh38
rs550154548 Chr.1: 37561643-37561643 on Build GRCh38
rs186497242 Chr.1: 37561946-37561946 on Build GRCh38
rs964592054 Chr.1: 37561850-37561850 on Build GRCh38
rs770923052 Chr.1: 37561622-37561622 on Build GRCh38
rs764522053 Chr.1: 37561775-37561775 on Build GRCh38
rs755768380 Chr.1: 37561656-37561656 on Build GRCh38
rs766211268 Chr.1: 37561727-37561727 on Build GRCh38
rs149439344 Chr.1: 37561634-37561634 on Build GRCh38
rs1011276080 Chr.1: 37561833-37561833 on Build GRCh38
rs746866631 Chr.1: 37561744-37561744 on Build GRCh38
rs779963478 Chr.1: 37561743-37561743 on Build GRCh38
rs971089694 Chr.1: 37561553-37561553 on Build GRCh38
rs745761540 Chr.1: 37561672-37561672 on Build GRCh38
rs765317340 Chr.1: 37561638-37561638 on Build GRCh38
rs753041062 Chr.1: 37561706-37561706 on Build GRCh38
rs771741737 Chr.1: 37561632-37561632 on Build GRCh38

View Full Product Details