Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37576316-37576785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL2
Entrez Gene ID
29889
Gene Name
G protein nucleolar 2
Gene Aliases
HUMAUANTIG, NGP1, Ngp-1, Nog2, Nug2
UniGene
Hs.75528
Gene Chromosome Location
Chr.1: 37566812-37595985 on Build GRCh38
Transcripts
NM_001323623.1 NM_001323624.1 NM_013285.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5451317 c.992G>A p.R331H Chr.1: 37576474-37576474 on Build GRCh38
6500868 c.991C>T p.R331C Chr.1: 37576475-37576475 on Build GRCh38
2167339 c.1037_1038insA p.V347fs*15 Chr.1: 37576428-37576429 on Build GRCh38
909014 c.913C>T p.H305Y Chr.1: 37576553-37576553 on Build GRCh38
3489212 c.999G>A p.K333K Chr.1: 37576467-37576467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762059672 Chr.1: 37576470-37576470 on Build GRCh38
rs895317918 Chr.1: 37576699-37576699 on Build GRCh38
rs769918756 Chr.1: 37576348-37576348 on Build GRCh38
rs748503699 Chr.1: 37576514-37576514 on Build GRCh38
rs565624574 Chr.1: 37576362-37576362 on Build GRCh38
rs375264996 Chr.1: 37576583-37576583 on Build GRCh38
rs1047992908 Chr.1: 37576610-37576610 on Build GRCh38
rs891395193 Chr.1: 37576748-37576748 on Build GRCh38
rs760534262 Chr.1: 37576576-37576576 on Build GRCh38
rs748867114 Chr.1: 37576520-37576520 on Build GRCh38
rs944474255 Chr.1: 37576494-37576494 on Build GRCh38
rs186543210 Chr.1: 37576577-37576577 on Build GRCh38
rs190951209 Chr.1: 37576746-37576746 on Build GRCh38
rs780414444 Chr.1: 37576412-37576412 on Build GRCh38
rs775493630 Chr.1: 37576568-37576568 on Build GRCh38
rs762353423 Chr.1: 37576439-37576439 on Build GRCh38
rs781467964 Chr.1: 37576513-37576513 on Build GRCh38
rs757439118 Chr.1: 37576398-37576398 on Build GRCh38
rs990417221 Chr.1: 37576452-37576452 on Build GRCh38
rs546061104 Chr.1: 37576744-37576744 on Build GRCh38
rs751960084 Chr.1: 37576479-37576479 on Build GRCh38
rs890084417 Chr.1: 37576475-37576475 on Build GRCh38
rs886660744 Chr.1: 37576622-37576622 on Build GRCh38
rs769294233 Chr.1: 37576424-37576424 on Build GRCh38
rs754076899 Chr.1: 37576395-37576395 on Build GRCh38
rs766939284 Chr.1: 37576477-37576477 on Build GRCh38
rs141806224 Chr.1: 37576442-37576442 on Build GRCh38
rs375170017 Chr.1: 37576423-37576423 on Build GRCh38
rs922937926 Chr.1: 37576543-37576543 on Build GRCh38
rs575899245 Chr.1: 37576566-37576566 on Build GRCh38
rs771998184 Chr.1: 37576550-37576550 on Build GRCh38
rs768861763 Chr.1: 37576417-37576417 on Build GRCh38
rs563137550 Chr.1: 37576648-37576648 on Build GRCh38
rs774128082 Chr.1: 37576445-37576445 on Build GRCh38
rs780844756 Chr.1: 37576418-37576418 on Build GRCh38
rs755382724 Chr.1: 37576486-37576486 on Build GRCh38
rs537747530 Chr.1: 37576354-37576354 on Build GRCh38
rs752770880 Chr.1: 37576414-37576414 on Build GRCh38
rs745739016 Chr.1: 37576542-37576542 on Build GRCh38
rs143930447 Chr.1: 37576738-37576738 on Build GRCh38
rs774370676 Chr.1: 37576535-37576535 on Build GRCh38
rs367824963 Chr.1: 37576406-37576406 on Build GRCh38
rs534836182 Chr.1: 37576416-37576416 on Build GRCh38
rs772772644 Chr.1: 37576434-37576434 on Build GRCh38
rs371779270 Chr.1: 37576515-37576515 on Build GRCh38
rs766310974 Chr.1: 37576587-37576587 on Build GRCh38
rs752111692 Chr.1: 37576588-37576588 on Build GRCh38
rs368064874 Chr.1: 37576448-37576448 on Build GRCh38
rs544829956 Chr.1: 37576729-37576729 on Build GRCh38
rs1035039684 Chr.1: 37576667-37576667 on Build GRCh38
rs755499723 Chr.1: 37576589-37576589 on Build GRCh38
rs779079142 Chr.1: 37576402-37576402 on Build GRCh38
rs770309135 Chr.1: 37576533-37576533 on Build GRCh38
rs759409971 Chr.1: 37576447-37576447 on Build GRCh38
rs979624572 Chr.1: 37576756-37576756 on Build GRCh38
rs946846657 Chr.1: 37576413-37576413 on Build GRCh38
rs577154570 Chr.1: 37576405-37576405 on Build GRCh38
rs1037859925 Chr.1: 37576665-37576665 on Build GRCh38
rs932938993 Chr.1: 37576573-37576573 on Build GRCh38
rs1012270431 Chr.1: 37576404-37576404 on Build GRCh38
rs374280864 Chr.1: 37576382-37576382 on Build GRCh38
rs201288080 Chr.1: 37576581-37576581 on Build GRCh38
rs764418451 Chr.1: 37576385-37576385 on Build GRCh38
rs775607774 Chr.1: 37576692-37576692 on Build GRCh38
rs750589653 Chr.1: 37576474-37576474 on Build GRCh38
rs767995591 Chr.1: 37576598-37576598 on Build GRCh38
rs770008051 Chr.1: 37576409-37576410 on Build GRCh38
rs112886394 Chr.1: 37576358-37576358 on Build GRCh38
rs140122153 Chr.1: 37576455-37576455 on Build GRCh38

View Full Product Details