Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37692618-37693109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA8
Entrez Gene ID
55143
Gene Name
cell division cycle associated 8
Gene Aliases
BOR, BOREALIN, DasraB, MESRGP
UniGene
Hs.524571
Gene Chromosome Location
Chr.1: 37692401-37709719 on Build GRCh38
Transcripts
NM_001256875.1 NM_018101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
680840 c.180G>T p.R60R Chr.1: 37692990-37692990 on Build GRCh38
5687195 c.12G>T p.R4S Chr.1: 37692702-37692702 on Build GRCh38
6863337 c.157C>G p.L53V Chr.1: 37692967-37692967 on Build GRCh38
7360432 c.104G>T p.R35L Chr.1: 37692914-37692914 on Build GRCh38
6210043 c.152A>C p.D51A Chr.1: 37692962-37692962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771261679 Chr.1: 37692724-37692724 on Build GRCh38
rs886135934 Chr.1: 37692805-37692805 on Build GRCh38
rs758966650 Chr.1: 37693075-37693075 on Build GRCh38
rs371464994 Chr.1: 37692884-37692884 on Build GRCh38
rs368533643 Chr.1: 37692725-37692725 on Build GRCh38
rs547981035 Chr.1: 37693051-37693051 on Build GRCh38
rs371780976 Chr.1: 37692893-37692893 on Build GRCh38
rs369811875 Chr.1: 37693033-37693033 on Build GRCh38
rs754788952 Chr.1: 37692896-37692896 on Build GRCh38
rs767204470 Chr.1: 37692648-37692648 on Build GRCh38
rs756131181 Chr.1: 37692823-37692823 on Build GRCh38
rs376553755 Chr.1: 37692686-37692686 on Build GRCh38
rs11558853 Chr.1: 37692673-37692673 on Build GRCh38
rs148168022 Chr.1: 37692975-37692975 on Build GRCh38
rs893062282 Chr.1: 37692742-37692742 on Build GRCh38
rs749828490 Chr.1: 37692834-37692834 on Build GRCh38
rs770780603 Chr.1: 37692994-37692994 on Build GRCh38
rs577147430 Chr.1: 37692976-37692976 on Build GRCh38
rs373741354 Chr.1: 37692674-37692674 on Build GRCh38
rs761888225 Chr.1: 37692948-37692948 on Build GRCh38
rs753696028 Chr.1: 37692668-37692668 on Build GRCh38
rs991001815 Chr.1: 37692849-37692849 on Build GRCh38
rs201410024 Chr.1: 37692744-37692744 on Build GRCh38
rs765850249 Chr.1: 37693060-37693060 on Build GRCh38
rs764094289 Chr.1: 37692763-37692763 on Build GRCh38
rs532376046 Chr.1: 37692837-37692837 on Build GRCh38
rs17851453 Chr.1: 37692726-37692726 on Build GRCh38
rs764120288 Chr.1: 37692906-37692906 on Build GRCh38
rs1034797886 Chr.1: 37693085-37693085 on Build GRCh38
rs755241952 Chr.1: 37692817-37692817 on Build GRCh38
rs936678581 Chr.1: 37692701-37692701 on Build GRCh38
rs775164073 Chr.1: 37692756-37692756 on Build GRCh38
rs768323749 Chr.1: 37692961-37692961 on Build GRCh38
rs753936003 Chr.1: 37692913-37692913 on Build GRCh38
rs112129534 Chr.1: 37693084-37693084 on Build GRCh38
rs746188983 Chr.1: 37692861-37692861 on Build GRCh38
rs367778850 Chr.1: 37692878-37692878 on Build GRCh38
rs371414920 Chr.1: 37692862-37692862 on Build GRCh38
rs780601986 Chr.1: 37693076-37693076 on Build GRCh38
rs371566364 Chr.1: 37693086-37693086 on Build GRCh38
rs766636904 Chr.1: 37692684-37692684 on Build GRCh38
rs759253865 Chr.1: 37692647-37692647 on Build GRCh38
rs779438508 Chr.1: 37692931-37692931 on Build GRCh38
rs757730266 Chr.1: 37692914-37692914 on Build GRCh38
rs966430697 Chr.1: 37692856-37692856 on Build GRCh38
rs200641284 Chr.1: 37693070-37693070 on Build GRCh38
rs758775604 Chr.1: 37692938-37692938 on Build GRCh38
rs749680803 Chr.1: 37692721-37692721 on Build GRCh38
rs1047973605 Chr.1: 37693048-37693048 on Build GRCh38
rs565289223 Chr.1: 37692818-37692818 on Build GRCh38
rs764076384 Chr.1: 37692655-37692655 on Build GRCh38
rs943287088 Chr.1: 37692984-37692984 on Build GRCh38
rs543224748 Chr.1: 37692746-37692746 on Build GRCh38
rs781286827 Chr.1: 37692694-37692694 on Build GRCh38
rs550448779 Chr.1: 37692791-37692791 on Build GRCh38
rs757685321 Chr.1: 37692731-37692754 on Build GRCh38
rs949512137 Chr.1: 37692669-37692669 on Build GRCh38
rs764332205 Chr.1: 37693020-37693020 on Build GRCh38
rs757755403 Chr.1: 37692855-37692855 on Build GRCh38
rs760744832 Chr.1: 37692652-37692652 on Build GRCh38
rs901655647 Chr.1: 37692787-37692787 on Build GRCh38
rs373643046 Chr.1: 37692872-37692872 on Build GRCh38
rs754608220 Chr.1: 37693080-37693080 on Build GRCh38
rs756634145 Chr.1: 37692711-37692711 on Build GRCh38
rs1005881070 Chr.1: 37692814-37692814 on Build GRCh38
rs771828918 Chr.1: 37692751-37692751 on Build GRCh38
rs750820280 Chr.1: 37692935-37692935 on Build GRCh38
rs200041937 Chr.1: 37692759-37692759 on Build GRCh38
rs1021289280 Chr.1: 37692843-37692843 on Build GRCh38
rs748201253 Chr.1: 37692705-37692705 on Build GRCh38
rs959622013 Chr.1: 37692829-37692829 on Build GRCh38
rs755184966 Chr.1: 37692690-37692690 on Build GRCh38
rs746528094 Chr.1: 37692866-37692866 on Build GRCh38
rs778078602 Chr.1: 37692715-37692715 on Build GRCh38
rs776371441 Chr.1: 37692774-37692774 on Build GRCh38
rs751022039 Chr.1: 37693072-37693072 on Build GRCh38
rs775110691 Chr.1: 37692651-37692651 on Build GRCh38
rs780615091 Chr.1: 37692967-37692967 on Build GRCh38
rs532039712 Chr.1: 37692784-37692784 on Build GRCh38
rs762865531 Chr.1: 37692985-37692985 on Build GRCh38
rs780652027 Chr.1: 37692874-37692880 on Build GRCh38
rs760860469 Chr.1: 37693013-37693013 on Build GRCh38
rs768105558 Chr.1: 37692869-37692869 on Build GRCh38
rs779197552 Chr.1: 37692859-37692859 on Build GRCh38
rs142159015 Chr.1: 37692969-37692969 on Build GRCh38
rs923961297 Chr.1: 37692697-37692697 on Build GRCh38
rs997700945 Chr.1: 37692790-37692790 on Build GRCh38
rs751930421 Chr.1: 37693079-37693079 on Build GRCh38
rs368058384 Chr.1: 37692891-37692891 on Build GRCh38
rs769747539 Chr.1: 37692877-37692877 on Build GRCh38
rs776659425 Chr.1: 37692740-37692740 on Build GRCh38
rs142003803 Chr.1: 37693012-37693012 on Build GRCh38
rs746722077 Chr.1: 37692954-37692954 on Build GRCh38
rs375268350 Chr.1: 37692901-37692901 on Build GRCh38
rs1011424052 Chr.1: 37692838-37692838 on Build GRCh38
rs776195938 Chr.1: 37692870-37692870 on Build GRCh38
rs750186595 Chr.1: 37692813-37692813 on Build GRCh38

View Full Product Details