Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37716262-37716759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA10
Entrez Gene ID
284656
Gene Name
EPH receptor A10
Gene Aliases
-
UniGene
Hs.129435
Gene Chromosome Location
Chr.1: 37713888-37765152 on Build GRCh38
Transcripts
NM_001099439.1 XM_017001080.1

SNP Detail

SNP ID Chromosome Location
rs183049110 Chr.1: 37716651-37716651 on Build GRCh38
rs904893869 Chr.1: 37716544-37716544 on Build GRCh38
rs930926031 Chr.1: 37716526-37716526 on Build GRCh38
rs528465328 Chr.1: 37716488-37716488 on Build GRCh38
rs904511474 Chr.1: 37716686-37716686 on Build GRCh38
rs1056998939 Chr.1: 37716430-37716430 on Build GRCh38
rs891861198 Chr.1: 37716435-37716435 on Build GRCh38
rs927200779 Chr.1: 37716583-37716583 on Build GRCh38
rs900676 Chr.1: 37716463-37716463 on Build GRCh38
rs563504782 Chr.1: 37716370-37716370 on Build GRCh38
rs504518 Chr.1: 37716661-37716661 on Build GRCh38
rs557212188 Chr.1: 37716593-37716593 on Build GRCh38
rs1056156680 Chr.1: 37716589-37716589 on Build GRCh38
rs772232974 Chr.1: 37716538-37716538 on Build GRCh38
rs969082634 Chr.1: 37716407-37716407 on Build GRCh38
rs879696479 Chr.1: 37716374-37716374 on Build GRCh38
rs905674412 Chr.1: 37716417-37716417 on Build GRCh38
rs774258151 Chr.1: 37716317-37716317 on Build GRCh38
rs140074960 Chr.1: 37716655-37716655 on Build GRCh38
rs931018035 Chr.1: 37716566-37716566 on Build GRCh38
rs938596574 Chr.1: 37716618-37716618 on Build GRCh38
rs1034800215 Chr.1: 37716352-37716352 on Build GRCh38
rs997195309 Chr.1: 37716677-37716677 on Build GRCh38
rs953430032 Chr.1: 37716461-37716461 on Build GRCh38
rs769157479 Chr.1: 37716732-37716732 on Build GRCh38
rs1044691893 Chr.1: 37716574-37716574 on Build GRCh38
rs1043190064 Chr.1: 37716676-37716676 on Build GRCh38
rs747958854 Chr.1: 37716481-37716481 on Build GRCh38
rs1003277617 Chr.1: 37716347-37716347 on Build GRCh38
rs1028978791 Chr.1: 37716697-37716697 on Build GRCh38
rs542027058 Chr.1: 37716287-37716287 on Build GRCh38
rs775569875 Chr.1: 37716643-37716643 on Build GRCh38
rs35037613 Chr.1: 37716562-37716562 on Build GRCh38
rs750033198 Chr.1: 37716379-37716379 on Build GRCh38
rs533644513 Chr.1: 37716669-37716669 on Build GRCh38
rs557752622 Chr.1: 37716659-37716659 on Build GRCh38
rs1002736629 Chr.1: 37716419-37716419 on Build GRCh38
rs502776 Chr.1: 37716492-37716492 on Build GRCh38
rs978780818 Chr.1: 37716431-37716431 on Build GRCh38
rs761455894 Chr.1: 37716457-37716457 on Build GRCh38
rs1029426653 Chr.1: 37716737-37716737 on Build GRCh38
rs774690800 Chr.1: 37716490-37716490 on Build GRCh38
rs757978080 Chr.1: 37716437-37716437 on Build GRCh38
rs535913803 Chr.1: 37716549-37716549 on Build GRCh38
rs866553187 Chr.1: 37716728-37716728 on Build GRCh38
rs568467430 Chr.1: 37716507-37716507 on Build GRCh38
rs531128878 Chr.1: 37716378-37716378 on Build GRCh38
rs2279347 Chr.1: 37716476-37716476 on Build GRCh38
rs904943009 Chr.1: 37716333-37716333 on Build GRCh38
rs553423662 Chr.1: 37716714-37716714 on Build GRCh38
rs779683866 Chr.1: 37716455-37716455 on Build GRCh38
rs574843569 Chr.1: 37716344-37716345 on Build GRCh38

View Full Product Details