Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37716639-37717145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA10
Entrez Gene ID
284656
Gene Name
EPH receptor A10
Gene Aliases
-
UniGene
Hs.129435
Gene Chromosome Location
Chr.1: 37713888-37765152 on Build GRCh38
Transcripts
NM_001099439.1 XM_017001080.1

SNP Detail

SNP ID Chromosome Location
rs866479841 Chr.1: 37716884-37716884 on Build GRCh38
rs904511474 Chr.1: 37716686-37716686 on Build GRCh38
rs751123654 Chr.1: 37717107-37717107 on Build GRCh38
rs539849065 Chr.1: 37716750-37716750 on Build GRCh38
rs939030809 Chr.1: 37716850-37716850 on Build GRCh38
rs546127004 Chr.1: 37717075-37717075 on Build GRCh38
rs926264673 Chr.1: 37716829-37716829 on Build GRCh38
rs762299254 Chr.1: 37716858-37716858 on Build GRCh38
rs542518654 Chr.1: 37717024-37717024 on Build GRCh38
rs879611097 Chr.1: 37716766-37716766 on Build GRCh38
rs972582696 Chr.1: 37717099-37717099 on Build GRCh38
rs1056100202 Chr.1: 37716851-37716851 on Build GRCh38
rs542572221 Chr.1: 37717048-37717048 on Build GRCh38
rs556960593 Chr.1: 37716870-37716870 on Build GRCh38
rs504518 Chr.1: 37716661-37716661 on Build GRCh38
rs528738252 Chr.1: 37717079-37717079 on Build GRCh38
rs980265463 Chr.1: 37717100-37717100 on Build GRCh38
rs573669089 Chr.1: 37716971-37716971 on Build GRCh38
rs1043297056 Chr.1: 37716999-37716999 on Build GRCh38
rs201170276 Chr.1: 37716786-37716787 on Build GRCh38
rs897985556 Chr.1: 37717065-37717065 on Build GRCh38
rs997195309 Chr.1: 37716677-37716677 on Build GRCh38
rs1028255918 Chr.1: 37716986-37716986 on Build GRCh38
rs769157479 Chr.1: 37716732-37716732 on Build GRCh38
rs1043190064 Chr.1: 37716676-37716676 on Build GRCh38
rs903707392 Chr.1: 37716976-37716976 on Build GRCh38
rs762547669 Chr.1: 37717023-37717023 on Build GRCh38
rs1028978791 Chr.1: 37716697-37716697 on Build GRCh38
rs533644513 Chr.1: 37716669-37716669 on Build GRCh38
rs1009099724 Chr.1: 37717031-37717031 on Build GRCh38
rs879189362 Chr.1: 37716993-37716993 on Build GRCh38
rs76466607 Chr.1: 37716984-37716984 on Build GRCh38
rs187430442 Chr.1: 37717078-37717078 on Build GRCh38
rs979766906 Chr.1: 37716853-37716853 on Build GRCh38
rs1029426653 Chr.1: 37716737-37716737 on Build GRCh38
rs890492130 Chr.1: 37717025-37717025 on Build GRCh38
rs866553187 Chr.1: 37716728-37716728 on Build GRCh38
rs1019043463 Chr.1: 37716755-37716755 on Build GRCh38
rs367820146 Chr.1: 37716765-37716765 on Build GRCh38
rs112895732 Chr.1: 37717008-37717009 on Build GRCh38
rs930893888 Chr.1: 37716801-37716801 on Build GRCh38
rs145671538 Chr.1: 37716797-37716797 on Build GRCh38
rs932062016 Chr.1: 37717012-37717012 on Build GRCh38
rs543037156 Chr.1: 37716763-37716763 on Build GRCh38
rs777120944 Chr.1: 37716808-37716808 on Build GRCh38
rs972251018 Chr.1: 37716794-37716794 on Build GRCh38
rs765499239 Chr.1: 37716944-37716944 on Build GRCh38
rs530990886 Chr.1: 37717039-37717039 on Build GRCh38
rs553423662 Chr.1: 37716714-37716714 on Build GRCh38
rs952454996 Chr.1: 37717083-37717083 on Build GRCh38

View Full Product Details