Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37719336-37719836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA10
Entrez Gene ID
284656
Gene Name
EPH receptor A10
Gene Aliases
-
UniGene
Hs.129435
Gene Chromosome Location
Chr.1: 37713888-37765152 on Build GRCh38
Transcripts
NM_001099439.1 XM_017001080.1 XM_017001081.1

SNP Detail

SNP ID Chromosome Location
rs749809109 Chr.1: 37719638-37719638 on Build GRCh38
rs3086618 Chr.1: 37719686-37719691 on Build GRCh38
rs372786497 Chr.1: 37719520-37719520 on Build GRCh38
rs777340871 Chr.1: 37719401-37719401 on Build GRCh38
rs375262595 Chr.1: 37719507-37719507 on Build GRCh38
rs1039643033 Chr.1: 37719645-37719645 on Build GRCh38
rs934460180 Chr.1: 37719802-37719802 on Build GRCh38
rs1004616062 Chr.1: 37719446-37719446 on Build GRCh38
rs138095126 Chr.1: 37719444-37719444 on Build GRCh38
rs748968137 Chr.1: 37719430-37719430 on Build GRCh38
rs767063294 Chr.1: 37719489-37719489 on Build GRCh38
rs1054682031 Chr.1: 37719812-37719812 on Build GRCh38
rs781456516 Chr.1: 37719532-37719532 on Build GRCh38
rs540944109 Chr.1: 37719715-37719715 on Build GRCh38
rs956776760 Chr.1: 37719565-37719565 on Build GRCh38
rs377304713 Chr.1: 37719529-37719529 on Build GRCh38
rs777654930 Chr.1: 37719428-37719428 on Build GRCh38
rs763173206 Chr.1: 37719466-37719466 on Build GRCh38
rs971476167 Chr.1: 37719746-37719746 on Build GRCh38
rs760777175 Chr.1: 37719393-37719393 on Build GRCh38
rs956670099 Chr.1: 37719791-37719794 on Build GRCh38
rs763651960 Chr.1: 37719570-37719570 on Build GRCh38
rs1016455110 Chr.1: 37719461-37719461 on Build GRCh38
rs746123800 Chr.1: 37719374-37719374 on Build GRCh38
rs373587096 Chr.1: 37719406-37719406 on Build GRCh38
rs1038552817 Chr.1: 37719712-37719713 on Build GRCh38
rs376550997 Chr.1: 37719409-37719409 on Build GRCh38
rs757192600 Chr.1: 37719579-37719579 on Build GRCh38
rs748175541 Chr.1: 37719534-37719534 on Build GRCh38
rs572328372 Chr.1: 37719437-37719437 on Build GRCh38
rs760165773 Chr.1: 37719713-37719713 on Build GRCh38
rs142918194 Chr.1: 37719395-37719396 on Build GRCh38
rs983016689 Chr.1: 37719380-37719380 on Build GRCh38
rs766755359 Chr.1: 37719467-37719467 on Build GRCh38
rs774992843 Chr.1: 37719567-37719567 on Build GRCh38
rs78866494 Chr.1: 37719724-37719724 on Build GRCh38
rs934490487 Chr.1: 37719537-37719537 on Build GRCh38
rs751281419 Chr.1: 37719482-37719482 on Build GRCh38
rs775641899 Chr.1: 37719386-37719386 on Build GRCh38
rs1017087730 Chr.1: 37719723-37719723 on Build GRCh38
rs150031151 Chr.1: 37719732-37719733 on Build GRCh38
rs899851873 Chr.1: 37719663-37719663 on Build GRCh38
rs755206454 Chr.1: 37719530-37719530 on Build GRCh38
rs1045901654 Chr.1: 37719523-37719523 on Build GRCh38
rs368098004 Chr.1: 37719487-37719487 on Build GRCh38
rs777880005 Chr.1: 37719634-37719634 on Build GRCh38
rs201129658 Chr.1: 37719571-37719571 on Build GRCh38
rs754542097 Chr.1: 37719413-37719413 on Build GRCh38
rs1047784760 Chr.1: 37719664-37719664 on Build GRCh38
rs748392542 Chr.1: 37719628-37719628 on Build GRCh38
rs765686994 Chr.1: 37719403-37719403 on Build GRCh38
rs518583 Chr.1: 37719665-37719665 on Build GRCh38
rs368522699 Chr.1: 37719517-37719517 on Build GRCh38
rs543219786 Chr.1: 37719541-37719541 on Build GRCh38
rs750207192 Chr.1: 37719590-37719590 on Build GRCh38
rs762701567 Chr.1: 37719407-37719407 on Build GRCh38
rs770350297 Chr.1: 37719538-37719538 on Build GRCh38
rs752672196 Chr.1: 37719426-37719426 on Build GRCh38
rs763878482 Chr.1: 37719502-37719502 on Build GRCh38
rs200423720 Chr.1: 37719540-37719540 on Build GRCh38
rs397839472 Chr.1: 37719692-37719697 on Build GRCh38
rs776825559 Chr.1: 37719445-37719445 on Build GRCh38
rs774676111 Chr.1: 37719547-37719547 on Build GRCh38
rs369198752 Chr.1: 37719715-37719716 on Build GRCh38
rs760160351 Chr.1: 37719569-37719569 on Build GRCh38
rs991721572 Chr.1: 37719731-37719731 on Build GRCh38
rs375221623 Chr.1: 37719602-37719602 on Build GRCh38
rs770164053 Chr.1: 37719456-37719456 on Build GRCh38
rs35909785 Chr.1: 37719497-37719497 on Build GRCh38
rs756216586 Chr.1: 37719630-37719630 on Build GRCh38
rs900677 Chr.1: 37719737-37719737 on Build GRCh38
rs757064255 Chr.1: 37719515-37719515 on Build GRCh38
rs768727314 Chr.1: 37719439-37719440 on Build GRCh38
rs915603480 Chr.1: 37719449-37719449 on Build GRCh38
rs571247403 Chr.1: 37719385-37719385 on Build GRCh38
rs370503923 Chr.1: 37719747-37719748 on Build GRCh38
rs563374536 Chr.1: 37719764-37719764 on Build GRCh38
rs746173401 Chr.1: 37719659-37719659 on Build GRCh38
rs771379554 Chr.1: 37719643-37719643 on Build GRCh38
rs989682155 Chr.1: 37719811-37719811 on Build GRCh38
rs554160164 Chr.1: 37719410-37719410 on Build GRCh38
rs370268519 Chr.1: 37719438-37719438 on Build GRCh38
rs755956421 Chr.1: 37719427-37719427 on Build GRCh38
rs772144361 Chr.1: 37719375-37719375 on Build GRCh38
rs773634070 Chr.1: 37719457-37719457 on Build GRCh38
rs767237675 Chr.1: 37719522-37719522 on Build GRCh38
rs760592748 Chr.1: 37719393-37719401 on Build GRCh38
rs559333164 Chr.1: 37719716-37719716 on Build GRCh38
rs781509600 Chr.1: 37719626-37719626 on Build GRCh38
rs199558258 Chr.1: 37719566-37719566 on Build GRCh38
rs776848180 Chr.1: 37719538-37719541 on Build GRCh38
rs149811548 Chr.1: 37719698-37719699 on Build GRCh38
rs756079971 Chr.1: 37719501-37719501 on Build GRCh38
rs1030163258 Chr.1: 37719679-37719679 on Build GRCh38
rs768840008 Chr.1: 37719400-37719400 on Build GRCh38
rs201284300 Chr.1: 37719679-37719680 on Build GRCh38
rs762374343 Chr.1: 37719402-37719402 on Build GRCh38
rs765143179 Chr.1: 37719580-37719580 on Build GRCh38
rs1011895356 Chr.1: 37719474-37719474 on Build GRCh38
rs915234192 Chr.1: 37719365-37719365 on Build GRCh38
rs924350852 Chr.1: 37719795-37719795 on Build GRCh38
rs369507793 Chr.1: 37719655-37719655 on Build GRCh38
rs747227181 Chr.1: 37719439-37719439 on Build GRCh38

View Full Product Details