Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37997877-37998385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL3
Entrez Gene ID
2275
Gene Name
four and a half LIM domains 3
Gene Aliases
SLIM2
UniGene
Hs.57687
Gene Chromosome Location
Chr.1: 37996765-38005515 on Build GRCh38
Transcripts
NM_001243878.1 NM_004468.4 XM_017000675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6663938 c.482G>A p.R161H Chr.1: 37997982-37997982 on Build GRCh38
909093 c.402G>A p.Q134Q Chr.1: 37998062-37998062 on Build GRCh38
6663939 c.430G>A p.D144N Chr.1: 37998034-37998034 on Build GRCh38
535000 c.427C>T p.P143S Chr.1: 37998037-37998037 on Build GRCh38
5423851 c.482G>C p.R161P Chr.1: 37997982-37997982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769779355 Chr.1: 37997942-37997942 on Build GRCh38
rs752590357 Chr.1: 37998063-37998068 on Build GRCh38
rs767156444 Chr.1: 37998057-37998061 on Build GRCh38
rs551683092 Chr.1: 37998329-37998329 on Build GRCh38
rs112743291 Chr.1: 37997962-37997962 on Build GRCh38
rs765355705 Chr.1: 37998049-37998049 on Build GRCh38
rs370875801 Chr.1: 37998287-37998287 on Build GRCh38
rs759264091 Chr.1: 37997933-37997934 on Build GRCh38
rs777013710 Chr.1: 37998127-37998127 on Build GRCh38
rs147927855 Chr.1: 37997978-37997978 on Build GRCh38
rs1046502323 Chr.1: 37998311-37998311 on Build GRCh38
rs754673207 Chr.1: 37998014-37998014 on Build GRCh38
rs1046923759 Chr.1: 37998351-37998351 on Build GRCh38
rs998671748 Chr.1: 37998354-37998355 on Build GRCh38
rs773389055 Chr.1: 37998139-37998139 on Build GRCh38
rs1017327506 Chr.1: 37998148-37998149 on Build GRCh38
rs766773447 Chr.1: 37998061-37998061 on Build GRCh38
rs746282274 Chr.1: 37997925-37997925 on Build GRCh38
rs761995906 Chr.1: 37998048-37998048 on Build GRCh38
rs777587205 Chr.1: 37998027-37998027 on Build GRCh38
rs924191692 Chr.1: 37998063-37998063 on Build GRCh38
rs758720934 Chr.1: 37997930-37997930 on Build GRCh38
rs759232548 Chr.1: 37998040-37998040 on Build GRCh38
rs1053477961 Chr.1: 37998080-37998080 on Build GRCh38
rs758707411 Chr.1: 37997988-37997988 on Build GRCh38
rs762663278 Chr.1: 37997955-37997955 on Build GRCh38
rs557494177 Chr.1: 37998271-37998271 on Build GRCh38
rs779582302 Chr.1: 37997931-37997931 on Build GRCh38
rs773131228 Chr.1: 37997954-37997954 on Build GRCh38
rs369957166 Chr.1: 37998165-37998165 on Build GRCh38
rs1040792526 Chr.1: 37998133-37998133 on Build GRCh38
rs371636908 Chr.1: 37997900-37997900 on Build GRCh38
rs895239579 Chr.1: 37997976-37997976 on Build GRCh38
rs989728952 Chr.1: 37998292-37998292 on Build GRCh38
rs150056091 Chr.1: 37998102-37998102 on Build GRCh38
rs998724140 Chr.1: 37998022-37998022 on Build GRCh38
rs369640388 Chr.1: 37998020-37998020 on Build GRCh38
rs72923731 Chr.1: 37998272-37998272 on Build GRCh38
rs745727880 Chr.1: 37998118-37998118 on Build GRCh38
rs769872653 Chr.1: 37998136-37998136 on Build GRCh38
rs555483030 Chr.1: 37998318-37998318 on Build GRCh38
rs776158550 Chr.1: 37997937-37997937 on Build GRCh38
rs768081279 Chr.1: 37997936-37997936 on Build GRCh38
rs773772156 Chr.1: 37998337-37998337 on Build GRCh38
rs143651640 Chr.1: 37997914-37997914 on Build GRCh38
rs756167023 Chr.1: 37997928-37997931 on Build GRCh38
rs776923869 Chr.1: 37998045-37998045 on Build GRCh38
rs201163097 Chr.1: 37998068-37998068 on Build GRCh38
rs763635111 Chr.1: 37998182-37998182 on Build GRCh38
rs942310936 Chr.1: 37998301-37998301 on Build GRCh38
rs942017453 Chr.1: 37998286-37998286 on Build GRCh38
rs769060688 Chr.1: 37998044-37998044 on Build GRCh38
rs550454324 Chr.1: 37998237-37998237 on Build GRCh38
rs528129937 Chr.1: 37998245-37998245 on Build GRCh38
rs942466439 Chr.1: 37998112-37998112 on Build GRCh38
rs568848347 Chr.1: 37998265-37998265 on Build GRCh38
rs375305856 Chr.1: 37997982-37997982 on Build GRCh38
rs766974446 Chr.1: 37998145-37998145 on Build GRCh38
rs780423046 Chr.1: 37998003-37998003 on Build GRCh38
rs754432586 Chr.1: 37997916-37997916 on Build GRCh38
rs539230422 Chr.1: 37998270-37998270 on Build GRCh38
rs376716946 Chr.1: 37998074-37998074 on Build GRCh38
rs1041196100 Chr.1: 37998324-37998324 on Build GRCh38
rs775597521 Chr.1: 37997971-37997971 on Build GRCh38
rs113413826 Chr.1: 37998065-37998065 on Build GRCh38
rs373149981 Chr.1: 37998035-37998035 on Build GRCh38
rs778706057 Chr.1: 37998113-37998113 on Build GRCh38
rs752378172 Chr.1: 37998166-37998166 on Build GRCh38
rs202242166 Chr.1: 37998026-37998026 on Build GRCh38
rs372470458 Chr.1: 37998174-37998174 on Build GRCh38
rs929814194 Chr.1: 37998336-37998336 on Build GRCh38
rs895196068 Chr.1: 37998294-37998297 on Build GRCh38
rs766139864 Chr.1: 37997960-37997960 on Build GRCh38
rs112738307 Chr.1: 37998134-37998134 on Build GRCh38
rs779544224 Chr.1: 37997923-37997923 on Build GRCh38
rs746620576 Chr.1: 37997935-37997935 on Build GRCh38
rs535892413 Chr.1: 37998204-37998204 on Build GRCh38
rs200186953 Chr.1: 37998132-37998132 on Build GRCh38
rs111280408 Chr.1: 37998013-37998013 on Build GRCh38
rs774652683 Chr.1: 37998158-37998158 on Build GRCh38
rs755644231 Chr.1: 37998101-37998101 on Build GRCh38
rs1016318708 Chr.1: 37997915-37997915 on Build GRCh38
rs372151784 Chr.1: 37997977-37997977 on Build GRCh38
rs201132824 Chr.1: 37997973-37997973 on Build GRCh38
rs770955273 Chr.1: 37998107-37998107 on Build GRCh38
rs757882807 Chr.1: 37997918-37997918 on Build GRCh38
rs34220308 Chr.1: 37997907-37997908 on Build GRCh38
rs201707111 Chr.1: 37998034-37998034 on Build GRCh38
rs750920418 Chr.1: 37998053-37998053 on Build GRCh38
rs780828480 Chr.1: 37998072-37998072 on Build GRCh38
rs771932815 Chr.1: 37998126-37998126 on Build GRCh38
rs759499164 Chr.1: 37998295-37998295 on Build GRCh38
rs749200855 Chr.1: 37998030-37998030 on Build GRCh38
rs770280329 Chr.1: 37998209-37998209 on Build GRCh38
rs767468643 Chr.1: 37998307-37998307 on Build GRCh38
rs991318672 Chr.1: 37998238-37998238 on Build GRCh38
rs888337165 Chr.1: 37998183-37998183 on Build GRCh38
rs959355524 Chr.1: 37998232-37998232 on Build GRCh38

View Full Product Details