Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38005488-38006001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL3
Entrez Gene ID
2275
Gene Name
four and a half LIM domains 3
Gene Aliases
SLIM2
UniGene
Hs.57687
Gene Chromosome Location
Chr.1: 37996765-38005515 on Build GRCh38
Transcripts
NM_001243878.1 NM_004468.4

SNP Detail

SNP ID Chromosome Location
rs372760725 Chr.1: 38005895-38005895 on Build GRCh38
rs935728855 Chr.1: 38005724-38005724 on Build GRCh38
rs939679380 Chr.1: 38005901-38005901 on Build GRCh38
rs899925110 Chr.1: 38005600-38005600 on Build GRCh38
rs558726541 Chr.1: 38005825-38005825 on Build GRCh38
rs992723938 Chr.1: 38005576-38005576 on Build GRCh38
rs573361377 Chr.1: 38005789-38005789 on Build GRCh38
rs1051253324 Chr.1: 38005880-38005880 on Build GRCh38
rs529237828 Chr.1: 38005936-38005936 on Build GRCh38
rs917208022 Chr.1: 38005583-38005583 on Build GRCh38
rs554127864 Chr.1: 38005560-38005561 on Build GRCh38
rs886788518 Chr.1: 38005886-38005886 on Build GRCh38
rs546195671 Chr.1: 38005565-38005565 on Build GRCh38
rs138931272 Chr.1: 38005719-38005719 on Build GRCh38
rs994471284 Chr.1: 38005828-38005828 on Build GRCh38
rs969907876 Chr.1: 38005609-38005609 on Build GRCh38
rs913875792 Chr.1: 38005522-38005522 on Build GRCh38
rs1052888487 Chr.1: 38005749-38005749 on Build GRCh38
rs1027321881 Chr.1: 38005836-38005836 on Build GRCh38
rs890328642 Chr.1: 38005744-38005744 on Build GRCh38
rs1050681949 Chr.1: 38005736-38005736 on Build GRCh38
rs960723217 Chr.1: 38005571-38005571 on Build GRCh38
rs980309712 Chr.1: 38005620-38005620 on Build GRCh38
rs914167166 Chr.1: 38005958-38005958 on Build GRCh38
rs946715364 Chr.1: 38005559-38005559 on Build GRCh38
rs995969097 Chr.1: 38005661-38005661 on Build GRCh38
rs912886727 Chr.1: 38005756-38005756 on Build GRCh38
rs561889413 Chr.1: 38005935-38005935 on Build GRCh38
rs921432527 Chr.1: 38005867-38005867 on Build GRCh38
rs898271723 Chr.1: 38005977-38005977 on Build GRCh38
rs961660270 Chr.1: 38005798-38005798 on Build GRCh38
rs1009216206 Chr.1: 38005769-38005769 on Build GRCh38
rs969109284 Chr.1: 38005850-38005850 on Build GRCh38
rs1038427311 Chr.1: 38005939-38005939 on Build GRCh38
rs980582517 Chr.1: 38005858-38005859 on Build GRCh38
rs988773375 Chr.1: 38005904-38005904 on Build GRCh38
rs768743141 Chr.1: 38005821-38005821 on Build GRCh38
rs934249355 Chr.1: 38005879-38005879 on Build GRCh38
rs540718936 Chr.1: 38005807-38005807 on Build GRCh38
rs925730657 Chr.1: 38005570-38005571 on Build GRCh38

View Full Product Details