Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38875734-38876270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA9
Entrez Gene ID
81025
Gene Name
gap junction protein alpha 9
Gene Aliases
CX58, CX59, GJA10
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38874067-38881626 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Gene Symbol
GJA9-MYCBP
Entrez Gene ID
100527950
Gene Name
GJA9-MYCBP readthrough
Gene Aliases
-
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38862490-38881626 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Gene Symbol
LOC105378663
Entrez Gene ID
105378663
Gene Name
uncharacterized LOC105378663
Gene Aliases
-
UniGene
Hs.585419
Gene Chromosome Location
Chr.1: 38873401-38876226 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622870 c.196T>C p.Y66H Chr.1: 38875903-38875903 on Build GRCh38
213987 c.296G>A p.R99Q Chr.1: 38875803-38875803 on Build GRCh38
4577312 c.198C>T p.Y66Y Chr.1: 38875901-38875901 on Build GRCh38
1728852 c.285T>A p.H95Q Chr.1: 38875814-38875814 on Build GRCh38
6575312 c.90C>T p.F30F Chr.1: 38876009-38876009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747759782 Chr.1: 38876096-38876096 on Build GRCh38
rs762871063 Chr.1: 38875826-38875826 on Build GRCh38
rs143122810 Chr.1: 38875811-38875811 on Build GRCh38
rs762926129 Chr.1: 38875920-38875920 on Build GRCh38
rs756966331 Chr.1: 38875850-38875850 on Build GRCh38
rs753546933 Chr.1: 38875818-38875827 on Build GRCh38
rs772779659 Chr.1: 38876004-38876004 on Build GRCh38
rs946381668 Chr.1: 38876051-38876051 on Build GRCh38
rs61732191 Chr.1: 38876077-38876077 on Build GRCh38
rs192931754 Chr.1: 38876207-38876207 on Build GRCh38
rs954962882 Chr.1: 38875827-38875827 on Build GRCh38
rs773304022 Chr.1: 38875910-38875910 on Build GRCh38
rs760275668 Chr.1: 38875804-38875804 on Build GRCh38
rs771780519 Chr.1: 38876144-38876144 on Build GRCh38
rs1030532243 Chr.1: 38876097-38876097 on Build GRCh38
rs755193871 Chr.1: 38876115-38876115 on Build GRCh38
rs866186693 Chr.1: 38875778-38875778 on Build GRCh38
rs377340906 Chr.1: 38875972-38875972 on Build GRCh38
rs766227667 Chr.1: 38875830-38875830 on Build GRCh38
rs774733173 Chr.1: 38876187-38876187 on Build GRCh38
rs375613201 Chr.1: 38876046-38876046 on Build GRCh38
rs151296913 Chr.1: 38875868-38875868 on Build GRCh38
rs200580999 Chr.1: 38875983-38875983 on Build GRCh38
rs764198005 Chr.1: 38875937-38875937 on Build GRCh38
rs781523950 Chr.1: 38876139-38876139 on Build GRCh38
rs781561295 Chr.1: 38876094-38876094 on Build GRCh38
rs750293236 Chr.1: 38875825-38875825 on Build GRCh38
rs765256162 Chr.1: 38875957-38875957 on Build GRCh38
rs771335080 Chr.1: 38875777-38875777 on Build GRCh38
rs751995734 Chr.1: 38876082-38876082 on Build GRCh38
rs757826964 Chr.1: 38875858-38875858 on Build GRCh38
rs148850123 Chr.1: 38875844-38875844 on Build GRCh38
rs910567333 Chr.1: 38875808-38875808 on Build GRCh38
rs1030499677 Chr.1: 38875819-38875819 on Build GRCh38
rs779358239 Chr.1: 38875861-38875861 on Build GRCh38
rs543180516 Chr.1: 38875765-38875765 on Build GRCh38
rs973391142 Chr.1: 38875909-38875909 on Build GRCh38
rs765307533 Chr.1: 38876060-38876060 on Build GRCh38
rs202056633 Chr.1: 38875900-38875900 on Build GRCh38
rs753987673 Chr.1: 38875943-38875943 on Build GRCh38
rs199814335 Chr.1: 38876108-38876108 on Build GRCh38
rs771749345 Chr.1: 38875795-38875795 on Build GRCh38
rs981950283 Chr.1: 38875970-38875970 on Build GRCh38
rs769442681 Chr.1: 38876002-38876002 on Build GRCh38
rs778778259 Chr.1: 38875897-38875897 on Build GRCh38
rs774282927 Chr.1: 38875925-38875925 on Build GRCh38
rs142342464 Chr.1: 38876021-38876021 on Build GRCh38
rs540022787 Chr.1: 38876103-38876103 on Build GRCh38
rs373082020 Chr.1: 38876066-38876066 on Build GRCh38
rs963632125 Chr.1: 38876173-38876173 on Build GRCh38
rs758838655 Chr.1: 38875969-38875969 on Build GRCh38
rs777973040 Chr.1: 38875842-38875842 on Build GRCh38
rs780678258 Chr.1: 38875987-38875987 on Build GRCh38
rs1032182624 Chr.1: 38876040-38876040 on Build GRCh38
rs201757000 Chr.1: 38875901-38875901 on Build GRCh38
rs374468698 Chr.1: 38876142-38876142 on Build GRCh38
rs755330574 Chr.1: 38876089-38876089 on Build GRCh38
rs780466744 Chr.1: 38875978-38875978 on Build GRCh38
rs758941326 Chr.1: 38875924-38875924 on Build GRCh38
rs776744931 Chr.1: 38876041-38876041 on Build GRCh38
rs772196845 Chr.1: 38876106-38876107 on Build GRCh38
rs927783567 Chr.1: 38875986-38875986 on Build GRCh38
rs1013236585 Chr.1: 38876001-38876001 on Build GRCh38
rs1055290795 Chr.1: 38876221-38876221 on Build GRCh38
rs774247987 Chr.1: 38876028-38876028 on Build GRCh38
rs759216370 Chr.1: 38876034-38876034 on Build GRCh38
rs549336317 Chr.1: 38876226-38876226 on Build GRCh38
rs536962551 Chr.1: 38875810-38875810 on Build GRCh38
rs377622565 Chr.1: 38876110-38876110 on Build GRCh38
rs369560277 Chr.1: 38876037-38876037 on Build GRCh38
rs761878672 Chr.1: 38875812-38875812 on Build GRCh38
rs768191273 Chr.1: 38875893-38875893 on Build GRCh38
rs780279014 Chr.1: 38876108-38876109 on Build GRCh38
rs984094201 Chr.1: 38875829-38875829 on Build GRCh38
rs746205982 Chr.1: 38875789-38875789 on Build GRCh38
rs779390361 Chr.1: 38875782-38875782 on Build GRCh38
rs745744469 Chr.1: 38876018-38876018 on Build GRCh38
rs752979539 Chr.1: 38875833-38875833 on Build GRCh38
rs780057071 Chr.1: 38876049-38876049 on Build GRCh38
rs368829532 Chr.1: 38875776-38875776 on Build GRCh38
rs894796341 Chr.1: 38875993-38875993 on Build GRCh38
rs772452202 Chr.1: 38875884-38875884 on Build GRCh38
rs777336193 Chr.1: 38875763-38875766 on Build GRCh38
rs766850745 Chr.1: 38876081-38876081 on Build GRCh38
rs1015595830 Chr.1: 38876159-38876159 on Build GRCh38
rs747787069 Chr.1: 38875996-38875996 on Build GRCh38
rs747590326 Chr.1: 38875895-38875895 on Build GRCh38
rs746681825 Chr.1: 38875888-38875888 on Build GRCh38
rs935292941 Chr.1: 38876008-38876008 on Build GRCh38
rs780341770 Chr.1: 38875886-38875886 on Build GRCh38
rs765072825 Chr.1: 38875824-38875824 on Build GRCh38
rs942227281 Chr.1: 38876213-38876213 on Build GRCh38
rs146021626 Chr.1: 38875803-38875803 on Build GRCh38
rs746337208 Chr.1: 38875879-38875879 on Build GRCh38
rs750945760 Chr.1: 38875967-38875967 on Build GRCh38
rs759991434 Chr.1: 38876222-38876222 on Build GRCh38
rs775290456 Chr.1: 38876148-38876148 on Build GRCh38
rs982149649 Chr.1: 38876227-38876227 on Build GRCh38
rs376469931 Chr.1: 38875953-38875953 on Build GRCh38
rs761012137 Chr.1: 38875935-38875935 on Build GRCh38
rs747322908 Chr.1: 38875980-38875980 on Build GRCh38

View Full Product Details