Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78991560-78992065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANT1
Entrez Gene ID
124583
Gene Name
calcium activated nucleotidase 1
Gene Aliases
DBQD, DBQD1, SCAN-1, SCAN1, SHAPY
UniGene
Hs.8859
Gene Chromosome Location
Chr.17: 78991716-79009817 on Build GRCh38
Transcripts
NM_001159772.1 NM_001159773.1 NM_138793.3 XM_005257021.1 XM_005257022.1 XM_006721683.1 XM_011524291.1 XM_011524293.1 XM_011524294.1 XM_011524295.2

SNP Detail

SNP ID Chromosome Location
rs537539803 Chr.17: 78991672-78991672 on Build GRCh38
rs111469424 Chr.17: 78991709-78991709 on Build GRCh38
rs992884665 Chr.17: 78991895-78991895 on Build GRCh38
rs1047991438 Chr.17: 78991970-78991970 on Build GRCh38
rs1019334005 Chr.17: 78991739-78991739 on Build GRCh38
rs998440550 Chr.17: 78991945-78991945 on Build GRCh38
rs969031812 Chr.17: 78991767-78991767 on Build GRCh38
rs948713644 Chr.17: 78991679-78991679 on Build GRCh38
rs886618987 Chr.17: 78991716-78991716 on Build GRCh38
rs1048035881 Chr.17: 78991887-78991889 on Build GRCh38
rs575323231 Chr.17: 78991820-78991820 on Build GRCh38
rs978708895 Chr.17: 78991797-78991797 on Build GRCh38
rs1048355852 Chr.17: 78991715-78991715 on Build GRCh38
rs1127654 Chr.17: 78991813-78991813 on Build GRCh38
rs886578068 Chr.17: 78992010-78992010 on Build GRCh38
rs553206625 Chr.17: 78991703-78991703 on Build GRCh38
rs767743350 Chr.17: 78991995-78991995 on Build GRCh38
rs184171534 Chr.17: 78991818-78991818 on Build GRCh38
rs1033008009 Chr.17: 78992012-78992012 on Build GRCh38
rs930858010 Chr.17: 78991948-78991948 on Build GRCh38
rs934698652 Chr.17: 78991623-78991623 on Build GRCh38
rs955876669 Chr.17: 78991822-78991822 on Build GRCh38
rs1045045345 Chr.17: 78991638-78991638 on Build GRCh38
rs1003892803 Chr.17: 78991727-78991727 on Build GRCh38
rs750853920 Chr.17: 78992016-78992016 on Build GRCh38
rs543886383 Chr.17: 78991914-78991914 on Build GRCh38
rs976657833 Chr.17: 78991707-78991707 on Build GRCh38
rs965126466 Chr.17: 78991742-78991742 on Build GRCh38
rs759735342 Chr.17: 78991907-78991907 on Build GRCh38
rs746625993 Chr.17: 78991633-78991633 on Build GRCh38
rs572958719 Chr.17: 78991705-78991705 on Build GRCh38
rs770771968 Chr.17: 78991652-78991652 on Build GRCh38
rs925968692 Chr.17: 78991726-78991726 on Build GRCh38
rs896005051 Chr.17: 78991611-78991611 on Build GRCh38
rs1044790463 Chr.17: 78991933-78991933 on Build GRCh38
rs1026667137 Chr.17: 78991683-78991686 on Build GRCh38
rs3744168 Chr.17: 78991681-78991681 on Build GRCh38
rs1028220573 Chr.17: 78991955-78991955 on Build GRCh38
rs1053262281 Chr.17: 78991585-78991585 on Build GRCh38
rs920641639 Chr.17: 78991946-78991946 on Build GRCh38
rs904819823 Chr.17: 78991668-78991668 on Build GRCh38
rs953927445 Chr.17: 78991968-78991968 on Build GRCh38
rs950326052 Chr.17: 78991616-78991616 on Build GRCh38
rs189295347 Chr.17: 78991699-78991699 on Build GRCh38
rs14791 Chr.17: 78991766-78991766 on Build GRCh38
rs192546263 Chr.17: 78991756-78991756 on Build GRCh38
rs762435941 Chr.17: 78991870-78991870 on Build GRCh38
rs888177794 Chr.17: 78991686-78991687 on Build GRCh38
rs539762065 Chr.17: 78991701-78991701 on Build GRCh38
rs978999441 Chr.17: 78991588-78991588 on Build GRCh38
rs770133112 Chr.17: 78991687-78991690 on Build GRCh38
rs776264858 Chr.17: 78991678-78991678 on Build GRCh38
rs930919281 Chr.17: 78991882-78991883 on Build GRCh38
rs1057240892 Chr.17: 78991637-78991637 on Build GRCh38
rs924502370 Chr.17: 78991620-78991620 on Build GRCh38
rs996896433 Chr.17: 78991744-78991744 on Build GRCh38
rs1015420400 Chr.17: 78991688-78991688 on Build GRCh38
rs948807964 Chr.17: 78991912-78991912 on Build GRCh38
rs1044621266 Chr.17: 78991865-78991865 on Build GRCh38
rs760876325 Chr.17: 78992033-78992033 on Build GRCh38

View Full Product Details