Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79022827-79023322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF1
Entrez Gene ID
114897
Gene Name
C1q and tumor necrosis factor related protein 1
Gene Aliases
CTRP1, GIP, ZSIG37
UniGene
Hs.201398
Gene Chromosome Location
Chr.17: 79022934-79049788 on Build GRCh38
Transcripts
NM_153372.1 NR_040018.1 NR_040019.1

Gene Symbol
C1QTNF1-AS1
Entrez Gene ID
100507410
Gene Name
C1QTNF1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.666723
Gene Chromosome Location
Chr.17: 79019209-79027655 on Build GRCh38
Transcripts
NM_153372.1 NR_040018.1 NR_040019.1

SNP Detail

SNP ID Chromosome Location
rs566634753 Chr.17: 79022934-79022934 on Build GRCh38
rs539692397 Chr.17: 79023133-79023133 on Build GRCh38
rs950129373 Chr.17: 79022856-79022856 on Build GRCh38
rs557813459 Chr.17: 79023022-79023022 on Build GRCh38
rs544989571 Chr.17: 79023177-79023177 on Build GRCh38
rs1026331906 Chr.17: 79023261-79023261 on Build GRCh38
rs778734121 Chr.17: 79023056-79023056 on Build GRCh38
rs560287337 Chr.17: 79023251-79023251 on Build GRCh38
rs770491940 Chr.17: 79023120-79023120 on Build GRCh38
rs987613886 Chr.17: 79023079-79023079 on Build GRCh38
rs947323226 Chr.17: 79022898-79022898 on Build GRCh38
rs76280673 Chr.17: 79023175-79023175 on Build GRCh38
rs908008910 Chr.17: 79023219-79023219 on Build GRCh38
rs746375295 Chr.17: 79023247-79023247 on Build GRCh38
rs568653614 Chr.17: 79022897-79022897 on Build GRCh38
rs77696401 Chr.17: 79022924-79022924 on Build GRCh38
rs1012949457 Chr.17: 79023232-79023232 on Build GRCh38
rs934590610 Chr.17: 79023205-79023205 on Build GRCh38
rs367706747 Chr.17: 79023173-79023173 on Build GRCh38
rs1043402155 Chr.17: 79022907-79022907 on Build GRCh38
rs1056857397 Chr.17: 79023222-79023222 on Build GRCh38
rs533756743 Chr.17: 79023116-79023116 on Build GRCh38
rs184116558 Chr.17: 79023140-79023140 on Build GRCh38
rs564781932 Chr.17: 79022880-79022880 on Build GRCh38
rs58251672 Chr.17: 79023164-79023164 on Build GRCh38
rs920932575 Chr.17: 79023157-79023157 on Build GRCh38
rs1012960921 Chr.17: 79022967-79022967 on Build GRCh38
rs943730486 Chr.17: 79023139-79023139 on Build GRCh38
rs1040624586 Chr.17: 79023246-79023246 on Build GRCh38
rs564669870 Chr.17: 79023191-79023191 on Build GRCh38
rs536620589 Chr.17: 79022889-79022889 on Build GRCh38
rs943782799 Chr.17: 79022868-79022868 on Build GRCh38
rs188926154 Chr.17: 79023196-79023196 on Build GRCh38
rs1049176017 Chr.17: 79023211-79023211 on Build GRCh38
rs571509004 Chr.17: 79023060-79023060 on Build GRCh38
rs998298135 Chr.17: 79023053-79023053 on Build GRCh38
rs557041282 Chr.17: 79022957-79022957 on Build GRCh38
rs770217260 Chr.17: 79023285-79023285 on Build GRCh38
rs182393325 Chr.17: 79022950-79022950 on Build GRCh38
rs1027011960 Chr.17: 79023058-79023058 on Build GRCh38
rs927291029 Chr.17: 79023190-79023190 on Build GRCh38
rs953769976 Chr.17: 79023097-79023097 on Build GRCh38
rs955883294 Chr.17: 79023149-79023149 on Build GRCh38
rs553960554 Chr.17: 79023117-79023117 on Build GRCh38
rs75224210 Chr.17: 79022891-79022891 on Build GRCh38
rs191877011 Chr.17: 79023041-79023041 on Build GRCh38
rs540431306 Chr.17: 79023229-79023229 on Build GRCh38
rs1035507612 Chr.17: 79022964-79022964 on Build GRCh38
rs553555257 Chr.17: 79023258-79023258 on Build GRCh38
rs1028052265 Chr.17: 79023057-79023057 on Build GRCh38
rs987725311 Chr.17: 79023287-79023287 on Build GRCh38
rs950912178 Chr.17: 79023284-79023284 on Build GRCh38
rs73411201 Chr.17: 79023018-79023018 on Build GRCh38
rs187883448 Chr.17: 79022966-79022966 on Build GRCh38
rs905967140 Chr.17: 79022906-79022906 on Build GRCh38
rs753687177 Chr.17: 79022849-79022849 on Build GRCh38
rs570528544 Chr.17: 79023025-79023025 on Build GRCh38
rs1049229144 Chr.17: 79022935-79022935 on Build GRCh38

View Full Product Details