Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79079141-79079680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENGASE
Entrez Gene ID
64772
Gene Name
endo-beta-N-acetylglucosaminidase
Gene Aliases
-
UniGene
Hs.29288
Gene Chromosome Location
Chr.17: 79074812-79088603 on Build GRCh38
Transcripts
NM_001042573.2 XM_006722017.3 XM_006722019.3 XM_017024961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853671 c.514delC p.P173fs*61 Chr.17: 79079586-79079586 on Build GRCh38
985339 c.475G>A p.D159N Chr.17: 79079547-79079547 on Build GRCh38
4915160 c.548A>G p.H183R Chr.17: 79079620-79079620 on Build GRCh38
6217495 c.417-3C>G p.? Chr.17: 79079486-79079486 on Build GRCh38
5493649 c.558C>T p.C186C Chr.17: 79079630-79079630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902489382 Chr.17: 79079250-79079250 on Build GRCh38
rs376231051 Chr.17: 79079499-79079499 on Build GRCh38
rs879683420 Chr.17: 79079518-79079518 on Build GRCh38
rs759269703 Chr.17: 79079495-79079495 on Build GRCh38
rs747589127 Chr.17: 79079455-79079455 on Build GRCh38
rs770960453 Chr.17: 79079618-79079618 on Build GRCh38
rs948226483 Chr.17: 79079273-79079273 on Build GRCh38
rs3744183 Chr.17: 79079585-79079585 on Build GRCh38
rs895425995 Chr.17: 79079313-79079313 on Build GRCh38
rs778084868 Chr.17: 79079610-79079610 on Build GRCh38
rs369871716 Chr.17: 79079475-79079475 on Build GRCh38
rs376818573 Chr.17: 79079458-79079458 on Build GRCh38
rs538827895 Chr.17: 79079212-79079212 on Build GRCh38
rs760631965 Chr.17: 79079580-79079580 on Build GRCh38
rs748924351 Chr.17: 79079446-79079446 on Build GRCh38
rs777323809 Chr.17: 79079438-79079438 on Build GRCh38
rs950420733 Chr.17: 79079541-79079541 on Build GRCh38
rs1038925383 Chr.17: 79079175-79079175 on Build GRCh38
rs752592601 Chr.17: 79079501-79079501 on Build GRCh38
rs1017020869 Chr.17: 79079415-79079415 on Build GRCh38
rs375383634 Chr.17: 79079631-79079631 on Build GRCh38
rs36073943 Chr.17: 79079577-79079577 on Build GRCh38
rs766201900 Chr.17: 79079486-79079486 on Build GRCh38
rs372884772 Chr.17: 79079504-79079504 on Build GRCh38
rs202152183 Chr.17: 79079503-79079503 on Build GRCh38
rs775355581 Chr.17: 79079642-79079642 on Build GRCh38
rs868411253 Chr.17: 79079251-79079251 on Build GRCh38
rs1026032407 Chr.17: 79079456-79079456 on Build GRCh38
rs759359839 Chr.17: 79079575-79079575 on Build GRCh38
rs368323172 Chr.17: 79079657-79079657 on Build GRCh38
rs918125959 Chr.17: 79079166-79079166 on Build GRCh38
rs763212636 Chr.17: 79079484-79079484 on Build GRCh38
rs915038629 Chr.17: 79079612-79079612 on Build GRCh38
rs781671656 Chr.17: 79079595-79079595 on Build GRCh38
rs748638220 Chr.17: 79079547-79079547 on Build GRCh38
rs768648762 Chr.17: 79079656-79079656 on Build GRCh38
rs533819742 Chr.17: 79079350-79079350 on Build GRCh38
rs371083441 Chr.17: 79079601-79079601 on Build GRCh38
rs531430835 Chr.17: 79079632-79079632 on Build GRCh38
rs750991833 Chr.17: 79079651-79079652 on Build GRCh38
rs569626929 Chr.17: 79079483-79079483 on Build GRCh38
rs551033482 Chr.17: 79079200-79079200 on Build GRCh38
rs201649577 Chr.17: 79079576-79079576 on Build GRCh38
rs570990126 Chr.17: 79079244-79079244 on Build GRCh38
rs920204820 Chr.17: 79079296-79079296 on Build GRCh38
rs893888602 Chr.17: 79079183-79079183 on Build GRCh38
rs776811675 Chr.17: 79079557-79079557 on Build GRCh38
rs748177351 Chr.17: 79079473-79079473 on Build GRCh38
rs780618705 Chr.17: 79079527-79079527 on Build GRCh38
rs901640976 Chr.17: 79079490-79079490 on Build GRCh38
rs1042469771 Chr.17: 79079625-79079625 on Build GRCh38
rs3744185 Chr.17: 79079591-79079591 on Build GRCh38
rs562365525 Chr.17: 79079630-79079630 on Build GRCh38
rs777218263 Chr.17: 79079462-79079462 on Build GRCh38
rs3744184 Chr.17: 79079588-79079588 on Build GRCh38
rs368644397 Chr.17: 79079345-79079345 on Build GRCh38
rs113488517 Chr.17: 79079585-79079585 on Build GRCh38
rs1057107520 Chr.17: 79079299-79079299 on Build GRCh38
rs760591687 Chr.17: 79079648-79079648 on Build GRCh38
rs201376470 Chr.17: 79079543-79079543 on Build GRCh38
rs201681138 Chr.17: 79079546-79079546 on Build GRCh38
rs140106265 Chr.17: 79079588-79079589 on Build GRCh38
rs775865887 Chr.17: 79079309-79079309 on Build GRCh38
rs779130522 Chr.17: 79079620-79079620 on Build GRCh38
rs3744182 Chr.17: 79079549-79079549 on Build GRCh38
rs573889051 Chr.17: 79079590-79079590 on Build GRCh38
rs758741817 Chr.17: 79079447-79079447 on Build GRCh38
rs1012368854 Chr.17: 79079394-79079394 on Build GRCh38
rs931611708 Chr.17: 79079624-79079624 on Build GRCh38
rs185872878 Chr.17: 79079421-79079421 on Build GRCh38
rs148385630 Chr.17: 79079587-79079587 on Build GRCh38
rs11547583 Chr.17: 79079629-79079629 on Build GRCh38
rs756932775 Chr.17: 79079522-79079522 on Build GRCh38
rs753476951 Chr.17: 79079521-79079521 on Build GRCh38
rs547070133 Chr.17: 79079374-79079374 on Build GRCh38
rs992280476 Chr.17: 79079165-79079166 on Build GRCh38
rs747386843 Chr.17: 79079531-79079531 on Build GRCh38
rs956839686 Chr.17: 79079165-79079165 on Build GRCh38
rs535730398 Chr.17: 79079425-79079425 on Build GRCh38
rs558315648 Chr.17: 79079564-79079564 on Build GRCh38
rs538416262 Chr.17: 79079550-79079550 on Build GRCh38
rs1042498948 Chr.17: 79079215-79079215 on Build GRCh38
rs1048745908 Chr.17: 79079412-79079412 on Build GRCh38
rs749391575 Chr.17: 79079611-79079611 on Build GRCh38
rs756639142 Chr.17: 79079607-79079607 on Build GRCh38
rs1003560570 Chr.17: 79079291-79079291 on Build GRCh38
rs373761972 Chr.17: 79079494-79079494 on Build GRCh38

View Full Product Details