Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79081815-79082311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENGASE
Entrez Gene ID
64772
Gene Name
endo-beta-N-acetylglucosaminidase
Gene Aliases
-
UniGene
Hs.29288
Gene Chromosome Location
Chr.17: 79074812-79088603 on Build GRCh38
Transcripts
NM_001042573.2 XM_006722017.3 XM_006722018.3 XM_006722019.3 XM_017024961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6082670 c.453G>T p.V151V Chr.17: 79082036-79082036 on Build GRCh38
6148508 c.329C>T p.S110F Chr.17: 79081912-79081912 on Build GRCh38
1386518 c.1007A>G p.N336S Chr.17: 79082032-79082032 on Build GRCh38
3680600 c.468C>T p.F156F Chr.17: 79082051-79082051 on Build GRCh38
7307297 c.1027G>C p.D343H Chr.17: 79082052-79082052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746053305 Chr.17: 79082130-79082130 on Build GRCh38
rs894577904 Chr.17: 79082043-79082043 on Build GRCh38
rs781739164 Chr.17: 79082036-79082036 on Build GRCh38
rs747110192 Chr.17: 79081859-79081859 on Build GRCh38
rs887952942 Chr.17: 79081844-79081844 on Build GRCh38
rs769381499 Chr.17: 79082165-79082165 on Build GRCh38
rs1036976639 Chr.17: 79082148-79082148 on Build GRCh38
rs151057576 Chr.17: 79082244-79082244 on Build GRCh38
rs752263128 Chr.17: 79081974-79081974 on Build GRCh38
rs768661797 Chr.17: 79081929-79081929 on Build GRCh38
rs1049182210 Chr.17: 79081837-79081837 on Build GRCh38
rs568411413 Chr.17: 79082195-79082195 on Build GRCh38
rs548593245 Chr.17: 79082185-79082185 on Build GRCh38
rs751971083 Chr.17: 79082033-79082033 on Build GRCh38
rs753581791 Chr.17: 79082025-79082025 on Build GRCh38
rs868239839 Chr.17: 79082166-79082166 on Build GRCh38
rs768731692 Chr.17: 79082143-79082143 on Build GRCh38
rs545736939 Chr.17: 79082272-79082272 on Build GRCh38
rs570593632 Chr.17: 79082259-79082259 on Build GRCh38
rs557191567 Chr.17: 79081914-79081914 on Build GRCh38
rs753917240 Chr.17: 79082113-79082113 on Build GRCh38
rs755777491 Chr.17: 79081902-79081902 on Build GRCh38
rs776501262 Chr.17: 79082062-79082063 on Build GRCh38
rs772941280 Chr.17: 79082166-79082167 on Build GRCh38
rs769591458 Chr.17: 79082164-79082166 on Build GRCh38
rs563498854 Chr.17: 79082079-79082079 on Build GRCh38
rs754506733 Chr.17: 79081894-79081894 on Build GRCh38
rs779038846 Chr.17: 79082057-79082057 on Build GRCh38
rs201060954 Chr.17: 79081897-79081897 on Build GRCh38
rs760034039 Chr.17: 79081977-79081977 on Build GRCh38
rs1032810670 Chr.17: 79081962-79081962 on Build GRCh38
rs776771656 Chr.17: 79081860-79081860 on Build GRCh38
rs760960499 Chr.17: 79082196-79082196 on Build GRCh38
rs766993020 Chr.17: 79081969-79081969 on Build GRCh38
rs74838690 Chr.17: 79082144-79082144 on Build GRCh38
rs771713028 Chr.17: 79082062-79082062 on Build GRCh38
rs539183056 Chr.17: 79082261-79082261 on Build GRCh38
rs763836526 Chr.17: 79081983-79081983 on Build GRCh38
rs374647992 Chr.17: 79081936-79081936 on Build GRCh38
rs774312624 Chr.17: 79081961-79081961 on Build GRCh38
rs76374707 Chr.17: 79082051-79082051 on Build GRCh38
rs368691882 Chr.17: 79082068-79082068 on Build GRCh38
rs764527447 Chr.17: 79082112-79082112 on Build GRCh38
rs367929579 Chr.17: 79081916-79081916 on Build GRCh38
rs375906857 Chr.17: 79081856-79081856 on Build GRCh38
rs772980246 Chr.17: 79081880-79081880 on Build GRCh38
rs754292056 Chr.17: 79082220-79082220 on Build GRCh38
rs1029293135 Chr.17: 79082029-79082029 on Build GRCh38
rs766310358 Chr.17: 79081891-79081891 on Build GRCh38
rs574786825 Chr.17: 79082034-79082034 on Build GRCh38
rs774118597 Chr.17: 79082004-79082004 on Build GRCh38
rs540871052 Chr.17: 79081992-79081992 on Build GRCh38
rs767751473 Chr.17: 79082108-79082108 on Build GRCh38
rs371426986 Chr.17: 79081919-79081919 on Build GRCh38
rs12937557 Chr.17: 79081987-79081987 on Build GRCh38
rs546248936 Chr.17: 79082132-79082132 on Build GRCh38
rs369637200 Chr.17: 79082014-79082014 on Build GRCh38
rs753488111 Chr.17: 79081986-79081986 on Build GRCh38
rs114380445 Chr.17: 79081867-79081867 on Build GRCh38
rs764989886 Chr.17: 79082090-79082090 on Build GRCh38
rs373702682 Chr.17: 79081955-79081955 on Build GRCh38
rs748379604 Chr.17: 79081938-79081938 on Build GRCh38
rs771268814 Chr.17: 79081895-79081895 on Build GRCh38
rs770778793 Chr.17: 79082003-79082003 on Build GRCh38
rs776270994 Chr.17: 79082084-79082084 on Build GRCh38
rs769723401 Chr.17: 79082000-79082000 on Build GRCh38
rs756599393 Chr.17: 79082111-79082111 on Build GRCh38
rs768769329 Chr.17: 79081900-79081902 on Build GRCh38
rs377455066 Chr.17: 79082109-79082109 on Build GRCh38
rs761556855 Chr.17: 79082026-79082026 on Build GRCh38
rs539067115 Chr.17: 79082162-79082162 on Build GRCh38
rs757374905 Chr.17: 79082224-79082224 on Build GRCh38
rs750721256 Chr.17: 79082228-79082228 on Build GRCh38
rs751407622 Chr.17: 79082253-79082253 on Build GRCh38
rs541914209 Chr.17: 79081840-79081840 on Build GRCh38
rs749489623 Chr.17: 79082002-79082002 on Build GRCh38
rs764615478 Chr.17: 79082205-79082205 on Build GRCh38
rs776070069 Chr.17: 79081849-79081849 on Build GRCh38
rs372228025 Chr.17: 79082009-79082009 on Build GRCh38
rs539480225 Chr.17: 79081963-79081963 on Build GRCh38
rs762768889 Chr.17: 79081945-79081945 on Build GRCh38
rs200250575 Chr.17: 79082010-79082010 on Build GRCh38
rs745704710 Chr.17: 79082061-79082061 on Build GRCh38
rs748051438 Chr.17: 79082064-79082099 on Build GRCh38
rs913327896 Chr.17: 79081975-79081975 on Build GRCh38
rs779214847 Chr.17: 79082124-79082124 on Build GRCh38
rs781332098 Chr.17: 79081935-79081935 on Build GRCh38
rs977282099 Chr.17: 79082236-79082236 on Build GRCh38
rs772701622 Chr.17: 79082093-79082093 on Build GRCh38
rs770664186 Chr.17: 79081951-79081951 on Build GRCh38
rs762680748 Chr.17: 79081889-79081889 on Build GRCh38
rs768315931 Chr.17: 79081944-79081944 on Build GRCh38
rs770070562 Chr.17: 79081940-79081940 on Build GRCh38
rs371024567 Chr.17: 79081892-79081892 on Build GRCh38
rs532512544 Chr.17: 79082118-79082118 on Build GRCh38
rs748266062 Chr.17: 79081999-79081999 on Build GRCh38
rs1317421 Chr.17: 79082027-79082027 on Build GRCh38
rs377404114 Chr.17: 79082063-79082063 on Build GRCh38
rs780724905 Chr.17: 79081920-79081920 on Build GRCh38
rs571627330 Chr.17: 79081893-79081893 on Build GRCh38
rs1019347747 Chr.17: 79082156-79082156 on Build GRCh38
rs369431239 Chr.17: 79082047-79082047 on Build GRCh38
rs765195705 Chr.17: 79082225-79082225 on Build GRCh38
rs753098384 Chr.17: 79082039-79082039 on Build GRCh38
rs550805462 Chr.17: 79082249-79082249 on Build GRCh38
rs572772099 Chr.17: 79081990-79081990 on Build GRCh38
rs746236035 Chr.17: 79081917-79081917 on Build GRCh38
rs762482086 Chr.17: 79082183-79082183 on Build GRCh38
rs780395616 Chr.17: 79081857-79081858 on Build GRCh38
rs757483206 Chr.17: 79082052-79082052 on Build GRCh38
rs201591624 Chr.17: 79082046-79082046 on Build GRCh38
rs889428801 Chr.17: 79082258-79082258 on Build GRCh38
rs140962076 Chr.17: 79082274-79082274 on Build GRCh38
rs759194033 Chr.17: 79082005-79082005 on Build GRCh38
rs765011610 Chr.17: 79081869-79081869 on Build GRCh38
rs780538822 Chr.17: 79081989-79081989 on Build GRCh38
rs200007139 Chr.17: 79082088-79082088 on Build GRCh38
rs541406332 Chr.17: 79081991-79081991 on Build GRCh38
rs759969519 Chr.17: 79082094-79082094 on Build GRCh38
rs568346088 Chr.17: 79081859-79081859 on Build GRCh38
rs1006782695 Chr.17: 79082040-79082040 on Build GRCh38
rs772969091 Chr.17: 79082172-79082172 on Build GRCh38
rs779705442 Chr.17: 79082138-79082138 on Build GRCh38
rs188378332 Chr.17: 79081928-79081928 on Build GRCh38

View Full Product Details