Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79082942-79083436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENGASE
Entrez Gene ID
64772
Gene Name
endo-beta-N-acetylglucosaminidase
Gene Aliases
-
UniGene
Hs.29288
Gene Chromosome Location
Chr.17: 79074812-79088603 on Build GRCh38
Transcripts
NM_001042573.2 XM_006722017.3 XM_006722018.3 XM_006722019.3 XM_017024961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6556722 c.1041G>T p.S347S Chr.17: 79083022-79083022 on Build GRCh38
2746986 c.1041G>A p.S347S Chr.17: 79083022-79083022 on Build GRCh38
437693 c.1040C>T p.S347L Chr.17: 79083021-79083021 on Build GRCh38
5387534 c.1071C>T p.S357S Chr.17: 79083052-79083052 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779609931 Chr.17: 79083125-79083125 on Build GRCh38
rs149186913 Chr.17: 79083035-79083035 on Build GRCh38
rs915810203 Chr.17: 79083227-79083227 on Build GRCh38
rs536062149 Chr.17: 79083331-79083331 on Build GRCh38
rs749364805 Chr.17: 79083255-79083255 on Build GRCh38
rs992818029 Chr.17: 79083278-79083278 on Build GRCh38
rs774560857 Chr.17: 79082990-79082990 on Build GRCh38
rs116227347 Chr.17: 79083348-79083348 on Build GRCh38
rs901868945 Chr.17: 79083345-79083345 on Build GRCh38
rs376481257 Chr.17: 79083119-79083119 on Build GRCh38
rs780165565 Chr.17: 79083146-79083146 on Build GRCh38
rs1048890562 Chr.17: 79083309-79083309 on Build GRCh38
rs749773037 Chr.17: 79082976-79082976 on Build GRCh38
rs994654039 Chr.17: 79083143-79083143 on Build GRCh38
rs766058419 Chr.17: 79083006-79083009 on Build GRCh38
rs757200943 Chr.17: 79083031-79083031 on Build GRCh38
rs760434544 Chr.17: 79083011-79083011 on Build GRCh38
rs768727793 Chr.17: 79083338-79083338 on Build GRCh38
rs562247127 Chr.17: 79083124-79083124 on Build GRCh38
rs199569776 Chr.17: 79082994-79082994 on Build GRCh38
rs916956724 Chr.17: 79083289-79083289 on Build GRCh38
rs1038995607 Chr.17: 79083305-79083305 on Build GRCh38
rs765097018 Chr.17: 79083034-79083034 on Build GRCh38
rs540137073 Chr.17: 79083053-79083053 on Build GRCh38
rs774031539 Chr.17: 79083015-79083016 on Build GRCh38
rs746261640 Chr.17: 79083186-79083186 on Build GRCh38
rs200703027 Chr.17: 79083036-79083036 on Build GRCh38
rs567133724 Chr.17: 79083306-79083306 on Build GRCh38
rs754645846 Chr.17: 79083112-79083112 on Build GRCh38
rs762074112 Chr.17: 79083190-79083190 on Build GRCh38
rs143092634 Chr.17: 79083106-79083106 on Build GRCh38
rs376610611 Chr.17: 79083005-79083005 on Build GRCh38
rs780311359 Chr.17: 79083252-79083252 on Build GRCh38
rs528636638 Chr.17: 79083364-79083364 on Build GRCh38
rs571140326 Chr.17: 79083221-79083221 on Build GRCh38
rs370035126 Chr.17: 79083052-79083052 on Build GRCh38
rs760943455 Chr.17: 79083079-79083079 on Build GRCh38
rs775806290 Chr.17: 79083000-79083000 on Build GRCh38
rs746346141 Chr.17: 79083126-79083126 on Build GRCh38
rs776495838 Chr.17: 79083089-79083089 on Build GRCh38
rs761445623 Chr.17: 79083091-79083091 on Build GRCh38
rs750313342 Chr.17: 79083098-79083098 on Build GRCh38
rs370885670 Chr.17: 79082980-79082980 on Build GRCh38
rs756476314 Chr.17: 79083049-79083049 on Build GRCh38
rs763914630 Chr.17: 79083018-79083018 on Build GRCh38
rs188871787 Chr.17: 79083097-79083097 on Build GRCh38
rs746655293 Chr.17: 79083046-79083046 on Build GRCh38
rs966435925 Chr.17: 79083215-79083215 on Build GRCh38
rs371968786 Chr.17: 79083006-79083006 on Build GRCh38
rs1051838142 Chr.17: 79083281-79083281 on Build GRCh38
rs370432979 Chr.17: 79083022-79083022 on Build GRCh38
rs1021089353 Chr.17: 79083138-79083138 on Build GRCh38
rs941848248 Chr.17: 79083021-79083021 on Build GRCh38
rs924203978 Chr.17: 79083251-79083252 on Build GRCh38
rs902670757 Chr.17: 79083344-79083344 on Build GRCh38
rs373590967 Chr.17: 79082999-79082999 on Build GRCh38
rs201551149 Chr.17: 79083172-79083172 on Build GRCh38
rs540152956 Chr.17: 79083228-79083228 on Build GRCh38
rs181617117 Chr.17: 79083244-79083244 on Build GRCh38
rs772341733 Chr.17: 79083128-79083128 on Build GRCh38
rs946961894 Chr.17: 79083332-79083332 on Build GRCh38
rs148218660 Chr.17: 79083198-79083198 on Build GRCh38
rs771579537 Chr.17: 79083067-79083067 on Build GRCh38
rs919074773 Chr.17: 79083245-79083245 on Build GRCh38
rs368162113 Chr.17: 79083149-79083149 on Build GRCh38
rs943349037 Chr.17: 79083302-79083302 on Build GRCh38
rs967115873 Chr.17: 79082975-79082975 on Build GRCh38
rs751237626 Chr.17: 79083110-79083110 on Build GRCh38
rs905586615 Chr.17: 79083304-79083304 on Build GRCh38
rs745965568 Chr.17: 79083070-79083070 on Build GRCh38
rs201695253 Chr.17: 79083081-79083081 on Build GRCh38
rs779707926 Chr.17: 79083041-79083041 on Build GRCh38
rs756336564 Chr.17: 79083222-79083222 on Build GRCh38
rs748135381 Chr.17: 79083388-79083388 on Build GRCh38
rs747001584 Chr.17: 79083150-79083150 on Build GRCh38
rs912016596 Chr.17: 79083301-79083301 on Build GRCh38
rs1000991974 Chr.17: 79083403-79083403 on Build GRCh38
rs368796338 Chr.17: 79083077-79083077 on Build GRCh38
rs774957644 Chr.17: 79083068-79083068 on Build GRCh38
rs374695310 Chr.17: 79083071-79083071 on Build GRCh38

View Full Product Details