Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79741711-79742204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP7
Entrez Gene ID
339221
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Gene Aliases
ALK-SMase, E-NPP 7, NPP-7, NPP7
UniGene
Hs.114084
Gene Chromosome Location
Chr.17: 79730919-79742926 on Build GRCh38
Transcripts
NM_178543.4 XM_011524737.1

SNP Detail

SNP ID Chromosome Location
rs1055561816 Chr.17: 79742128-79742128 on Build GRCh38
rs1006448929 Chr.17: 79741952-79741952 on Build GRCh38
rs535129115 Chr.17: 79742143-79742143 on Build GRCh38
rs112143890 Chr.17: 79742156-79742156 on Build GRCh38
rs1017991846 Chr.17: 79742013-79742013 on Build GRCh38
rs555753239 Chr.17: 79742072-79742072 on Build GRCh38
rs945903105 Chr.17: 79741837-79741837 on Build GRCh38
rs894154622 Chr.17: 79741909-79741909 on Build GRCh38
rs914515412 Chr.17: 79742061-79742061 on Build GRCh38
rs1037944216 Chr.17: 79742141-79742141 on Build GRCh38
rs967172554 Chr.17: 79742065-79742065 on Build GRCh38
rs958046942 Chr.17: 79742039-79742039 on Build GRCh38
rs547135173 Chr.17: 79741841-79741841 on Build GRCh38
rs911136621 Chr.17: 79741758-79741758 on Build GRCh38
rs533962179 Chr.17: 79742161-79742162 on Build GRCh38
rs375874280 Chr.17: 79741858-79741858 on Build GRCh38
rs928401924 Chr.17: 79742079-79742079 on Build GRCh38
rs553146039 Chr.17: 79742168-79742168 on Build GRCh38
rs1030407099 Chr.17: 79741972-79741972 on Build GRCh38
rs374383034 Chr.17: 79742108-79742108 on Build GRCh38
rs890635478 Chr.17: 79742170-79742170 on Build GRCh38
rs565869422 Chr.17: 79741850-79741850 on Build GRCh38
rs3194491 Chr.17: 79741979-79741979 on Build GRCh38
rs78180981 Chr.17: 79741772-79741772 on Build GRCh38
rs113917231 Chr.17: 79742110-79742110 on Build GRCh38
rs915555728 Chr.17: 79742133-79742133 on Build GRCh38
rs990004406 Chr.17: 79742054-79742054 on Build GRCh38
rs914380127 Chr.17: 79741820-79741820 on Build GRCh38
rs567519564 Chr.17: 79742093-79742093 on Build GRCh38
rs551149709 Chr.17: 79741959-79741959 on Build GRCh38
rs66677409 Chr.17: 79741735-79741735 on Build GRCh38
rs993677239 Chr.17: 79742162-79742162 on Build GRCh38
rs941700262 Chr.17: 79742136-79742136 on Build GRCh38
rs533003093 Chr.17: 79741885-79741885 on Build GRCh38
rs569648604 Chr.17: 79741965-79741965 on Build GRCh38
rs977693271 Chr.17: 79742066-79742066 on Build GRCh38
rs145036727 Chr.17: 79741793-79741793 on Build GRCh38
rs994099816 Chr.17: 79741969-79741969 on Build GRCh38
rs1007642853 Chr.17: 79741981-79741981 on Build GRCh38
rs371908908 Chr.17: 79742028-79742028 on Build GRCh38
rs938354022 Chr.17: 79741889-79741889 on Build GRCh38
rs782795363 Chr.17: 79742122-79742122 on Build GRCh38
rs1055907935 Chr.17: 79741893-79741893 on Build GRCh38
rs34313979 Chr.17: 79742038-79742038 on Build GRCh38
rs36081630 Chr.17: 79741865-79741866 on Build GRCh38
rs897976292 Chr.17: 79741968-79741968 on Build GRCh38
rs574390017 Chr.17: 79742105-79742105 on Build GRCh38

View Full Product Details