Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80036548-80037053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC40
Entrez Gene ID
55036
Gene Name
coiled-coil domain containing 40
Gene Aliases
CILD15, FAP172
UniGene
Hs.202542
Gene Chromosome Location
Chr.17: 80036632-80100613 on Build GRCh38
Transcripts
NM_001243342.1 NM_017950.3 XM_005257492.4 XM_011524963.2 XM_011524965.2 XM_017024807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364320 c.28C>T p.R10W Chr.17: 80036690-80036690 on Build GRCh38
5364318 c.28C>T p.R10W Chr.17: 80036690-80036690 on Build GRCh38
4428182 c.10C>T p.P4S Chr.17: 80036672-80036672 on Build GRCh38
4428184 c.10C>T p.P4S Chr.17: 80036672-80036672 on Build GRCh38
4428183 c.10C>T p.P4S Chr.17: 80036672-80036672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894855944 Chr.17: 80037015-80037015 on Build GRCh38
rs1008672106 Chr.17: 80036711-80036711 on Build GRCh38
rs1019182046 Chr.17: 80036726-80036726 on Build GRCh38
rs1040663604 Chr.17: 80036874-80036874 on Build GRCh38
rs556946520 Chr.17: 80036960-80036960 on Build GRCh38
rs985612411 Chr.17: 80036943-80036943 on Build GRCh38
rs751298635 Chr.17: 80036685-80036685 on Build GRCh38
rs577056774 Chr.17: 80036703-80036703 on Build GRCh38
rs754850477 Chr.17: 80036686-80036686 on Build GRCh38
rs934195206 Chr.17: 80036696-80036696 on Build GRCh38
rs922758159 Chr.17: 80036691-80036691 on Build GRCh38
rs947349275 Chr.17: 80036581-80036581 on Build GRCh38
rs955520403 Chr.17: 80036936-80036936 on Build GRCh38
rs930922047 Chr.17: 80036904-80036904 on Build GRCh38
rs1053692064 Chr.17: 80036595-80036595 on Build GRCh38
rs993241819 Chr.17: 80037031-80037031 on Build GRCh38
rs752426610 Chr.17: 80036693-80036693 on Build GRCh38
rs747590301 Chr.17: 80036625-80036625 on Build GRCh38
rs920840793 Chr.17: 80036880-80036880 on Build GRCh38
rs750300695 Chr.17: 80036682-80036682 on Build GRCh38
rs769092437 Chr.17: 80036702-80036702 on Build GRCh38
rs900256515 Chr.17: 80036963-80036963 on Build GRCh38
rs990128534 Chr.17: 80036968-80036968 on Build GRCh38
rs1033464259 Chr.17: 80036621-80036621 on Build GRCh38
rs961110015 Chr.17: 80036877-80036877 on Build GRCh38
rs778919758 Chr.17: 80037007-80037007 on Build GRCh38
rs377599647 Chr.17: 80036690-80036690 on Build GRCh38
rs999425121 Chr.17: 80037001-80037001 on Build GRCh38
rs1054971493 Chr.17: 80037014-80037015 on Build GRCh38
rs1000180694 Chr.17: 80036583-80036583 on Build GRCh38
rs3752042 Chr.17: 80036614-80036614 on Build GRCh38
rs766369570 Chr.17: 80036683-80036683 on Build GRCh38
rs768751527 Chr.17: 80036659-80036659 on Build GRCh38
rs3752041 Chr.17: 80036749-80036749 on Build GRCh38
rs1005182080 Chr.17: 80036869-80036869 on Build GRCh38
rs893734225 Chr.17: 80036599-80036599 on Build GRCh38
rs541624280 Chr.17: 80036774-80036774 on Build GRCh38
rs1009374307 Chr.17: 80036600-80036600 on Build GRCh38
rs772706089 Chr.17: 80036699-80036699 on Build GRCh38
rs746401466 Chr.17: 80036637-80036637 on Build GRCh38
rs776947186 Chr.17: 80036673-80036673 on Build GRCh38
rs996015616 Chr.17: 80036964-80036964 on Build GRCh38
rs545826383 Chr.17: 80036896-80036896 on Build GRCh38
rs906358406 Chr.17: 80036804-80036804 on Build GRCh38
rs1003293180 Chr.17: 80036620-80036620 on Build GRCh38
rs765319797 Chr.17: 80036681-80036681 on Build GRCh38
rs1003407666 Chr.17: 80036808-80036808 on Build GRCh38
rs886187552 Chr.17: 80036705-80036705 on Build GRCh38
rs970428657 Chr.17: 80036616-80036616 on Build GRCh38
rs781242178 Chr.17: 80037008-80037008 on Build GRCh38
rs917341011 Chr.17: 80036852-80036852 on Build GRCh38
rs1022123068 Chr.17: 80036979-80036979 on Build GRCh38
rs187049659 Chr.17: 80036912-80036912 on Build GRCh38
rs747300417 Chr.17: 80036701-80036701 on Build GRCh38
rs772509954 Chr.17: 80036642-80036642 on Build GRCh38
rs924636801 Chr.17: 80036787-80036787 on Build GRCh38
rs771732758 Chr.17: 80036684-80036685 on Build GRCh38
rs948877269 Chr.17: 80036868-80036868 on Build GRCh38
rs533801756 Chr.17: 80036680-80036680 on Build GRCh38
rs939136912 Chr.17: 80037012-80037012 on Build GRCh38
rs544319033 Chr.17: 80036736-80036736 on Build GRCh38
rs945290088 Chr.17: 80036761-80036761 on Build GRCh38
rs868696844 Chr.17: 80036813-80036813 on Build GRCh38
rs967971099 Chr.17: 80036766-80036766 on Build GRCh38
rs755531523 Chr.17: 80036697-80036697 on Build GRCh38
rs746596590 Chr.17: 80036622-80036622 on Build GRCh38
rs574583489 Chr.17: 80036763-80036763 on Build GRCh38
rs776665408 Chr.17: 80036708-80036708 on Build GRCh38
rs1042685548 Chr.17: 80037003-80037003 on Build GRCh38
rs527569429 Chr.17: 80036882-80036882 on Build GRCh38
rs3752040 Chr.17: 80036911-80036911 on Build GRCh38
rs551907438 Chr.17: 80036645-80036645 on Build GRCh38
rs967917466 Chr.17: 80036998-80036998 on Build GRCh38
rs1017011849 Chr.17: 80036874-80036875 on Build GRCh38
rs73437681 Chr.17: 80036627-80036627 on Build GRCh38
rs1044968221 Chr.17: 80036876-80036876 on Build GRCh38
rs1005709657 Chr.17: 80037021-80037021 on Build GRCh38
rs776004675 Chr.17: 80036644-80036644 on Build GRCh38
rs896013335 Chr.17: 80036640-80036640 on Build GRCh38
rs549872389 Chr.17: 80036962-80036962 on Build GRCh38
rs913223135 Chr.17: 80036986-80036986 on Build GRCh38
rs181824359 Chr.17: 80036809-80036809 on Build GRCh38
rs762106770 Chr.17: 80036720-80036720 on Build GRCh38
rs780956509 Chr.17: 80036623-80036623 on Build GRCh38
rs558574170 Chr.17: 80036698-80036698 on Build GRCh38

View Full Product Details