Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80138892-80139400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A3
Entrez Gene ID
9775
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
DDX48, MUK34, NMP265, NUK34, RCPS, eIF4AIII
UniGene
Hs.389649
Gene Chromosome Location
Chr.17: 80135214-80147183 on Build GRCh38
Transcripts
NM_014740.3 XM_011525522.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437725 c.610G>T p.V204L Chr.17: 80139139-80139139 on Build GRCh38
5640431 c.651C>T p.I217I Chr.17: 80139098-80139098 on Build GRCh38
4170849 c.709A>G p.I237V Chr.17: 80139040-80139040 on Build GRCh38
6304283 c.659C>T p.T220M Chr.17: 80139090-80139090 on Build GRCh38
3937640 c.699G>A p.M233I Chr.17: 80139050-80139050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764301358 Chr.17: 80139100-80139100 on Build GRCh38
rs896024172 Chr.17: 80139254-80139254 on Build GRCh38
rs535401553 Chr.17: 80139080-80139080 on Build GRCh38
rs145491523 Chr.17: 80139316-80139316 on Build GRCh38
rs931787200 Chr.17: 80139366-80139366 on Build GRCh38
rs183195514 Chr.17: 80139317-80139317 on Build GRCh38
rs547093183 Chr.17: 80139250-80139250 on Build GRCh38
rs778792702 Chr.17: 80139009-80139009 on Build GRCh38
rs373315661 Chr.17: 80139116-80139116 on Build GRCh38
rs537273424 Chr.17: 80139013-80139013 on Build GRCh38
rs903277434 Chr.17: 80139221-80139221 on Build GRCh38
rs562180248 Chr.17: 80139358-80139358 on Build GRCh38
rs771388338 Chr.17: 80139044-80139044 on Build GRCh38
rs79962374 Chr.17: 80139319-80139319 on Build GRCh38
rs773665474 Chr.17: 80139038-80139038 on Build GRCh38
rs753819652 Chr.17: 80138981-80138981 on Build GRCh38
rs923251206 Chr.17: 80139353-80139353 on Build GRCh38
rs1018647461 Chr.17: 80139318-80139318 on Build GRCh38
rs376241915 Chr.17: 80139179-80139179 on Build GRCh38
rs915127631 Chr.17: 80139195-80139195 on Build GRCh38
rs915161033 Chr.17: 80139357-80139357 on Build GRCh38
rs59432707 Chr.17: 80139375-80139375 on Build GRCh38
rs925121184 Chr.17: 80139372-80139372 on Build GRCh38
rs903228897 Chr.17: 80138918-80138918 on Build GRCh38
rs965353057 Chr.17: 80139337-80139337 on Build GRCh38
rs1002416085 Chr.17: 80139268-80139268 on Build GRCh38
rs767842888 Chr.17: 80139088-80139088 on Build GRCh38
rs369529398 Chr.17: 80139175-80139175 on Build GRCh38
rs375014709 Chr.17: 80139143-80139143 on Build GRCh38
rs779611208 Chr.17: 80139173-80139173 on Build GRCh38
rs113448837 Chr.17: 80139272-80139272 on Build GRCh38
rs895971837 Chr.17: 80138945-80138945 on Build GRCh38
rs200424645 Chr.17: 80139037-80139037 on Build GRCh38
rs552140196 Chr.17: 80138958-80138958 on Build GRCh38
rs369642771 Chr.17: 80138935-80138935 on Build GRCh38
rs113268177 Chr.17: 80139024-80139024 on Build GRCh38
rs778713625 Chr.17: 80139203-80139223 on Build GRCh38
rs369049709 Chr.17: 80139089-80139089 on Build GRCh38
rs899377672 Chr.17: 80139269-80139269 on Build GRCh38
rs1052071284 Chr.17: 80139128-80139128 on Build GRCh38
rs931866843 Chr.17: 80139011-80139011 on Build GRCh38
rs1025951295 Chr.17: 80139273-80139273 on Build GRCh38
rs141667694 Chr.17: 80139068-80139068 on Build GRCh38
rs570258154 Chr.17: 80138986-80138986 on Build GRCh38
rs11559244 Chr.17: 80139036-80139036 on Build GRCh38
rs746463899 Chr.17: 80139018-80139018 on Build GRCh38
rs752824112 Chr.17: 80138978-80138978 on Build GRCh38
rs934759225 Chr.17: 80139227-80139227 on Build GRCh38
rs761661436 Chr.17: 80139261-80139261 on Build GRCh38
rs981158695 Chr.17: 80139258-80139258 on Build GRCh38
rs139991637 Chr.17: 80139199-80139199 on Build GRCh38
rs191691560 Chr.17: 80138957-80138957 on Build GRCh38
rs753672167 Chr.17: 80139107-80139107 on Build GRCh38
rs779738577 Chr.17: 80139017-80139017 on Build GRCh38
rs373215829 Chr.17: 80139010-80139010 on Build GRCh38
rs376084901 Chr.17: 80139193-80139193 on Build GRCh38
rs974507686 Chr.17: 80139340-80139340 on Build GRCh38
rs146210677 Chr.17: 80139140-80139140 on Build GRCh38
rs147471756 Chr.17: 80138929-80138929 on Build GRCh38
rs771468313 Chr.17: 80139201-80139201 on Build GRCh38
rs1023647850 Chr.17: 80139265-80139265 on Build GRCh38
rs976394872 Chr.17: 80139347-80139347 on Build GRCh38
rs543288496 Chr.17: 80139341-80139341 on Build GRCh38
rs574463592 Chr.17: 80139047-80139047 on Build GRCh38
rs372300628 Chr.17: 80138980-80138980 on Build GRCh38
rs1043039034 Chr.17: 80139215-80139215 on Build GRCh38
rs757104118 Chr.17: 80139007-80139011 on Build GRCh38
rs760771097 Chr.17: 80139095-80139095 on Build GRCh38
rs750039941 Chr.17: 80139202-80139203 on Build GRCh38
rs923060108 Chr.17: 80138959-80138959 on Build GRCh38
rs201917263 Chr.17: 80139040-80139040 on Build GRCh38
rs529393794 Chr.17: 80139368-80139368 on Build GRCh38
rs756324175 Chr.17: 80138979-80138979 on Build GRCh38
rs371370527 Chr.17: 80139202-80139202 on Build GRCh38
rs911756594 Chr.17: 80139094-80139094 on Build GRCh38
rs1013196586 Chr.17: 80138947-80138947 on Build GRCh38

View Full Product Details