Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81086357-81086854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2
Entrez Gene ID
10458
Gene Name
BAI1 associated protein 2
Gene Aliases
BAP2, FLAF3, IRSP53
UniGene
Hs.128316
Gene Chromosome Location
Chr.17: 81035147-81117432 on Build GRCh38
Transcripts
NM_001144888.1 NM_006340.2 NM_017450.2 NM_017451.2 XM_005256943.1 XM_005256944.1 XM_005256945.1 XM_005256948.3 XM_006721635.1 XM_006721636.1 XM_006721637.2 XM_011524193.1 XM_011524194.1 XM_017024017.1 XM_017024018.1 XM_017024019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937651 c.394G>A p.D132N Chr.17: 81086485-81086485 on Build GRCh38
2804345 c.397G>A p.A133T Chr.17: 81086488-81086488 on Build GRCh38
6267386 c.449G>T p.G150V Chr.17: 81086540-81086540 on Build GRCh38
3773340 c.404A>G p.D135G Chr.17: 81086495-81086495 on Build GRCh38
6021697 c.412C>G p.Q138E Chr.17: 81086503-81086503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757268353 Chr.17: 81086593-81086593 on Build GRCh38
rs1029629177 Chr.17: 81086829-81086829 on Build GRCh38
rs777916497 Chr.17: 81086488-81086488 on Build GRCh38
rs561571272 Chr.17: 81086480-81086480 on Build GRCh38
rs1019112293 Chr.17: 81086634-81086634 on Build GRCh38
rs530620776 Chr.17: 81086640-81086640 on Build GRCh38
rs774407310 Chr.17: 81086413-81086413 on Build GRCh38
rs538221314 Chr.17: 81086741-81086741 on Build GRCh38
rs979471510 Chr.17: 81086651-81086651 on Build GRCh38
rs1036619432 Chr.17: 81086791-81086791 on Build GRCh38
rs775403797 Chr.17: 81086427-81086427 on Build GRCh38
rs577550065 Chr.17: 81086389-81086389 on Build GRCh38
rs771110857 Chr.17: 81086508-81086508 on Build GRCh38
rs886761949 Chr.17: 81086592-81086592 on Build GRCh38
rs779072690 Chr.17: 81086607-81086607 on Build GRCh38
rs117737110 Chr.17: 81086639-81086639 on Build GRCh38
rs774265446 Chr.17: 81086512-81086512 on Build GRCh38
rs939632704 Chr.17: 81086785-81086785 on Build GRCh38
rs749303669 Chr.17: 81086406-81086406 on Build GRCh38
rs776492137 Chr.17: 81086565-81086565 on Build GRCh38
rs772086481 Chr.17: 81086526-81086526 on Build GRCh38
rs749461725 Chr.17: 81086505-81086505 on Build GRCh38
rs949215483 Chr.17: 81086411-81086411 on Build GRCh38
rs773315796 Chr.17: 81086396-81086396 on Build GRCh38
rs917794978 Chr.17: 81086705-81086705 on Build GRCh38
rs756848734 Chr.17: 81086448-81086448 on Build GRCh38
rs759389844 Chr.17: 81086418-81086418 on Build GRCh38
rs141506955 Chr.17: 81086647-81086647 on Build GRCh38
rs11539642 Chr.17: 81086487-81086487 on Build GRCh38
rs754131303 Chr.17: 81086597-81086597 on Build GRCh38
rs981052585 Chr.17: 81086722-81086722 on Build GRCh38
rs1029457611 Chr.17: 81086384-81086384 on Build GRCh38
rs182333422 Chr.17: 81086434-81086434 on Build GRCh38
rs988034255 Chr.17: 81086695-81086695 on Build GRCh38
rs992231638 Chr.17: 81086723-81086723 on Build GRCh38
rs1040853504 Chr.17: 81086742-81086742 on Build GRCh38
rs956684044 Chr.17: 81086687-81086687 on Build GRCh38
rs551077297 Chr.17: 81086593-81086593 on Build GRCh38
rs532382372 Chr.17: 81086588-81086588 on Build GRCh38
rs190091807 Chr.17: 81086388-81086388 on Build GRCh38
rs374586268 Chr.17: 81086619-81086619 on Build GRCh38
rs201271163 Chr.17: 81086395-81086395 on Build GRCh38
rs371384882 Chr.17: 81086449-81086449 on Build GRCh38
rs765595067 Chr.17: 81086616-81086616 on Build GRCh38
rs917747643 Chr.17: 81086726-81086726 on Build GRCh38
rs908219715 Chr.17: 81086769-81086769 on Build GRCh38
rs567112212 Chr.17: 81086650-81086650 on Build GRCh38
rs953405967 Chr.17: 81086385-81086385 on Build GRCh38
rs770789583 Chr.17: 81086408-81086408 on Build GRCh38
rs879295310 Chr.17: 81086709-81086709 on Build GRCh38
rs947893278 Chr.17: 81086727-81086727 on Build GRCh38
rs750628284 Chr.17: 81086611-81086612 on Build GRCh38
rs745868193 Chr.17: 81086525-81086525 on Build GRCh38
rs748476838 Chr.17: 81086485-81086485 on Build GRCh38
rs778109095 Chr.17: 81086594-81086594 on Build GRCh38
rs146668397 Chr.17: 81086447-81086447 on Build GRCh38
rs765573261 Chr.17: 81086416-81086417 on Build GRCh38
rs761337520 Chr.17: 81086576-81086576 on Build GRCh38
rs767387128 Chr.17: 81086425-81086425 on Build GRCh38
rs534635236 Chr.17: 81086668-81086668 on Build GRCh38
rs571242350 Chr.17: 81086740-81086740 on Build GRCh38
rs1015154545 Chr.17: 81086601-81086601 on Build GRCh38
rs767999334 Chr.17: 81086534-81086534 on Build GRCh38
rs367841125 Chr.17: 81086536-81086536 on Build GRCh38
rs186604902 Chr.17: 81086577-81086577 on Build GRCh38
rs968417718 Chr.17: 81086703-81086703 on Build GRCh38
rs545115518 Chr.17: 81086390-81086390 on Build GRCh38
rs200297110 Chr.17: 81086549-81086549 on Build GRCh38
rs919868732 Chr.17: 81086732-81086732 on Build GRCh38
rs41292398 Chr.17: 81086674-81086674 on Build GRCh38
rs926470807 Chr.17: 81086731-81086731 on Build GRCh38
rs930952322 Chr.17: 81086734-81086734 on Build GRCh38
rs1008006044 Chr.17: 81086633-81086633 on Build GRCh38
rs769562078 Chr.17: 81086764-81086764 on Build GRCh38
rs371413726 Chr.17: 81086589-81086589 on Build GRCh38
rs768555901 Chr.17: 81086562-81086562 on Build GRCh38
rs748415517 Chr.17: 81086394-81086394 on Build GRCh38
rs7210438 Chr.17: 81086380-81086380 on Build GRCh38
rs757400683 Chr.17: 81086598-81086598 on Build GRCh38
rs780197095 Chr.17: 81086630-81086630 on Build GRCh38
rs752146377 Chr.17: 81086464-81086464 on Build GRCh38
rs991970933 Chr.17: 81086543-81086543 on Build GRCh38
rs35895585 Chr.17: 81086553-81086553 on Build GRCh38
rs760425980 Chr.17: 81086428-81086428 on Build GRCh38
rs140045781 Chr.17: 81086484-81086484 on Build GRCh38

View Full Product Details