Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81103362-81103849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2
Entrez Gene ID
10458
Gene Name
BAI1 associated protein 2
Gene Aliases
BAP2, FLAF3, IRSP53
UniGene
Hs.128316
Gene Chromosome Location
Chr.17: 81035147-81117432 on Build GRCh38
Transcripts
NM_001144888.1 NM_006340.2 NM_017450.2 NM_017451.2 XM_005256943.1 XM_005256944.1 XM_005256945.1 XM_005256948.3 XM_006721635.1 XM_006721636.1 XM_006721637.2 XM_011524193.1 XM_011524194.1 XM_011524195.1 XM_011524196.1 XM_011524197.1 XM_017024017.1 XM_017024018.1 XM_017024019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609779 c.719C>T p.A240V Chr.17: 81103578-81103578 on Build GRCh38
301743 c.710C>T p.P237L Chr.17: 81103569-81103569 on Build GRCh38
6609778 c.719C>T p.A240V Chr.17: 81103578-81103578 on Build GRCh38
4508826 c.788C>T p.S263F Chr.17: 81103647-81103647 on Build GRCh38
6609741 c.805G>A p.D269N Chr.17: 81103664-81103664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904013066 Chr.17: 81103790-81103790 on Build GRCh38
rs1008483164 Chr.17: 81103393-81103393 on Build GRCh38
rs745320058 Chr.17: 81103518-81103518 on Build GRCh38
rs767216738 Chr.17: 81103642-81103642 on Build GRCh38
rs916489305 Chr.17: 81103654-81103654 on Build GRCh38
rs763520836 Chr.17: 81103730-81103730 on Build GRCh38
rs146997101 Chr.17: 81103579-81103579 on Build GRCh38
rs780259021 Chr.17: 81103612-81103612 on Build GRCh38
rs772891818 Chr.17: 81103578-81103578 on Build GRCh38
rs538951036 Chr.17: 81103675-81103675 on Build GRCh38
rs1022352094 Chr.17: 81103603-81103603 on Build GRCh38
rs755745862 Chr.17: 81103657-81103657 on Build GRCh38
rs757987780 Chr.17: 81103519-81103519 on Build GRCh38
rs201786144 Chr.17: 81103577-81103577 on Build GRCh38
rs765037803 Chr.17: 81103516-81103516 on Build GRCh38
rs200210885 Chr.17: 81103717-81103717 on Build GRCh38
rs750986627 Chr.17: 81103674-81103674 on Build GRCh38
rs748362686 Chr.17: 81103690-81103690 on Build GRCh38
rs944146140 Chr.17: 81103786-81103786 on Build GRCh38
rs752268332 Chr.17: 81103656-81103656 on Build GRCh38
rs1031453012 Chr.17: 81103635-81103635 on Build GRCh38
rs779694736 Chr.17: 81103524-81103524 on Build GRCh38
rs200833670 Chr.17: 81103702-81103702 on Build GRCh38
rs992068315 Chr.17: 81103474-81103474 on Build GRCh38
rs778478562 Chr.17: 81103451-81103451 on Build GRCh38
rs760753131 Chr.17: 81103480-81103480 on Build GRCh38
rs369167632 Chr.17: 81103530-81103530 on Build GRCh38
rs1031065700 Chr.17: 81103795-81103795 on Build GRCh38
rs762599873 Chr.17: 81103785-81103786 on Build GRCh38
rs772199606 Chr.17: 81103553-81103553 on Build GRCh38
rs778303736 Chr.17: 81103384-81103384 on Build GRCh38
rs373270072 Chr.17: 81103636-81103636 on Build GRCh38
rs375070592 Chr.17: 81103575-81103575 on Build GRCh38
rs777695629 Chr.17: 81103699-81103699 on Build GRCh38
rs924372424 Chr.17: 81103487-81103487 on Build GRCh38
rs1022322355 Chr.17: 81103409-81103409 on Build GRCh38
rs554601074 Chr.17: 81103814-81103814 on Build GRCh38
rs371777908 Chr.17: 81103473-81103473 on Build GRCh38
rs371555896 Chr.17: 81103742-81103742 on Build GRCh38
rs760374560 Chr.17: 81103715-81103715 on Build GRCh38
rs1023575047 Chr.17: 81103830-81103830 on Build GRCh38
rs766248709 Chr.17: 81103630-81103630 on Build GRCh38
rs755203182 Chr.17: 81103685-81103685 on Build GRCh38
rs992100996 Chr.17: 81103640-81103640 on Build GRCh38
rs1021269875 Chr.17: 81103464-81103464 on Build GRCh38
rs746177192 Chr.17: 81103552-81103552 on Build GRCh38
rs746349769 Chr.17: 81103609-81103609 on Build GRCh38
rs751642315 Chr.17: 81103763-81103763 on Build GRCh38
rs753245697 Chr.17: 81103726-81103726 on Build GRCh38
rs143001141 Chr.17: 81103774-81103776 on Build GRCh38
rs773970956 Chr.17: 81103631-81103631 on Build GRCh38
rs767161879 Chr.17: 81103594-81103594 on Build GRCh38
rs146622763 Chr.17: 81103385-81103385 on Build GRCh38
rs751155844 Chr.17: 81103525-81103525 on Build GRCh38
rs975095013 Chr.17: 81103462-81103462 on Build GRCh38
rs747167307 Chr.17: 81103558-81103558 on Build GRCh38
rs758866130 Chr.17: 81103610-81103610 on Build GRCh38
rs771464994 Chr.17: 81103465-81103465 on Build GRCh38
rs1000056858 Chr.17: 81103794-81103794 on Build GRCh38
rs774203588 Chr.17: 81103709-81103709 on Build GRCh38
rs780498773 Chr.17: 81103678-81103678 on Build GRCh38
rs866376390 Chr.17: 81103551-81103551 on Build GRCh38
rs536348523 Chr.17: 81103787-81103787 on Build GRCh38
rs149587657 Chr.17: 81103706-81103706 on Build GRCh38
rs776695608 Chr.17: 81103619-81103619 on Build GRCh38
rs756243479 Chr.17: 81103748-81103748 on Build GRCh38
rs753812724 Chr.17: 81103492-81103492 on Build GRCh38
rs772298350 Chr.17: 81103476-81103476 on Build GRCh38
rs775437751 Chr.17: 81103477-81103477 on Build GRCh38
rs138233728 Chr.17: 81103663-81103663 on Build GRCh38
rs751267807 Chr.17: 81103589-81103589 on Build GRCh38
rs762765620 Chr.17: 81103628-81103628 on Build GRCh38
rs1043838592 Chr.17: 81103753-81103753 on Build GRCh38
rs767042934 Chr.17: 81103517-81103517 on Build GRCh38
rs781447173 Chr.17: 81103689-81103689 on Build GRCh38
rs752044874 Chr.17: 81103741-81103741 on Build GRCh38
rs75955278 Chr.17: 81103493-81103493 on Build GRCh38
rs754657609 Chr.17: 81103593-81103593 on Build GRCh38
rs961021218 Chr.17: 81103801-81103801 on Build GRCh38
rs920176083 Chr.17: 81103755-81103755 on Build GRCh38
rs752217288 Chr.17: 81103596-81103596 on Build GRCh38
rs775350782 Chr.17: 81103714-81103714 on Build GRCh38
rs749861902 Chr.17: 81103543-81103543 on Build GRCh38
rs768770966 Chr.17: 81103570-81103570 on Build GRCh38
rs978471733 Chr.17: 81103431-81103431 on Build GRCh38
rs759347952 Chr.17: 81103712-81103712 on Build GRCh38
rs774838970 Chr.17: 81103467-81103467 on Build GRCh38
rs751854175 Chr.17: 81103681-81103681 on Build GRCh38
rs764147330 Chr.17: 81103482-81103482 on Build GRCh38
rs1013565343 Chr.17: 81103811-81103811 on Build GRCh38
rs992770877 Chr.17: 81103511-81103511 on Build GRCh38
rs749264291 Chr.17: 81103757-81103757 on Build GRCh38
rs757794937 Chr.17: 81103546-81103546 on Build GRCh38
rs150398192 Chr.17: 81103627-81103627 on Build GRCh38
rs141395686 Chr.17: 81103613-81103613 on Build GRCh38
rs749811041 Chr.17: 81103459-81103459 on Build GRCh38
rs768826195 Chr.17: 81103617-81103617 on Build GRCh38
rs775840355 Chr.17: 81103555-81103555 on Build GRCh38
rs943691615 Chr.17: 81103718-81103718 on Build GRCh38
rs766736894 Chr.17: 81103734-81103734 on Build GRCh38
rs761769705 Chr.17: 81103576-81103576 on Build GRCh38
rs779050963 Chr.17: 81103764-81103764 on Build GRCh38
rs764496449 Chr.17: 81103784-81103784 on Build GRCh38
rs766415384 Chr.17: 81103754-81103755 on Build GRCh38
rs777942500 Chr.17: 81103752-81103752 on Build GRCh38
rs968087488 Chr.17: 81103411-81103411 on Build GRCh38
rs111840759 Chr.17: 81103436-81103436 on Build GRCh38
rs571846743 Chr.17: 81103786-81103787 on Build GRCh38
rs757844412 Chr.17: 81103601-81103601 on Build GRCh38
rs748164415 Chr.17: 81103622-81103622 on Build GRCh38
rs770918645 Chr.17: 81103761-81103761 on Build GRCh38
rs746634655 Chr.17: 81103792-81103792 on Build GRCh38
rs781502660 Chr.17: 81103746-81103746 on Build GRCh38
rs769846726 Chr.17: 81103624-81103624 on Build GRCh38
rs371666322 Chr.17: 81103531-81103531 on Build GRCh38
rs779452421 Chr.17: 81103549-81103549 on Build GRCh38
rs769953866 Chr.17: 81103693-81103693 on Build GRCh38

View Full Product Details