Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81120076-81120616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AATK
Entrez Gene ID
9625
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP
UniGene
Hs.514575
Gene Chromosome Location
Chr.17: 81117295-81166072 on Build GRCh38
Transcripts
NM_001080395.2 NM_004920.2 XM_006722193.2 XM_006722194.2 XM_006722195.2 XM_006722196.2 XM_011525509.2 XM_011525510.2 XM_011525511.2 XM_017025421.1 XM_017025422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7415114 c.3670G>T p.D1224Y Chr.17: 81120266-81120266 on Build GRCh38
1564139 c.3679C>T p.R1227C Chr.17: 81120257-81120257 on Build GRCh38
7408830 c.3364G>C p.G1122R Chr.17: 81120572-81120572 on Build GRCh38
4646569 c.3404G>C p.G1135A Chr.17: 81120532-81120532 on Build GRCh38
5363812 c.3404delG p.G1135fs*172 Chr.17: 81120532-81120532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370976937 Chr.17: 81120313-81120313 on Build GRCh38
rs751219940 Chr.17: 81120127-81120127 on Build GRCh38
rs962651428 Chr.17: 81120149-81120149 on Build GRCh38
rs111744243 Chr.17: 81120545-81120545 on Build GRCh38
rs767076655 Chr.17: 81120131-81120131 on Build GRCh38
rs760158729 Chr.17: 81120209-81120209 on Build GRCh38
rs746857718 Chr.17: 81120431-81120431 on Build GRCh38
rs56360567 Chr.17: 81120363-81120363 on Build GRCh38
rs773298565 Chr.17: 81120585-81120585 on Build GRCh38
rs776928858 Chr.17: 81120578-81120578 on Build GRCh38
rs767063580 Chr.17: 81120297-81120297 on Build GRCh38
rs577652593 Chr.17: 81120163-81120163 on Build GRCh38
rs778512295 Chr.17: 81120351-81120351 on Build GRCh38
rs143364400 Chr.17: 81120444-81120444 on Build GRCh38
rs1012231618 Chr.17: 81120135-81120135 on Build GRCh38
rs745760208 Chr.17: 81120349-81120349 on Build GRCh38
rs575537754 Chr.17: 81120234-81120234 on Build GRCh38
rs1035416601 Chr.17: 81120100-81120100 on Build GRCh38
rs55793641 Chr.17: 81120458-81120458 on Build GRCh38
rs761982339 Chr.17: 81120483-81120483 on Build GRCh38
rs750355431 Chr.17: 81120426-81120426 on Build GRCh38
rs748897440 Chr.17: 81120542-81120542 on Build GRCh38
rs1025846649 Chr.17: 81120580-81120580 on Build GRCh38
rs779354524 Chr.17: 81120162-81120162 on Build GRCh38
rs761084908 Chr.17: 81120474-81120474 on Build GRCh38
rs548877317 Chr.17: 81120204-81120204 on Build GRCh38
rs747448646 Chr.17: 81120557-81120557 on Build GRCh38
rs556627095 Chr.17: 81120579-81120579 on Build GRCh38
rs369187278 Chr.17: 81120556-81120556 on Build GRCh38
rs755368457 Chr.17: 81120231-81120231 on Build GRCh38
rs1018473008 Chr.17: 81120460-81120460 on Build GRCh38
rs111881294 Chr.17: 81120510-81120510 on Build GRCh38
rs769038810 Chr.17: 81120576-81120576 on Build GRCh38
rs772731013 Chr.17: 81120467-81120467 on Build GRCh38
rs202227008 Chr.17: 81120420-81120420 on Build GRCh38
rs766304985 Chr.17: 81120589-81120589 on Build GRCh38
rs747739874 Chr.17: 81120454-81120454 on Build GRCh38
rs768708455 Chr.17: 81120170-81120170 on Build GRCh38
rs765250764 Chr.17: 81120489-81120489 on Build GRCh38
rs372164584 Chr.17: 81120429-81120429 on Build GRCh38
rs751487460 Chr.17: 81120505-81120505 on Build GRCh38
rs538310773 Chr.17: 81120506-81120506 on Build GRCh38
rs748347875 Chr.17: 81120240-81120240 on Build GRCh38
rs903090298 Chr.17: 81120463-81120463 on Build GRCh38
rs778877532 Chr.17: 81120345-81120345 on Build GRCh38
rs776229948 Chr.17: 81120292-81120292 on Build GRCh38
rs1023793942 Chr.17: 81120370-81120370 on Build GRCh38
rs768783665 Chr.17: 81120106-81120106 on Build GRCh38
rs755316163 Chr.17: 81120312-81120312 on Build GRCh38
rs758714054 Chr.17: 81120311-81120319 on Build GRCh38
rs767796214 Chr.17: 81120383-81120383 on Build GRCh38
rs757323806 Chr.17: 81120341-81120341 on Build GRCh38
rs972549056 Chr.17: 81120152-81120152 on Build GRCh38
rs774728297 Chr.17: 81120369-81120369 on Build GRCh38
rs557107438 Chr.17: 81120183-81120183 on Build GRCh38
rs775203891 Chr.17: 81120208-81120208 on Build GRCh38
rs767115629 Chr.17: 81120198-81120198 on Build GRCh38
rs764382577 Chr.17: 81120479-81120479 on Build GRCh38
rs768561727 Chr.17: 81120440-81120440 on Build GRCh38
rs763724298 Chr.17: 81120288-81120288 on Build GRCh38
rs755618434 Chr.17: 81120154-81120154 on Build GRCh38
rs368792123 Chr.17: 81120326-81120326 on Build GRCh38
rs750813468 Chr.17: 81120272-81120274 on Build GRCh38
rs772677815 Chr.17: 81120471-81120471 on Build GRCh38
rs1056627786 Chr.17: 81120555-81120555 on Build GRCh38
rs758843423 Chr.17: 81120168-81120168 on Build GRCh38
rs770905516 Chr.17: 81120248-81120248 on Build GRCh38
rs773131429 Chr.17: 81120117-81120117 on Build GRCh38
rs761382681 Chr.17: 81120296-81120296 on Build GRCh38
rs760304700 Chr.17: 81120286-81120286 on Build GRCh38
rs770746038 Chr.17: 81120191-81120191 on Build GRCh38
rs375456982 Chr.17: 81120377-81120377 on Build GRCh38
rs1025394136 Chr.17: 81120212-81120212 on Build GRCh38
rs1041556592 Chr.17: 81120461-81120461 on Build GRCh38
rs766869636 Chr.17: 81120224-81120224 on Build GRCh38
rs192541131 Chr.17: 81120147-81120147 on Build GRCh38
rs374195702 Chr.17: 81120257-81120257 on Build GRCh38
rs755839232 Chr.17: 81120532-81120532 on Build GRCh38
rs562940571 Chr.17: 81120172-81120172 on Build GRCh38
rs184348609 Chr.17: 81120322-81120322 on Build GRCh38
rs539240235 Chr.17: 81120176-81120176 on Build GRCh38
rs749248581 Chr.17: 81120247-81120247 on Build GRCh38
rs752133891 Chr.17: 81120099-81120101 on Build GRCh38
rs745714755 Chr.17: 81120282-81120282 on Build GRCh38
rs763110575 Chr.17: 81120586-81120586 on Build GRCh38
rs61743713 Chr.17: 81120490-81120490 on Build GRCh38
rs377359465 Chr.17: 81120275-81120275 on Build GRCh38
rs1038339963 Chr.17: 81120415-81120415 on Build GRCh38
rs372185250 Chr.17: 81120364-81120364 on Build GRCh38
rs765533377 Chr.17: 81120584-81120584 on Build GRCh38
rs763638615 Chr.17: 81120210-81120210 on Build GRCh38
rs750722905 Chr.17: 81120504-81120504 on Build GRCh38
rs771887428 Chr.17: 81120283-81120283 on Build GRCh38
rs770816664 Chr.17: 81120225-81120225 on Build GRCh38
rs963174896 Chr.17: 81120375-81120375 on Build GRCh38
rs540245594 Chr.17: 81120331-81120331 on Build GRCh38
rs747276632 Chr.17: 81120445-81120447 on Build GRCh38
rs780479673 Chr.17: 81120104-81120104 on Build GRCh38
rs903120968 Chr.17: 81120358-81120358 on Build GRCh38
rs777262558 Chr.17: 81120541-81120541 on Build GRCh38
rs757851302 Chr.17: 81120246-81120248 on Build GRCh38
rs781393781 Chr.17: 81120444-81120445 on Build GRCh38
rs752095600 Chr.17: 81120306-81120306 on Build GRCh38
rs199725264 Chr.17: 81120384-81120384 on Build GRCh38
rs750951008 Chr.17: 81120164-81120164 on Build GRCh38
rs947313694 Chr.17: 81120327-81120327 on Build GRCh38
rs564244999 Chr.17: 81120416-81120416 on Build GRCh38
rs758929898 Chr.17: 81120219-81120219 on Build GRCh38
rs776528669 Chr.17: 81120107-81120107 on Build GRCh38
rs200255480 Chr.17: 81120243-81120243 on Build GRCh38
rs774083842 Chr.17: 81120193-81120193 on Build GRCh38
rs572926712 Chr.17: 81120323-81120323 on Build GRCh38
rs759149213 Chr.17: 81120129-81120129 on Build GRCh38
rs759354596 Chr.17: 81120194-81120194 on Build GRCh38
rs370038265 Chr.17: 81120480-81120480 on Build GRCh38
rs779223813 Chr.17: 81120245-81120246 on Build GRCh38
rs775858843 Chr.17: 81120473-81120473 on Build GRCh38
rs752197081 Chr.17: 81120133-81120133 on Build GRCh38
rs756153426 Chr.17: 81120242-81120242 on Build GRCh38
rs368515584 Chr.17: 81120382-81120382 on Build GRCh38
rs768583972 Chr.17: 81120441-81120441 on Build GRCh38
rs760998878 Chr.17: 81120400-81120400 on Build GRCh38
rs747261863 Chr.17: 81120169-81120169 on Build GRCh38
rs370847936 Chr.17: 81120218-81120218 on Build GRCh38
rs371044067 Chr.17: 81120447-81120447 on Build GRCh38
rs895167121 Chr.17: 81120134-81120134 on Build GRCh38
rs989822492 Chr.17: 81120291-81120291 on Build GRCh38
rs376545697 Chr.17: 81120113-81120113 on Build GRCh38
rs1026276034 Chr.17: 81120419-81120419 on Build GRCh38
rs747167873 Chr.17: 81120105-81120105 on Build GRCh38
rs768846587 Chr.17: 81120445-81120450 on Build GRCh38
rs113944067 Chr.17: 81120546-81120546 on Build GRCh38
rs746658657 Chr.17: 81120360-81120360 on Build GRCh38
rs761829409 Chr.17: 81120114-81120114 on Build GRCh38
rs780289093 Chr.17: 81120337-81120337 on Build GRCh38
rs758302958 Chr.17: 81120428-81120428 on Build GRCh38
rs55856613 Chr.17: 81120362-81120362 on Build GRCh38
rs758376255 Chr.17: 81120491-81120491 on Build GRCh38
rs775018366 Chr.17: 81120285-81120285 on Build GRCh38
rs747825658 Chr.17: 81120525-81120525 on Build GRCh38
rs776075353 Chr.17: 81120552-81120552 on Build GRCh38
rs530717750 Chr.17: 81120424-81120424 on Build GRCh38
rs769906823 Chr.17: 81120179-81120179 on Build GRCh38
rs916660983 Chr.17: 81120577-81120577 on Build GRCh38
rs749158694 Chr.17: 81120187-81120187 on Build GRCh38
rs200251873 Chr.17: 81120494-81120494 on Build GRCh38
rs745803589 Chr.17: 81120356-81120356 on Build GRCh38
rs781098815 Chr.17: 81120523-81120523 on Build GRCh38
rs769463517 Chr.17: 81120466-81120466 on Build GRCh38

View Full Product Details